Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01007109.hg.1 |
ENST00000373210.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01007109.hg.1 |
NM_017629.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01007109.hg.1 |
TC01000462.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007784.hg.1 |
ENST00000496257.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007784.hg.1 |
ENST00000496257.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007784.hg.1 |
ENST00000496257.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007784.hg.1 |
NM_017629.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007784.hg.1 |
NM_017629.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007784.hg.1 |
NM_017629.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007784.hg.1 |
NM_017629.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007784.hg.1 |
NM_017629.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007784.hg.1 |
NM_017629.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007784.hg.1 |
NM_017629.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007784.hg.1 |
NM_017629.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007784.hg.1 |
NM_017629.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007784.hg.1 |
NM_017629.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007784.hg.1 |
NM_017629.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007784.hg.1 |
NM_017629.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007784.hg.1 |
NM_017629.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007784.hg.1 |
NM_017629.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007784.hg.1 |
NM_017629.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007784.hg.1 |
NM_017629.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007784.hg.1 |
NM_017629.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007784.hg.1 |
NM_017629.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007784.hg.1 |
NM_017629.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007784.hg.1 |
NM_017629.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007784.hg.1 |
NM_017629.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007784.hg.1 |
NM_017629.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007784.hg.1 |
NM_017629.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007784.hg.1 |
NM_017629.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007784.hg.1 |
NM_017629.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007784.hg.1 |
NM_017629.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007784.hg.1 |
ENST00000373210.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007784.hg.1 |
ENST00000373210.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007784.hg.1 |
ENST00000373210.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007784.hg.1 |
ENST00000373210.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007784.hg.1 |
ENST00000373210.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007784.hg.1 |
ENST00000373210.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007784.hg.1 |
ENST00000373210.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007784.hg.1 |
ENST00000373210.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007784.hg.1 |
ENST00000373210.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007784.hg.1 |
ENST00000373210.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007784.hg.1 |
ENST00000373210.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007784.hg.1 |
ENST00000373210.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007784.hg.1 |
ENST00000373210.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007784.hg.1 |
ENST00000373210.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007784.hg.1 |
ENST00000373210.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007784.hg.1 |
ENST00000373210.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007784.hg.1 |
ENST00000373210.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007784.hg.1 |
ENST00000373210.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007784.hg.1 |
ENST00000373210.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007784.hg.1 |
ENST00000373210.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007784.hg.1 |
ENST00000373210.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007784.hg.1 |
ENST00000373210.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007784.hg.1 |
ENST00000373210.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007784.hg.1 |
ENST00000373210.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007784.hg.1 |
ENST00000373210.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007784.hg.1 |
ENST00000373210.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007784.hg.1 |
ENST00000373210.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007784.hg.1 |
ENST00000488778.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007784.hg.1 |
ENST00000488778.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007784.hg.1 |
ENST00000488778.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007784.hg.1 |
ENST00000488778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100154774.hg.1 |
ENST00000373210.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100154774.hg.1 |
NM_017629.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100154774.hg.1 |
TC0100007784.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100154774.hg.1 |
ENST00000373210.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100154774.hg.1 |
NM_017629.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007784.hg.1 |
ENST00000496257.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007784.hg.1 |
ENST00000496257.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007784.hg.1 |
ENST00000496257.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007784.hg.1 |
NM_017629.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007784.hg.1 |
NM_017629.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007784.hg.1 |
NM_017629.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007784.hg.1 |
NM_017629.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007784.hg.1 |
NM_017629.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007784.hg.1 |
NM_017629.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007784.hg.1 |
NM_017629.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007784.hg.1 |
NM_017629.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007784.hg.1 |
NM_017629.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007784.hg.1 |
NM_017629.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007784.hg.1 |
NM_017629.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007784.hg.1 |
NM_017629.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007784.hg.1 |
NM_017629.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007784.hg.1 |
NM_017629.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007784.hg.1 |
NM_017629.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007784.hg.1 |
NM_017629.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007784.hg.1 |
NM_017629.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007784.hg.1 |
NM_017629.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007784.hg.1 |
NM_017629.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007784.hg.1 |
NM_017629.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007784.hg.1 |
NM_017629.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007784.hg.1 |
NM_017629.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007784.hg.1 |
NM_017629.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007784.hg.1 |
NM_017629.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007784.hg.1 |
NM_017629.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007784.hg.1 |
NM_017629.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007784.hg.1 |
ENST00000373210.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007784.hg.1 |
ENST00000373210.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007784.hg.1 |
ENST00000373210.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007784.hg.1 |
ENST00000373210.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007784.hg.1 |
ENST00000373210.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007784.hg.1 |
ENST00000373210.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007784.hg.1 |
ENST00000373210.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007784.hg.1 |
ENST00000373210.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007784.hg.1 |
ENST00000373210.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007784.hg.1 |
ENST00000373210.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007784.hg.1 |
ENST00000373210.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007784.hg.1 |
ENST00000373210.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007784.hg.1 |
ENST00000373210.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007784.hg.1 |
ENST00000373210.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007784.hg.1 |
ENST00000373210.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007784.hg.1 |
ENST00000373210.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007784.hg.1 |
ENST00000373210.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007784.hg.1 |
ENST00000373210.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007784.hg.1 |
ENST00000373210.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007784.hg.1 |
ENST00000373210.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007784.hg.1 |
ENST00000373210.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007784.hg.1 |
ENST00000373210.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007784.hg.1 |
ENST00000373210.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007784.hg.1 |
ENST00000373210.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007784.hg.1 |
ENST00000373210.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007784.hg.1 |
ENST00000373210.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007784.hg.1 |
ENST00000373210.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007784.hg.1 |
ENST00000488778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007784.hg.1 |
ENST00000488778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007784.hg.1 |
ENST00000488778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007784.hg.1 |
ENST00000488778.1 |
Ensembl |