Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01061729_m1
          See other NR2F1-AS1 GE Assays ›
          Gene Symbol:
          NR2F1-AS1 Hs.457407-USP11 fusion
          Gene Name:
          NR2F1 antisense RNA 1
          FUSION
          Gene Aliases:
          -
          Hs.457407, USP11
          Chromosome Location:
          Chr.5: 93409356 - 93581320 on Build GRCh38
          Chromosome Location:
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Hs01061729_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: NR2F1-AS1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_021490.2 2 - 3 278 70
          NR_021491.2 2 - 3 278 70
          NR_109818.1 3 - 4 391 70
          NR_109819.1 2 - 3 953 70
          NR_109820.1 2 - 3 278 70
          NR_109821.1 2 - 3 278 70
          NR_109823.1 2 - 3 275 70
          NR_109824.1 2 - 3 278 70
          NR_109825.1 3 - 4 1058 70
          GenBank mRNA AK095719.1 - 3 - 4 369 70
          AK124699.1 - 2 - 3 1003 70
          AK311143.1 - 2 - 3 177 70
          AK311323.1 - 2 - 3 666 70
          BC010610.1 - 2 - 3 246 70
          BC018089.1 - 2 - 3 255 70
          BC042879.1 - 2 - 3 177 70
          BC044619.1 - 3 - 4 534 70
          BC048999.1 - 3 - 4 534 70
          BC144330.1 - 2 - 3 1553 70
          BC144331.1 - 2 - 3 1553 70
          BX647510.1 - 1 - 2 46 70
          DA718791.1 - 2 - 3 261 70
          DB257894.1 - 2 - 3 177 70
          HY108556.1 - 2 - 3 208 70
          Gene Symbol: Hs.457407-USP11 fusion
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK092258.1 - 688 70
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA DB503335.1 - 2 - 3 174 Chr. 5 - 93544834 - 93581216 [-] on Build GRCh38 70
          GenBankmRNA DA531679.1 - 2 - 3 329 Chr. 5 - 93543968 - 93581370 [-] on Build GRCh38 70
          GenBankmRNA BI463026.1 - 3 - 4 541 Chr. 5 - 93554872 - 93570826 [-] on Build GRCh38 70
          GenBankmRNA DA530156.1 - 2 - 3 527 Chr. 5 - 93554928 - 93580891 [-] on Build GRCh38 70
          GenBankmRNA DB045052.1 - 3 - 4 468 Chr. 5 - 93554901 - 93570793 [-] on Build GRCh38 70
          GenBankmRNA BM546601.1 - 343 Chr. - - [-] on Build 70
          GenBankmRNA DA271332.1 - 2 - 3 192 Chr. 5 - 93438714 - 93581234 [-] on Build GRCh38 70
          GenBankmRNA HY136165.1 - 3 - 4 235 Chr. 5 - 93553771 - 93581276 [-] on Build GRCh38 70
          GenBankmRNA DB486010.2 - 2 - 3 174 Chr. 5 - 93411386 - 93581216 [-] on Build GRCh38 70
          GenBankmRNA DB173308.1 - 2 - 3 428 Chr. 5 - 93553837 - 93580795 [-] on Build GRCh38 70
          GenBankmRNA DA533837.1 - 2 - 3 251 Chr. 5 - 93544815 - 93580618 [-] on Build GRCh38 70
          GenBankmRNA DA363045.1 - 2 - 3 322 Chr. 5 - 93553758 - 93580689 [-] on Build GRCh38 70
          GenBankmRNA DA134670.1 - 2 - 3 177 Chr. 5 - 93411404 - 93581219 [-] on Build GRCh38 70
          GenBankmRNA DA527197.1 - 2 - 3 261 Chr. 5 - 93411404 - 93581303 [-] on Build GRCh38 70
          GenBankmRNA DA706940.1 - 2 - 3 260 Chr. 5 - 93544828 - 93581300 [-] on Build GRCh38 70
          GenBankmRNA BX377086.2 - 2 - 3 181 Chr. 5 - 93543540 - 93581223 [-] on Build GRCh38 70
          GenBankmRNA BG701915.1 - 2 - 3 422 Chr. 5 - 93501042 - 93585650 [-] on Build GRCh38 70
          GenBankmRNA DA844094.1 - 2 - 3 257 Chr. 5 - 93544836 - 93581297 [-] on Build GRCh38 70
          GenBankmRNA AA375334.1 - 2 - 3 126 Chr. 5 - 93553829 - 93581168 [-] on Build GRCh38 70
          GenBankmRNA BI669922.1 - 2 - 3 182 Chr. 5 - 93411232 - 93581219 [-] on Build GRCh38 70
          GenBankmRNA DA518837.1 - 3 - 4 369 Chr. 5 - 93553785 - 93581297 [-] on Build GRCh38 70
          GenBankmRNA DB272535.1 - 2 - 3 279 Chr. 5 - 93544853 - 93580646 [-] on Build GRCh38 70
          GenBankmRNA DA545967.1 - 2 - 3 177 Chr. 5 - 93544816 - 93581219 [-] on Build GRCh38 70
          GenBankmRNA DA304607.1 - 2 - 3 164 Chr. 5 - 93411321 - 93581205 [-] on Build GRCh38 70
          GenBankmRNA DB498974.1 - 2 - 3 174 Chr. 5 - 93544802 - 93581216 [-] on Build GRCh38 70
          GenBankmRNA DA839566.1 - 1 - 2 68 Chr. 5 - 93432362 - 93563432 [-] on Build GRCh38 70
          GenBankmRNA HY141502.1 - 2 - 3 158 Chr. 5 - 93544815 - 93581199 [-] on Build GRCh38 70
          GenBankmRNA BG707159.1 - 2 - 3 182 Chr. 5 - 93411365 - 93581219 [-] on Build GRCh38 70
          GenBankmRNA DA388648.1 - 2 - 3 328 Chr. 5 - 93544889 - 93580695 [-] on Build GRCh38 70
          GenBankmRNA DA328801.1 - 2 - 3 179 Chr. 5 - 93543992 - 93581221 [-] on Build GRCh38 70
          GenBankmRNA DN993098.1 - 2 - 3 379 Chr. 5 - 93553814 - 93580737 [-] on Build GRCh38 70
          GenBankmRNA BI820847.1 - 2 - 3 228 Chr. 5 - 93342240 - 93580595 [-] on Build GRCh38 70
          GenBankmRNA DA517345.1 - 2 - 3 254 Chr. 5 - 93411434 - 93581295 [-] on Build GRCh38 70
          GenBankmRNA DB492381.1 - 2 - 3 174 Chr. 5 - 93544849 - 93581216 [-] on Build GRCh38 70
          GenBankmRNA DB475119.2 - 2 - 3 252 Chr. 5 - 93411324 - 93581297 [-] on Build GRCh38 70
          GenBankmRNA BX444443.2 - 2 - 3 817 Chr. 5 - 93553821 - 93581185 [-] on Build GRCh38 70
          GenBankmRNA DB177914.1 - 2 - 3 258 Chr. 5 - 93438700 - 93581300 [-] on Build GRCh38 70
          GenBankmRNA DB030965.1 - 3 - 4 437 Chr. 5 - 93554880 - 93570722 [-] on Build GRCh38 70

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream ARRDC3 antisense RNA 1 ARRDC3-AS1 -2.1mb Anti-sense Chr. 5 - 91380347 - 91420715 on NCBI Build GRCh38
          Downstream nuclear receptor subfamily 2 group F member 1 NR2F1 BBOAS,
          BBSOAS,
          COUP-TFI,
          EAR-3,
          EAR3,
          ERBAL3,
          NR2F2,
          SVP44,
          TCFCOUP1,
          TFCOUP1
          19.9kb Anti-sense Chr. 5 - 93583337 - 93594613 on NCBI Build GRCh38

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0