Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01063130_g1
          See other RAPGEF1 GE Assays ›
          Gene Symbol:
          RAPGEF1
          Gene Name:
          Rap guanine nucleotide exchange factor 1
          Gene Aliases:
          C3G, GRF2
          Chromosome Location:
          Chr.9: 131576770 - 131740068 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01063130_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RAPGEF1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001304275.1 NP_001291204.1 20 - 21 2923 75
          NM_005312.3 NP_005303.2 20 - 21 2882 75
          NM_198679.1 NP_941372.1 20 - 21 2971 75
          XM_005272186.4 XP_005272243.1 23 - 24 3557 75
          XM_005272191.3 XP_005272248.1 22 - 23 3400 75
          XM_006717067.3 XP_006717130.1 23 - 24 3519 75
          XM_006717072.3 XP_006717135.1 22 - 23 3415 75
          XM_006717074.3 XP_006717137.1 20 - 21 3009 75
          XM_011518569.2 XP_011516871.1 23 - 24 3644 75
          XM_011518570.2 XP_011516872.1 23 - 24 3533 75
          XM_011518571.2 XP_011516873.1 23 - 24 4204 75
          XM_011518572.2 XP_011516874.1 23 - 24 3383 75
          XM_011518573.2 XP_011516875.1 23 - 24 3516 75
          XM_011518574.2 XP_011516876.1 22 - 23 3443 75
          XM_011518575.2 XP_011516877.1 22 - 23 3440 75
          XM_011518576.2 XP_011516878.1 21 - 22 3185 75
          XM_011518577.2 XP_011516879.1 21 - 22 3182 75
          XM_011518578.2 XP_011516880.1 21 - 22 3119 75
          XM_011518579.2 XP_011516881.1 20 - 21 3026 75
          XM_017014633.1 XP_016870122.1 23 - 24 3529 75
          XM_017014634.1 XP_016870123.1 23 - 24 3380 75
          XM_017014635.1 XP_016870124.1 23 - 24 3434 75
          XM_017014636.1 XP_016870125.1 23 - 24 3405 75
          XM_017014637.1 XP_016870126.1 22 - 23 3377 75
          XM_017014638.1 XP_016870127.1 21 - 22 3284 75
          XM_017014639.1 XP_016870128.1 22 - 23 3278 75
          GenBank mRNA AK302763.1 - 12 - 13 1461 75
          BC041710.1 - 20 - 21 2850 6051007 75
          CR749354.1 - 20 - 21 2824 75
          D21239.1 - 20 - 21 2888 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0900011778.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC09001668.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene RAPGEF1

          Gene Ontology Categories:

          Function(s) Process(es)

          blood vessel development
          signal transduction
          transmembrane receptor protein tyrosine kinase signaling pathway
          positive regulation of neuron projection development
          Rap protein signal transduction
          nerve growth factor signaling pathway
          positive regulation of GTPase activity
          negative regulation of Ras protein signal transduction
          platelet-derived growth factor receptor signaling pathway
          negative regulation of protein kinase B signaling
          establishment of endothelial barrier
          negative regulation of ERK1 and ERK2 cascade
          cellular response to cAMP
          negative regulation of canonical Wnt signaling pathway
          cell-cell adhesion
          regulation of cell junction assembly
          cellular response to nerve growth factor stimulus
          negative regulation of neural precursor cell proliferation
          protein binding
          SH3 domain binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR09021228.hg.1 ENST00000266110.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09021228.hg.1 ENST00000337036.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09021228.hg.1 ENST00000357686.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09021228.hg.1 ENST00000372189.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09021228.hg.1 ENST00000372190.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09021228.hg.1 ENST00000372191.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09021228.hg.1 ENST00000372195.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09021228.hg.1 ENST00000398415.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09021228.hg.1 ENST00000411834.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09021228.hg.1 ENST00000431470.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09021228.hg.1 ENST00000437236.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09021228.hg.1 NM_005312.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09021228.hg.1 NM_198679.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09021228.hg.1 TC09001668.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0900180471.hg.1 ENST00000372189.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0900180471.hg.1 ENST00000372190.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0900180471.hg.1 ENST00000372195.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0900180471.hg.1 NM_001304275.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900180471.hg.1 NM_005312.3 Refseq
          Affymetrix Human Clariom D Assay PSR0900180471.hg.1 NM_198679.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900180471.hg.1 TC0900011778.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900180471.hg.1 ENST00000372195.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0900180471.hg.1 ENST00000372189.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0900180471.hg.1 ENST00000372190.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0900180471.hg.1 NM_001304275.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900180471.hg.1 NM_005312.3 Refseq
          Affymetrix Human Clariom D Assay PSR0900180471.hg.1 NM_198679.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline