Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01063853_m1
          See other IKBKE GE Assays ›
          Gene Symbol:
          IKBKE
          Gene Name:
          inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase epsilon
          Gene Aliases:
          IKK-E, IKK-i, IKKE, IKKI
          Chromosome Location:
          Chr.1: 206470243 - 206496890 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01063853_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: IKBKE
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001193321.1 NP_001180250.1 16 - 17 2191 67
          NM_001193322.1 NP_001180251.1 17 - 18 2310 67
          NM_014002.3 NP_054721.1 17 - 18 2310 67
          XM_005273356.2 XP_005273413.1 17 - 18 2306 67
          XM_017002868.1 XP_016858357.1 16 - 17 2306 67
          GenBank mRNA AB016590.1 - 15 - 16 1734 67
          AF241789.1 - 15 - 16 1732 67
          AK093798.1 - 11 - 12 1171 67
          BC105923.1 - 17 - 18 2058 40001562 67
          BC105924.1 - 17 - 18 2058 40001567 67
          BC107591.1 - 17 - 18 2058 67
          BC107812.1 - 16 - 17 1939 40001564 67
          D63485.1 - 17 - 18 2058 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100011374.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01001723.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene IKBKE

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          immune response
          I-kappaB phosphorylation
          intrinsic apoptotic signaling pathway in response to DNA damage
          negative regulation of type I interferon production
          response to type I interferon
          response to interferon-beta
          TRIF-dependent toll-like receptor signaling pathway
          NIK/NF-kappaB signaling
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          protein homooligomerization
          cellular response to virus
          protein kinase activity
          NF-kappaB-inducing kinase activity
          protein binding
          ATP binding
          IkappaB kinase activity
          ubiquitin protein ligase binding
          K48-linked polyubiquitin modification-dependent protein binding
          K63-linked polyubiquitin modification-dependent protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR01026886.hg.1 ENST00000367120.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01026886.hg.1 ENST00000462698.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01026886.hg.1 ENST00000537984.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01026886.hg.1 NM_001193321.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01026886.hg.1 NM_001193322.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01026886.hg.1 NM_014002.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01026886.hg.1 TC01001723.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0100183623.hg.1 ENST00000578328.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0100183623.hg.1 ENST00000581977.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0100183623.hg.1 ENST00000584998.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100183623.hg.1 NM_001193321.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100183623.hg.1 NM_001193322.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100183623.hg.1 NM_014002.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100183623.hg.1 TC0100011374.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100183623.hg.1 ENST00000584998.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100183623.hg.1 ENST00000578328.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0100183623.hg.1 ENST00000581977.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0100183623.hg.1 NM_001193321.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100183623.hg.1 NM_001193322.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100183623.hg.1 NM_014002.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0