Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01065413_m1
          See other PARP8 GE Assays ›
          Gene Symbol:
          PARP8
          Gene Name:
          poly(ADP-ribose) polymerase family member 8
          Gene Aliases:
          ARTD16, pART16
          Chromosome Location:
          Chr.5: 50665899 - 50846522 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01065413_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PARP8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001178055.1 NP_001171526.1 23 - 24 2415 96
          NM_001178056.1 NP_001171527.1 21 - 22 2265 96
          NM_024615.3 NP_078891.2 22 - 23 2391 96
          XM_005248596.4 XP_005248653.1 23 - 24 2771 96
          XM_011543631.2 XP_011541933.1 23 - 24 2304 96
          XM_011543632.2 XP_011541934.2 22 - 23 2225 96
          XM_011543633.2 XP_011541935.2 23 - 24 2351 96
          XM_011543634.2 XP_011541936.1 24 - 25 2900 96
          XM_011543635.2 XP_011541937.1 24 - 25 2421 96
          XM_011543636.2 XP_011541938.1 23 - 24 2285 96
          XM_011543640.2 XP_011541942.1 20 - 21 2175 96
          XM_011543641.2 XP_011541943.1 21 - 22 2152 96
          XM_011543642.2 XP_011541944.1 19 - 20 2139 96
          XM_011543643.2 XP_011541945.1 20 - 21 2580 96
          XM_017009850.1 XP_016865339.1 22 - 23 2851 96
          XM_017009851.1 XP_016865340.1 21 - 22 2291 96
          GenBank mRNA AK024961.1 - 12 - 13 1461 96
          AK225188.1 - 12 - 13 1461 96
          AK289369.1 - 22 - 23 2374 96
          AK298680.1 - 21 - 22 2254 96
          AK314466.1 - 23 - 24 2415 96
          AL834477.1 - 22 - 23 2350 96
          BC037386.1 - 19 - 20 2123 4830820 96
          BC075801.1 - 21 - 22 2224 5226828 96

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0500007337.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC05000209.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene PARP8

          Gene Ontology Categories:

          Process(es)

          NAD+ ADP-ribosyltransferase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR05002551.hg.1 ENST00000281631.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05002551.hg.1 ENST00000423185.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05002551.hg.1 ENST00000503561.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05002551.hg.1 ENST00000503750.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05002551.hg.1 ENST00000505554.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05002551.hg.1 ENST00000505697.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05002551.hg.1 ENST00000511363.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05002551.hg.1 ENST00000514067.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05002551.hg.1 ENST00000514342.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05002551.hg.1 NM_001178055.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05002551.hg.1 NM_001178056.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05002551.hg.1 NM_024615.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05002551.hg.1 TC05000209.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0500149594.hg.1 ENST00000281631.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0500149594.hg.1 ENST00000503561.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0500149594.hg.1 ENST00000503750.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0500149594.hg.1 ENST00000505554.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500149594.hg.1 ENST00000505697.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0500149594.hg.1 ENST00000511363.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0500149594.hg.1 ENST00000514067.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0500149594.hg.1 ENST00000514342.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0500149594.hg.1 ENST00000515166.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500149594.hg.1 NM_001178055.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500149594.hg.1 NM_001178056.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500149594.hg.1 NM_024615.3 Refseq
          Affymetrix Human Clariom D Assay PSR0500149594.hg.1 TC0500007337.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0500149594.hg.1 ENST00000505554.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500149594.hg.1 ENST00000281631.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0500149594.hg.1 ENST00000503561.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0500149594.hg.1 ENST00000503750.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0500149594.hg.1 ENST00000505697.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0500149594.hg.1 ENST00000511363.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0500149594.hg.1 ENST00000514067.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0500149594.hg.1 ENST00000514342.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0500149594.hg.1 ENST00000515166.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500149594.hg.1 NM_001178055.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500149594.hg.1 NM_001178056.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500149594.hg.1 NM_024615.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6c59565f7c-q2rrq:80/100.66.130.62:80.
          git-commit: 653a2953ec6bb2e3dd1e37289b36fc4e0ab15bfe
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline