Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01065687_s1
          See other RAPGEF4-AS1 GE Assays ›
          Gene Symbol:
          RAPGEF4-AS1
          Gene Name:
          RAPGEF4 antisense RNA 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 172723189 - 172736206 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01065687_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RAPGEF4-AS1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_026995.1 4 556 100
          GenBank mRNA AL157450.1 - 4 556 100

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream pyruvate dehydrogenase kinase 1 PDK1 -50.5kb Anti-sense Chr. 2 - 172555373 - 172672693 on NCBI Build GRCh38
          Downstream Rap guanine nucleotide exchange factor 4 RAPGEF4 CAMP-GEFII,
          CGEF2,
          EPAC,
          EPAC 2,
          EPAC2,
          Nbla00496
          7.8kb Anti-sense Chr. 2 - 172735282 - 173052893 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21802312 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01678249 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21802328 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21802314 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21802313 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01678232 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01678247 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21802303 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21802304 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21802327 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21802305 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21802315 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01678259 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21802324 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21802300 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01678252 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21802325 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21802329 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21802302 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21802334 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01678230 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21802330 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21802317 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01678239 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21802308 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 27669 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 494391 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 433694 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 832184 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 605024 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 11797 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3390367 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_4913 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_4912 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 53582 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P00529925 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3390367 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3390367 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21802402 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01678355 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21802428 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01678350 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21802419 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01678332 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01678328 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01678362 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21802415 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21802404 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21802438 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21802406 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01678363 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01678356 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21802408 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21802420 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01678344 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01678361 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21802448 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21802401 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21802432 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21802427 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01678367 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21802439 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01678366 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21802423 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21802397 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15356143 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21802421 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21802436 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21802400 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21802413 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01678329 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01678314 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01678344 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21802404 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01678350 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21802408 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21802432 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01678355 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01678356 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01678328 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21802421 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21802448 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01678362 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21802397 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21802415 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21802420 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21802402 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21802400 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21802401 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21802406 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01678367 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21802413 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21802419 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01678361 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21802428 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15356143 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21802438 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21802423 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21802427 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21802439 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01678314 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01678329 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21802436 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01678366 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01678332 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01678363 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P00529925 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P15930305 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P00529925 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P15930327 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P15930311 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P15930318 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01678336 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01678339 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01678237 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01678249 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01678239 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01678369 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01678346 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01678340 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15356121 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01678323 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01678318 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01678366 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01678332 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01678247 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01678234 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01678343 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01678329 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01678356 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01678334 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01678353 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01678253 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01678345 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01678320 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01678257 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01678314 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01678241 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01678363 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01678250 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01678351 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01678325 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01678341 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01678372 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01678355 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01678358 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01678229 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01678327 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01678260 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01678361 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15356119 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01678255 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01678348 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15356143 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01678315 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P00529925 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P15930311 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 1563528_PM_at - -
          Human Genome U133 Plus 2.0 Array 1563528_at - -
          Human Genome U133 Plus 2.0 Array 1563528_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563528_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563528_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563528_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563528_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563528_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563528_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563528_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563528_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563528_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563528_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563528_at2 - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A014629 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A014629 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 229841 - -
          ABI Human Genome Survey Microarray Version 2.0 231165 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0