Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01065756_s1
          See other LINC00974 GE Assays ›
          Gene Symbol:
          LINC00974
          Gene Name:
          long intergenic non-protein coding RNA 974
          Gene Aliases:
          -
          Chromosome Location:
          Chr.17: 41549606 - 41554495 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01065756_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LINC00974
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_038442.1 3 1405 67
          GenBank mRNA AK056318.1 - 3 1405 67

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream keratin 19 KRT19 CK19,
          K19,
          K1CS
          -21.2kb Sense Chr. 17 - 41523617 - 41528389 on NCBI Build GRCh38
          Downstream keratin 9 KRT9 CK-9,
          EPPK,
          K9
          15.0kb Sense Chr. 17 - 41565841 - 41572058 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1700010678.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC17001511.hg.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10323265 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10323287 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10323302 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10323249 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30895362 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10323283 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10323316 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10323300 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10323258 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16982233 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10323268 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10323314 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30895322 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10323255 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30895349 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10323267 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10323269 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10323246 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10323280 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10323279 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30895317 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10323298 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10323250 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10323273 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30895368 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30895372 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10323270 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10323312 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10323311 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30895383 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10323264 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30895326 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10323307 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10323289 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10323266 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10323247 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30895332 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30895324 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10323292 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10323260 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10323281 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10323294 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10323295 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10323304 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10323277 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30895327 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30895370 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30895392 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10323272 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30895386 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10323296 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30895357 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10323244 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10323303 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10323306 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30895329 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10323313 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10323285 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10323271 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10323284 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30895391 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10323276 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30895352 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3738849 - -
          Agilent Human Genome CGH Microarray 44B A_14_P109739 - -
          Agilent Human Genome CGH Microarray 44A A_14_P109739 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 49906 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3738849 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3738849 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_14_P109739 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_14_P109739 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 117254 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 98954 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P40817986 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03245372 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_14_P109739 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03245363 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR17020012.hg.1 ENST00000456403.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17020012.hg.1 NR_038442.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17020012.hg.1 TC17001511.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR17020013.hg.1 ENST00000456403.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17020013.hg.1 NR_038442.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17020013.hg.1 TC17001511.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR17020011.hg.1 ENST00000456403.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17020011.hg.1 NR_038442.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17020011.hg.1 TC17001511.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1700010678.hg.1 ENST00000456403.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010678.hg.1 ENST00000456403.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010678.hg.1 ENST00000456403.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010678.hg.1 NR_038442.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010678.hg.1 NR_038442.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010678.hg.1 NR_038442.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700184466.hg.1 ENST00000456403.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700184466.hg.1 NR_038442.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700184466.hg.1 TC1700010678.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700184466.hg.1 ENST00000456403.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700184466.hg.1 NR_038442.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700184465.hg.1 ENST00000456403.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700184465.hg.1 NR_038442.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700184465.hg.1 TC1700010678.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700184465.hg.1 ENST00000456403.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700184465.hg.1 NR_038442.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700184467.hg.1 ENST00000456403.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700184467.hg.1 NR_038442.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700184467.hg.1 TC1700010678.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700184467.hg.1 ENST00000456403.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700184467.hg.1 NR_038442.1 Refseq
          Affymetrix Human Clariom D Assay TC1700010678.hg.1 ENST00000456403.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700010678.hg.1 ENST00000456403.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700010678.hg.1 ENST00000456403.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700010678.hg.1 NR_038442.1 Refseq
          Affymetrix Human Clariom D Assay TC1700010678.hg.1 NR_038442.1 Refseq
          Affymetrix Human Clariom D Assay TC1700010678.hg.1 NR_038442.1 Refseq
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A008339 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A008339 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 275496 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-786bfc498d-47c8f:80/100.66.130.62:80.
          git-commit: 9eb01563f6748445c58f67612c67ecf6b1da7421
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0