Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01066233_m1
          See other PTK2 GE Assays ›
          Gene Symbol:
          PTK2
          Gene Name:
          protein tyrosine kinase 2
          Gene Aliases:
          FADK, FAK, FAK1, FRNK, PPP1R71, p125FAK, pp125FAK
          Chromosome Location:
          Chr.8: 140658382 - 141001313 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01066233_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PTK2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001316342.1 NP_001303271.1 3 - 4 505 154
          XM_005251013.2 XP_005251070.1 3 - 4 463 154
          XM_006716609.2 XP_006716672.1 4 - 5 566 154
          XM_006716610.2 XP_006716673.1 4 - 5 558 154
          XM_017013681.1 XP_016869170.1 4 - 5 566 154
          XM_017013682.1 XP_016869171.1 3 - 4 463 154
          XM_017013683.1 XP_016869172.1 4 - 5 566 154
          XM_017013684.1 XP_016869173.1 4 - 5 566 154
          XM_017013685.1 XP_016869174.1 4 - 5 510 154
          XM_017013686.1 XP_016869175.1 3 - 4 463 154
          XM_017013687.1 XP_016869176.1 4 - 5 566 154
          XM_017013688.1 XP_016869177.1 4 - 5 566 154
          XM_017013689.1 XP_016869178.1 3 - 4 431 154
          XM_017013690.1 XP_016869179.1 3 - 4 405 154
          GenBank mRNA AB209071.1 - 3 - 4 411 154
          DA994616.1 - 3 - 4 364 154

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          microtubule cytoskeleton organization
          angiogenesis
          vasculogenesis
          neuron migration
          placenta development
          regulation of protein phosphorylation
          positive regulation of protein phosphorylation
          heart morphogenesis
          cellular component disassembly involved in execution phase of apoptosis
          nuclear migration
          signal complex assembly
          epidermal growth factor receptor signaling pathway
          transforming growth factor beta receptor signaling pathway
          integrin-mediated signaling pathway
          axon guidance
          positive regulation of cell population proliferation
          regulation of cell shape
          embryo development
          regulation of endothelial cell migration
          regulation of epithelial cell migration
          positive regulation of phosphatidylinositol 3-kinase signaling
          peptidyl-tyrosine phosphorylation
          central nervous system neuron axonogenesis
          negative regulation of cell-cell adhesion
          establishment of cell polarity
          extracellular matrix organization
          positive regulation of cell migration
          regulation of cell adhesion mediated by integrin
          netrin-activated signaling pathway
          peptidyl-tyrosine autophosphorylation
          Fc-gamma receptor signaling pathway involved in phagocytosis
          regulation of cell population proliferation
          negative regulation of apoptotic process
          regulation of GTPase activity
          endothelial cell migration
          positive regulation of GTPase activity
          positive regulation of phosphatidylinositol 3-kinase activity
          innate immune response
          regulation of osteoblast differentiation
          positive regulation of protein kinase activity
          negative regulation of organ growth
          protein autophosphorylation
          vascular endothelial growth factor receptor signaling pathway
          ephrin receptor signaling pathway
          cell motility
          negative regulation of axonogenesis
          regulation of cytoskeleton organization
          regulation of focal adhesion assembly
          positive regulation of protein kinase B signaling
          negative regulation of synapse assembly
          growth hormone receptor signaling pathway
          regulation of substrate adhesion-dependent cell spreading
          positive regulation of ubiquitin-dependent protein catabolic process
          negative regulation of anoikis
          actin binding
          protein kinase activity
          protein tyrosine kinase activity
          non-membrane spanning protein tyrosine kinase activity
          signaling receptor binding
          protein binding
          ATP binding
          JUN kinase binding
          protein kinase binding
          SH2 domain binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline