Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01067421_m1
          See other MCM8 GE Assays ›
          Gene Symbol:
          MCM8
          Gene Name:
          minichromosome maintenance 8 homologous recombination repair factor
          Gene Aliases:
          C20orf154, POF10, dJ967N21.5
          Chromosome Location:
          Chr.20: 5950652 - 6000941 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01067421_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MCM8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001281520.1 NP_001268449.1 17 - 18 2672 107
          NM_001281521.1 NP_001268450.1 17 - 18 2738 107
          NM_001281522.1 NP_001268451.1 16 - 17 2477 107
          NM_032485.5 NP_115874.3 17 - 18 2618 107
          NM_182802.2 NP_877954.1 17 - 18 2624 107
          XM_017028105.1 XP_016883594.1 17 - 18 2767 107
          XM_017028106.1 XP_016883595.1 17 - 18 2584 107
          XM_017028107.1 XP_016883596.1 10 - 11 2857 107
          GenBank mRNA AJ439063.1 - 17 - 18 2618 107
          AK027644.1 - 11 - 12 1657 107
          AK314654.1 - 17 - 18 2300 107
          AY158211.1 - 17 - 18 2323 107
          BC005170.1 - 17 - 18 2323 107
          BC008830.2 - 14 - 15 1928 107
          BC080656.1 - 17 - 18 2323 4841271 107
          BC101054.1 - 16 - 17 2278 40008094 107
          BC101055.1 - 17 - 18 2539 40008095 107
          BC101056.1 - 17 - 18 2419 40008096 107
          BC101057.1 - 17 - 18 2473 40008097 107
          KU178803.1 - 11 - 12 1530 107
          KU178804.1 - 12 - 13 1621 107
          KU178805.1 - 10 - 11 1389 107

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC2000006628.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC20000063.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene MCM8

          Gene Ontology Categories:

          Function(s) Process(es)

          G1/S transition of mitotic cell cycle
          double-strand break repair via homologous recombination
          DNA replication
          cellular response to DNA damage stimulus
          female gamete generation
          male gamete generation
          DNA binding
          helicase activity
          protein binding
          ATP binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR20000954.hg.1 TC20000063.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR20000954.hg.1 ENST00000265187.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20000954.hg.1 ENST00000378883.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20000954.hg.1 ENST00000378886.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20000954.hg.1 ENST00000378896.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20000954.hg.1 NM_032485.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR20000954.hg.1 NM_182802.2 Refseq
          Affymetrix Human Clariom D Assay PSR2000145489.hg.1 ENST00000265187.4 Ensembl
          Affymetrix Human Clariom D Assay PSR2000145489.hg.1 ENST00000378883.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2000145489.hg.1 ENST00000378886.6 Ensembl
          Affymetrix Human Clariom D Assay PSR2000145489.hg.1 ENST00000378896.7 Ensembl
          Affymetrix Human Clariom D Assay PSR2000145489.hg.1 ENST00000610722.4 Ensembl
          Affymetrix Human Clariom D Assay PSR2000145489.hg.1 NM_001281520.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000145489.hg.1 NM_001281521.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000145489.hg.1 NM_001281522.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000145489.hg.1 NM_032485.5 Refseq
          Affymetrix Human Clariom D Assay PSR2000145489.hg.1 NM_182802.2 Refseq
          Affymetrix Human Clariom D Assay PSR2000145489.hg.1 NM_001281521.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000145489.hg.1 TC2000006628.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2000145489.hg.1 ENST00000265187.4 Ensembl
          Affymetrix Human Clariom D Assay PSR2000145489.hg.1 ENST00000378883.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2000145489.hg.1 ENST00000378886.6 Ensembl
          Affymetrix Human Clariom D Assay PSR2000145489.hg.1 ENST00000378896.7 Ensembl
          Affymetrix Human Clariom D Assay PSR2000145489.hg.1 ENST00000610722.4 Ensembl
          Affymetrix Human Clariom D Assay PSR2000145489.hg.1 NM_001281520.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000145489.hg.1 NM_001281522.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000145489.hg.1 NM_032485.5 Refseq
          Affymetrix Human Clariom D Assay PSR2000145489.hg.1 NM_182802.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0