Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06009967.hg.1 |
ENST00000369089.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06009967.hg.1 |
ENST00000369091.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06009967.hg.1 |
ENST00000369096.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06009967.hg.1 |
ENST00000456278.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06009967.hg.1 |
NM_001198.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06009967.hg.1 |
NM_182907.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06009967.hg.1 |
TC06000844.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
ENST00000369089.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
ENST00000369089.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
ENST00000369089.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
ENST00000369089.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
ENST00000369089.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
ENST00000369089.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
ENST00000369089.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
ENST00000369089.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
ENST00000369089.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
ENST00000369091.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
ENST00000369091.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
ENST00000369091.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
ENST00000369091.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
ENST00000369091.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
ENST00000369091.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
ENST00000369091.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
ENST00000369091.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
ENST00000369091.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
ENST00000369091.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
ENST00000369096.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
ENST00000369096.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
ENST00000369096.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
ENST00000369096.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
ENST00000369096.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
ENST00000369096.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
ENST00000369096.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
ENST00000369096.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
ENST00000369096.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
ENST00000369096.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
ENST00000369096.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
ENST00000369096.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
ENST00000369096.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
ENST00000369096.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
ENST00000369096.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
ENST00000424894.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
ENST00000424894.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
ENST00000424894.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
ENST00000450060.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
ENST00000450060.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
ENST00000450060.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
ENST00000481163.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
ENST00000481163.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
ENST00000481163.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
ENST00000489365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
ENST00000489365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
ENST00000489365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
ENST00000613999.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
ENST00000613999.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
ENST00000613999.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
ENST00000613999.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
ENST00000613999.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
ENST00000613999.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
ENST00000613999.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
ENST00000613999.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
ENST00000613999.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
ENST00000613999.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
ENST00000613999.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
ENST00000613999.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
ENST00000613999.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
NM_001198.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
NM_001198.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
NM_001198.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
NM_001198.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
NM_001198.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
NM_001198.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
NM_001198.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
NM_001198.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
NM_001198.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
NM_001198.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
NM_001198.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
NM_001198.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
NM_001198.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
NM_001198.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
NM_001198.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
NM_182907.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
NM_182907.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
NM_182907.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
NM_182907.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
NM_182907.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
NM_182907.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
NM_182907.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
NM_182907.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
NM_182907.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008972.hg.1 |
NM_182907.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
ENST00000369089.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
ENST00000369089.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
ENST00000369089.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
ENST00000369089.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
ENST00000369089.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
ENST00000369089.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
ENST00000369089.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
ENST00000369089.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
ENST00000369089.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
ENST00000369091.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
ENST00000369091.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
ENST00000369091.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
ENST00000369091.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
ENST00000369091.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
ENST00000369091.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
ENST00000369091.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
ENST00000369091.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
ENST00000369091.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
ENST00000369091.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
ENST00000369096.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
ENST00000369096.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
ENST00000369096.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
ENST00000369096.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
ENST00000369096.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
ENST00000369096.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
ENST00000369096.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
ENST00000369096.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
ENST00000369096.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
ENST00000369096.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
ENST00000369096.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
ENST00000369096.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
ENST00000369096.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
ENST00000369096.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
ENST00000369096.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
ENST00000424894.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
ENST00000424894.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
ENST00000424894.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
ENST00000450060.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
ENST00000450060.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
ENST00000450060.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
ENST00000481163.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
ENST00000481163.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
ENST00000481163.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
ENST00000489365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
ENST00000489365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
ENST00000489365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
ENST00000613999.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
ENST00000613999.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
ENST00000613999.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
ENST00000613999.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
ENST00000613999.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
ENST00000613999.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
ENST00000613999.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
ENST00000613999.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
ENST00000613999.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
ENST00000613999.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
ENST00000613999.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
ENST00000613999.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
ENST00000613999.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
NM_001198.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
NM_001198.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
NM_001198.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
NM_001198.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
NM_001198.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
NM_001198.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
NM_001198.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
NM_001198.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
NM_001198.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
NM_001198.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
NM_001198.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
NM_001198.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
NM_001198.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
NM_001198.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
NM_001198.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
NM_182907.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
NM_182907.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
NM_182907.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
NM_182907.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
NM_182907.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
NM_182907.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
NM_182907.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
NM_182907.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
NM_182907.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008972.hg.1 |
NM_182907.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600161885.hg.1 |
ENST00000369089.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600161885.hg.1 |
ENST00000369091.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600161885.hg.1 |
ENST00000369096.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600161885.hg.1 |
ENST00000613999.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600161885.hg.1 |
NM_001198.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600161885.hg.1 |
NM_182907.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600161885.hg.1 |
TC0600008972.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600161885.hg.1 |
ENST00000369096.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600161885.hg.1 |
ENST00000369089.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600161885.hg.1 |
ENST00000369091.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600161885.hg.1 |
ENST00000613999.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600161885.hg.1 |
NM_001198.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600161885.hg.1 |
NM_182907.2 |
Refseq |