Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01069007_g1
          See other SMG7-AS1 GE Assays ›
          Gene Symbol:
          SMG7-AS1
          Gene Name:
          SMG7 antisense RNA 1
          Gene Aliases:
          DKFZP564C196
          Chromosome Location:
          Chr.1: 183460874 - 183471982 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01069007_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SMG7-AS1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040063.1 4 - 5 1654 97
          GenBank mRNA BC032873.2 - 4 - 5 1418 97

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100016652.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01003615.hg.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20707200 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00647413 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20707197 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15150484 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20707185 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00647401 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20707211 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20707196 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00647404 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00647418 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20707195 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00647402 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20707206 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20707190 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00647392 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20707210 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00647414 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20707183 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00647419 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20707213 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 164849 AB020670.1 genbank_mrna_human
          Agilent SurePrint G3 Human CGH Microarray 4x180K 164849 AK023032.1 genbank_mrna_human
          Agilent SurePrint G3 Human CGH Microarray 4x180K 164849 BC071589.1 genbank_mrna_human
          Agilent SurePrint G3 Human CGH Microarray 4x180K 164849 NM_014913.2 refseq_mrna_human_NM
          Agilent SurePrint G3 Human CGH Microarray 8x60K 22470 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 498730 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 927108 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 30247 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 651105 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3461129 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P412789 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P281228 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 43047 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 4555 - -
          Agilent Human Genome CGH Microarray 44B A_14_P110947 - -
          Agilent Human Genome CGH Microarray 44A A_14_P110947 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 40750 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3461129 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P412789 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P281228 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_14_P110947 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_14_P110947 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P281228 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P412789 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 32313 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 20675 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_14_P110947 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P15368174 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P15368185 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P15368180 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P15368165 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P15368174 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_14_P110947 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 215953_PM_at - -
          Affymetrix HT HG-U133+ PM Array Plate 222384_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR01055603.hg.1 ENST00000421703.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01055603.hg.1 NR_040063.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01055603.hg.1 TC01003615.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0100016652.hg.1 ENST00000421703.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016652.hg.1 ENST00000421703.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016652.hg.1 ENST00000421703.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016652.hg.1 ENST00000421703.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016652.hg.1 ENST00000421703.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016652.hg.1 ENST00000421703.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016652.hg.1 ENST00000421703.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016652.hg.1 ENST00000421703.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016652.hg.1 ENST00000421703.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016652.hg.1 ENST00000421703.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016652.hg.1 ENST00000421703.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016652.hg.1 ENST00000421703.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016652.hg.1 ENST00000421703.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016652.hg.1 ENST00000421703.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016652.hg.1 ENST00000421703.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016652.hg.1 ENST00000421703.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016652.hg.1 ENST00000421703.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016652.hg.1 ENST00000421703.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016652.hg.1 ENST00000432837.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016652.hg.1 ENST00000432837.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016652.hg.1 ENST00000432837.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016652.hg.1 ENST00000432837.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016652.hg.1 ENST00000432837.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016652.hg.1 ENST00000432837.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016652.hg.1 ENST00000624060.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016652.hg.1 ENST00000624060.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016652.hg.1 ENST00000624060.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016652.hg.1 ENST00000624060.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016652.hg.1 ENST00000624060.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016652.hg.1 ENST00000624060.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016652.hg.1 ENST00000624060.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016652.hg.1 ENST00000624060.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016652.hg.1 NR_040063.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016652.hg.1 NR_040063.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016652.hg.1 NR_040063.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016652.hg.1 NR_040063.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016652.hg.1 NR_040063.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016652.hg.1 NR_040063.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016652.hg.1 NR_040063.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016652.hg.1 NR_040063.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016652.hg.1 NR_040063.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100016652.hg.1 NR_040063.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01055601.hg.1 ENST00000421703.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01055601.hg.1 NR_040063.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01055601.hg.1 TC01003615.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0100226316.hg.1 ENST00000421703.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100226316.hg.1 ENST00000421703.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100226316.hg.1 ENST00000432837.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100226316.hg.1 NR_040063.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100226316.hg.1 TC0100016652.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100226316.hg.1 NR_040063.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100226316.hg.1 ENST00000421703.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100226316.hg.1 ENST00000421703.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100226316.hg.1 ENST00000432837.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100226314.hg.1 ENST00000421703.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100226314.hg.1 ENST00000421703.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100226314.hg.1 NR_040063.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100226314.hg.1 TC0100016652.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100226314.hg.1 ENST00000421703.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100226314.hg.1 ENST00000421703.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100226314.hg.1 NR_040063.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100226317.hg.1 ENST00000421703.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100226317.hg.1 ENST00000421703.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100226317.hg.1 ENST00000432837.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100226317.hg.1 NR_040063.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100226317.hg.1 TC0100016652.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100226317.hg.1 ENST00000421703.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100226317.hg.1 ENST00000421703.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100226317.hg.1 ENST00000432837.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100226317.hg.1 NR_040063.1 Refseq
          Affymetrix Human Clariom D Assay TC0100016652.hg.1 ENST00000421703.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016652.hg.1 ENST00000421703.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016652.hg.1 ENST00000421703.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016652.hg.1 ENST00000421703.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016652.hg.1 ENST00000421703.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016652.hg.1 ENST00000421703.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016652.hg.1 ENST00000421703.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016652.hg.1 ENST00000421703.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016652.hg.1 ENST00000421703.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016652.hg.1 ENST00000421703.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016652.hg.1 ENST00000421703.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016652.hg.1 ENST00000421703.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016652.hg.1 ENST00000421703.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016652.hg.1 ENST00000421703.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016652.hg.1 ENST00000421703.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016652.hg.1 ENST00000421703.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016652.hg.1 ENST00000421703.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016652.hg.1 ENST00000421703.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016652.hg.1 ENST00000432837.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016652.hg.1 ENST00000432837.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016652.hg.1 ENST00000432837.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016652.hg.1 ENST00000432837.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016652.hg.1 ENST00000432837.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016652.hg.1 ENST00000432837.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016652.hg.1 ENST00000624060.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016652.hg.1 ENST00000624060.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016652.hg.1 ENST00000624060.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016652.hg.1 ENST00000624060.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016652.hg.1 ENST00000624060.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016652.hg.1 ENST00000624060.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016652.hg.1 ENST00000624060.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016652.hg.1 ENST00000624060.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016652.hg.1 NR_040063.1 Refseq
          Affymetrix Human Clariom D Assay TC0100016652.hg.1 NR_040063.1 Refseq
          Affymetrix Human Clariom D Assay TC0100016652.hg.1 NR_040063.1 Refseq
          Affymetrix Human Clariom D Assay TC0100016652.hg.1 NR_040063.1 Refseq
          Affymetrix Human Clariom D Assay TC0100016652.hg.1 NR_040063.1 Refseq
          Affymetrix Human Clariom D Assay TC0100016652.hg.1 NR_040063.1 Refseq
          Affymetrix Human Clariom D Assay TC0100016652.hg.1 NR_040063.1 Refseq
          Affymetrix Human Clariom D Assay TC0100016652.hg.1 NR_040063.1 Refseq
          Affymetrix Human Clariom D Assay TC0100016652.hg.1 NR_040063.1 Refseq
          Affymetrix Human Clariom D Assay TC0100016652.hg.1 NR_040063.1 Refseq
          Human Genome U133 Plus 2.0 Array 222384_at - -
          Human Genome U133 Plus 2.0 Array 222384_PM_at - -
          Human Genome U133 Plus 2.0 Array 215953_at - -
          Human Genome U133 Plus 2.0 Array 215953_PM_at - -
          Human Genome U133A 2.0 Array 215953_at - -
          Human Genome U133A 2.0 Array 222384_at - -
          Human Genome U133A 2.0 Array 215953_PM_at - -
          Human Genome U133A 2.0 Array 222384_PM_at - -
          AffymetrixPathwork Diagnostics Pathchip pw525759 - -
          AffymetrixPathwork Diagnostics Pathchip pw583197 - -
          Human Genome U95 Set 35516_PM_at - -
          Human Genome U95 Set 35516_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 222384_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 215953_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 215953_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 222384_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 215953_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 222384_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 222384_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 222384_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 215953_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 215953_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 222384_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 222384_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 215953_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 215953_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 222384_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 215953_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 215953_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 222384_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 215953_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 222384_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 215953_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 222384_at6 - -
          Human Genome U95 Set 68960_at - -
          Human Genome U95 Set 68960_PM_at - -
          Human Genome U133 Set 215953_at - -
          Human Genome U133 Set 215953_PM_at - -
          Human Genome U133 Set 222384_at - -
          Human Genome U133 Set 222384_PM_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A008745 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A008745 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1824647 AL050020.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1824647 AL050020.1 human_transcriptome
          Illumina Sentrix Human-6 v2 Expression BeadChip 380014 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_76417 - -
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1824647 AL050020.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1824647 AL050020.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0