Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02037603.hg.1 |
ENST00000389484.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02037603.hg.1 |
ENST00000544579.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02037603.hg.1 |
NM_018557.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02037603.hg.1 |
TC02002388.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0200205319.hg.1 |
ENST00000389484.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200205319.hg.1 |
ENST00000437977.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200205319.hg.1 |
ENST00000618808.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200205319.hg.1 |
NM_018557.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200205319.hg.1 |
TC0200014468.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200205319.hg.1 |
ENST00000389484.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200205319.hg.1 |
ENST00000437977.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200205319.hg.1 |
ENST00000618808.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200205319.hg.1 |
NM_018557.2 |
Refseq |