Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01069294_m1
          See other DMBT1 GE Assays ›
          Gene Symbol:
          DMBT1
          Gene Name:
          deleted in malignant brain tumors 1
          Gene Aliases:
          GP340, SAG, muclin
          Chromosome Location:
          Chr.10: 122560665 - 122643736 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01069294_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: DMBT1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001320644.1 NP_001307573.1 40 - 41 5006 66
          NM_004406.2 NP_004397.2 27 - 28 3122 66
          NM_007329.2 NP_015568.2 40 - 41 5006 66
          NM_017579.2 NP_060049.2 39 - 40 4976 66
          XM_006717660.3 XP_006717723.1 42 - 43 5333 66
          XM_006717665.3 XP_006717728.1 41 - 42 5285 66
          XM_011539388.2 XP_011537690.1 43 - 44 5381 66
          XM_011539389.2 XP_011537691.1 43 - 44 5381 66
          XM_011539390.2 XP_011537692.1 42 - 43 5351 66
          XM_011539391.2 XP_011537693.1 42 - 43 5351 66
          XM_011539392.2 XP_011537694.1 42 - 43 5351 66
          XM_011539393.2 XP_011537695.1 42 - 43 5351 66
          XM_011539394.2 XP_011537696.1 42 - 43 5351 66
          XM_011539395.2 XP_011537697.1 42 - 43 5351 66
          XM_011539396.2 XP_011537698.1 42 - 43 5333 66
          XM_011539398.2 XP_011537700.1 41 - 42 5318 66
          XM_011539399.2 XP_011537701.1 41 - 42 5285 66
          XM_011539400.2 XP_011537702.1 42 - 43 5057 66
          XM_011539401.2 XP_011537703.1 40 - 41 4994 66
          XM_011539402.2 XP_011537704.1 40 - 41 4994 66
          XM_011539403.2 XP_011537705.1 40 - 41 4994 66
          XM_011539405.2 XP_011537707.1 40 - 41 4995 66
          XM_011539407.2 XP_011537709.1 40 - 41 4988 66
          XM_011539408.2 XP_011537710.1 40 - 41 4988 66
          XM_011539409.2 XP_011537711.1 40 - 41 4988 66
          XM_011539411.2 XP_011537713.1 39 - 40 4964 66
          XM_011539413.2 XP_011537715.1 37 - 38 4600 66
          XM_011539414.2 XP_011537716.1 27 - 28 3101 66
          XM_017015798.1 XP_016871287.1 28 - 29 3643 66
          GenBank mRNA AF159456.1 - 40 - 41 5006 66
          AJ000342.1 - 27 - 28 3122 66
          AJ243212.1 - 40 - 41 5006 66
          AJ243224.1 - 39 - 40 4976 66
          AJ297935.1 - 40 - 41 4987 66
          BC153299.1 - 24 - 25 2740 40147087 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1000009155.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC10000890.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene DMBT1

          Gene Ontology Categories:

          Function(s) Process(es)

          pattern recognition receptor signaling pathway
          receptor-mediated endocytosis
          multicellular organism development
          protein transport
          epithelial cell differentiation
          induction of bacterial agglutination
          cellular protein metabolic process
          innate immune response
          defense response to virus
          scavenger receptor activity
          protein binding
          zymogen binding
          pattern recognition receptor activity
          calcium-dependent protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR10010888.hg.1 ENST00000327438.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10010888.hg.1 ENST00000330163.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10010888.hg.1 ENST00000338354.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10010888.hg.1 ENST00000339712.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10010888.hg.1 ENST00000339871.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10010888.hg.1 ENST00000341278.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10010888.hg.1 ENST00000344338.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10010888.hg.1 ENST00000344467.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10010888.hg.1 ENST00000368909.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10010888.hg.1 ENST00000368915.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10010888.hg.1 ENST00000368942.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10010888.hg.1 ENST00000368953.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10010888.hg.1 ENST00000368955.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10010888.hg.1 ENST00000368956.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10010888.hg.1 NM_004406.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10010888.hg.1 NM_007329.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10010888.hg.1 NM_017579.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10010888.hg.1 TC10000890.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1000162772.hg.1 ENST00000330163.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1000162772.hg.1 ENST00000338354.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1000162772.hg.1 ENST00000344338.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1000162772.hg.1 ENST00000368909.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1000162772.hg.1 ENST00000368955.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1000162772.hg.1 ENST00000368956.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1000162772.hg.1 ENST00000619379.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000162772.hg.1 NM_004406.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000162772.hg.1 NM_007329.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000162772.hg.1 NM_017579.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000162772.hg.1 TC1000009155.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000162772.hg.1 ENST00000330163.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1000162772.hg.1 ENST00000338354.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1000162772.hg.1 ENST00000344338.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1000162772.hg.1 ENST00000368909.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1000162772.hg.1 ENST00000368955.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1000162772.hg.1 ENST00000368956.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1000162772.hg.1 ENST00000619379.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000162772.hg.1 NM_004406.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000162772.hg.1 NM_007329.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000162772.hg.1 NM_017579.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-865d7754c7-mcq8j:80/100.66.130.62:80.
          git-commit: 7ff36f61b459eb8f02ba5a263f0e3ce367ca82d3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0