Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01069298_m1
          See other DMBT1 GE Assays ›
          Gene Symbol:
          DMBT1
          Gene Name:
          deleted in malignant brain tumors 1
          Gene Aliases:
          GP340, SAG, muclin
          Chromosome Location:
          Chr.10: 122560665 - 122643736 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01069298_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: DMBT1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001320644.1 NP_001307573.1 43 - 44 5386 69
          NM_004406.2 NP_004397.2 30 - 31 3502 69
          NM_007329.2 NP_015568.2 43 - 44 5386 69
          NM_017579.2 NP_060049.2 42 - 43 5356 69
          XM_006717660.3 XP_006717723.1 45 - 46 5713 69
          XM_006717665.3 XP_006717728.1 44 - 45 5665 69
          XM_011539388.2 XP_011537690.1 46 - 47 5761 69
          XM_011539389.2 XP_011537691.1 46 - 47 5761 69
          XM_011539390.2 XP_011537692.1 45 - 46 5731 69
          XM_011539391.2 XP_011537693.1 45 - 46 5731 69
          XM_011539392.2 XP_011537694.1 45 - 46 5731 69
          XM_011539393.2 XP_011537695.1 45 - 46 5731 69
          XM_011539394.2 XP_011537696.1 45 - 46 5731 69
          XM_011539395.2 XP_011537697.1 45 - 46 5731 69
          XM_011539396.2 XP_011537698.1 45 - 46 5713 69
          XM_011539398.2 XP_011537700.1 44 - 45 5698 69
          XM_011539399.2 XP_011537701.1 44 - 45 5665 69
          XM_011539400.2 XP_011537702.1 45 - 46 5437 69
          XM_011539401.2 XP_011537703.1 43 - 44 5374 69
          XM_011539402.2 XP_011537704.1 43 - 44 5374 69
          XM_011539403.2 XP_011537705.1 43 - 44 5374 69
          XM_011539405.2 XP_011537707.1 43 - 44 5375 69
          XM_011539407.2 XP_011537709.1 43 - 44 5368 69
          XM_011539408.2 XP_011537710.1 43 - 44 5368 69
          XM_011539409.2 XP_011537711.1 43 - 44 5368 69
          XM_011539410.2 XP_011537712.1 43 - 44 5368 69
          XM_011539411.2 XP_011537713.1 42 - 43 5344 69
          XM_011539413.2 XP_011537715.1 40 - 41 4980 69
          XM_011539414.2 XP_011537716.1 30 - 31 3481 69
          XM_011539415.2 XP_011537717.1 28 - 29 3435 69
          XM_017015798.1 XP_016871287.1 31 - 32 4023 69
          GenBank mRNA AF159456.1 - 43 - 44 5386 69
          AJ000342.1 - 30 - 31 3502 69
          AJ243212.1 - 43 - 44 5386 69
          AJ243224.1 - 42 - 43 5356 69
          AJ297935.1 - 43 - 44 5367 69
          AK304128.1 - 13 - 14 1236 69
          AK304149.1 - 17 - 18 1526 69
          BC153299.1 - 27 - 28 3120 40147087 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1000009155.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC10000890.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene DMBT1

          Gene Ontology Categories:

          Function(s) Process(es)

          pattern recognition receptor signaling pathway
          receptor-mediated endocytosis
          multicellular organism development
          protein transport
          epithelial cell differentiation
          induction of bacterial agglutination
          cellular protein metabolic process
          innate immune response
          defense response to virus
          scavenger receptor activity
          protein binding
          zymogen binding
          pattern recognition receptor activity
          calcium-dependent protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR10010891.hg.1 ENST00000327438.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10010891.hg.1 ENST00000330163.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10010891.hg.1 ENST00000338354.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10010891.hg.1 ENST00000339712.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10010891.hg.1 ENST00000339871.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10010891.hg.1 ENST00000341278.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10010891.hg.1 ENST00000344338.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10010891.hg.1 ENST00000344467.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10010891.hg.1 ENST00000359586.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10010891.hg.1 ENST00000368909.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10010891.hg.1 ENST00000368915.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10010891.hg.1 ENST00000368942.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10010891.hg.1 ENST00000368953.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10010891.hg.1 ENST00000368955.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10010891.hg.1 ENST00000368956.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10010891.hg.1 NM_004406.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10010891.hg.1 NM_007329.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10010891.hg.1 NM_017579.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10010891.hg.1 TC10000890.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1000162775.hg.1 ENST00000330163.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1000162775.hg.1 ENST00000338354.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1000162775.hg.1 ENST00000344338.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1000162775.hg.1 ENST00000359586.10 Ensembl
          Affymetrix Human Clariom D Assay PSR1000162775.hg.1 ENST00000368909.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1000162775.hg.1 ENST00000368955.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1000162775.hg.1 ENST00000368956.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1000162775.hg.1 ENST00000619379.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000162775.hg.1 NM_004406.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000162775.hg.1 NM_007329.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000162775.hg.1 NM_017579.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000162775.hg.1 TC1000009155.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000162775.hg.1 ENST00000359586.10 Ensembl
          Affymetrix Human Clariom D Assay PSR1000162775.hg.1 ENST00000330163.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1000162775.hg.1 ENST00000338354.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1000162775.hg.1 ENST00000344338.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1000162775.hg.1 ENST00000368909.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1000162775.hg.1 ENST00000368955.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1000162775.hg.1 ENST00000368956.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1000162775.hg.1 ENST00000619379.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000162775.hg.1 NM_004406.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000162775.hg.1 NM_007329.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000162775.hg.1 NM_017579.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-786bfc498d-47c8f:80/100.66.130.62:80.
          git-commit: 9eb01563f6748445c58f67612c67ecf6b1da7421
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0