Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19002247.hg.1 |
NM_001195259.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19002247.hg.1 |
TC19000141.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006865.hg.1 |
ENST00000564348.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006865.hg.1 |
ENST00000564348.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006865.hg.1 |
ENST00000564348.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006865.hg.1 |
ENST00000564348.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006865.hg.1 |
ENST00000564348.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006865.hg.1 |
ENST00000564348.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006865.hg.1 |
ENST00000564348.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006865.hg.1 |
ENST00000564348.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006865.hg.1 |
ENST00000564348.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006865.hg.1 |
ENST00000565886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006865.hg.1 |
ENST00000565886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006865.hg.1 |
ENST00000565886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006865.hg.1 |
ENST00000565886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006865.hg.1 |
ENST00000565886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006865.hg.1 |
ENST00000565886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006865.hg.1 |
ENST00000565886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006865.hg.1 |
ENST00000565886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006865.hg.1 |
ENST00000565886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006865.hg.1 |
ENST00000565886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006865.hg.1 |
ENST00000566166.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006865.hg.1 |
ENST00000566166.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006865.hg.1 |
ENST00000566166.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006865.hg.1 |
ENST00000566166.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006865.hg.1 |
ENST00000566166.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006865.hg.1 |
ENST00000566166.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006865.hg.1 |
NM_001195259.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006865.hg.1 |
NM_001195259.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006865.hg.1 |
NM_001195259.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006865.hg.1 |
NM_001195259.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006865.hg.1 |
NM_001195259.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006865.hg.1 |
NM_001195259.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006865.hg.1 |
NM_001195259.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006865.hg.1 |
NM_001195259.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006865.hg.1 |
NM_001195259.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006865.hg.1 |
NM_001195259.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900148667.hg.1 |
ENST00000564348.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900148667.hg.1 |
ENST00000565886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900148667.hg.1 |
ENST00000566166.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900148667.hg.1 |
NM_001195259.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900148667.hg.1 |
TC1900006865.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900148667.hg.1 |
NM_001195259.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900148667.hg.1 |
ENST00000564348.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900148667.hg.1 |
ENST00000565886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900148667.hg.1 |
ENST00000566166.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006865.hg.1 |
ENST00000564348.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006865.hg.1 |
ENST00000564348.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006865.hg.1 |
ENST00000564348.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006865.hg.1 |
ENST00000564348.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006865.hg.1 |
ENST00000564348.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006865.hg.1 |
ENST00000564348.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006865.hg.1 |
ENST00000564348.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006865.hg.1 |
ENST00000564348.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006865.hg.1 |
ENST00000564348.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006865.hg.1 |
ENST00000565886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006865.hg.1 |
ENST00000565886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006865.hg.1 |
ENST00000565886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006865.hg.1 |
ENST00000565886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006865.hg.1 |
ENST00000565886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006865.hg.1 |
ENST00000565886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006865.hg.1 |
ENST00000565886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006865.hg.1 |
ENST00000565886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006865.hg.1 |
ENST00000565886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006865.hg.1 |
ENST00000565886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006865.hg.1 |
ENST00000566166.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006865.hg.1 |
ENST00000566166.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006865.hg.1 |
ENST00000566166.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006865.hg.1 |
ENST00000566166.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006865.hg.1 |
ENST00000566166.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006865.hg.1 |
ENST00000566166.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006865.hg.1 |
NM_001195259.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006865.hg.1 |
NM_001195259.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006865.hg.1 |
NM_001195259.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006865.hg.1 |
NM_001195259.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006865.hg.1 |
NM_001195259.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006865.hg.1 |
NM_001195259.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006865.hg.1 |
NM_001195259.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006865.hg.1 |
NM_001195259.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006865.hg.1 |
NM_001195259.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006865.hg.1 |
NM_001195259.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900148670.hg.1 |
ENST00000564348.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900148670.hg.1 |
ENST00000565886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900148670.hg.1 |
ENST00000566166.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900148670.hg.1 |
NM_001195259.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900148670.hg.1 |
TC1900006865.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900148670.hg.1 |
ENST00000564348.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900148670.hg.1 |
ENST00000565886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900148670.hg.1 |
ENST00000566166.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900148670.hg.1 |
NM_001195259.1 |
Refseq |