Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NALCN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_052867.2 | NP_443099.1 | 35 - 36 | 4140 | 80 | |
XM_011521067.2 | XP_011519369.1 | 35 - 36 | 4080 | 80 | ||
XM_011521068.2 | XP_011519370.1 | 35 - 36 | 4063 | 80 | ||
XM_011521069.2 | XP_011519371.1 | 34 - 35 | 3993 | 80 | ||
XM_017020534.1 | XP_016876023.1 | 34 - 35 | 4024 | 80 | ||
XM_017020535.1 | XP_016876024.1 | 34 - 35 | 3966 | 80 | ||
XM_017020536.1 | XP_016876025.1 | 30 - 31 | 3892 | 80 | ||
XM_017020537.1 | XP_016876026.1 | 29 - 30 | 3281 | 80 | ||
GenBank mRNA | AK055506.1 | - | 2 - 3 | 197 | 80 | |
AK126621.1 | - | 16 - 17 | 1604 | 80 | ||
AK126718.1 | - | 8 - 9 | 741 | 80 | ||
AY141972.1 | - | 35 - 36 | 4034 | 80 |
Set Membership: |
![]() ![]() ![]() ![]() |