Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01076088_m1
          See other EGFR GE Assays ›
          Gene Symbol:
          EGFR
          Gene Name:
          epidermal growth factor receptor
          Gene Aliases:
          ERBB, ERBB1, HER1, NISBD2, PIG61, mENA
          Chromosome Location:
          Chr.7: 55019032 - 55207338 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01076088_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: EGFR
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_005228.3 NP_005219.2 3 - 4 671 71
          NM_201282.1 NP_958439.1 3 - 4 671 71
          NM_201283.1 NP_958440.1 3 - 4 671 71
          NM_201284.1 NP_958441.1 3 - 4 671 71
          GenBank mRNA AB209442.1 - 3 - 4 571 71
          AF125253.1 - 3 - 4 670 71
          AK127817.1 - 1 - 2 969 71
          AK225422.1 - 3 - 4 586 71
          AK290352.1 - 3 - 4 471 71
          AK294750.1 - 3 - 4 449 71
          AY550966.1 - 3 - 4 471 71
          AY698023.1 - 3 - 4 425 71
          AY698024.1 - 3 - 4 425 71
          DQ088980.1 - 3 - 4 471 71
          GU255993.1 - 3 - 4 468 71
          K03193.1 - 3 - 4 584 71
          U48722.1 - 3 - 4 669 71
          U95089.1 - 4 - 5 928 71
          X00588.1 - 3 - 4 611 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0700007596.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC07000328.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Protein Kinase Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene EGFR

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          activation of MAPKK activity
          cell morphogenesis
          ossification
          embryonic placenta development
          positive regulation of protein phosphorylation
          hair follicle development
          response to stress
          signal transduction
          cell surface receptor signaling pathway
          epidermal growth factor receptor signaling pathway
          activation of phospholipase C activity
          salivary gland morphogenesis
          learning or memory
          cell proliferation
          positive regulation of cell proliferation
          regulation of phosphatidylinositol 3-kinase signaling
          single organismal cell-cell adhesion
          peptidyl-tyrosine phosphorylation
          cerebral cortex cell migration
          positive regulation of cell growth
          positive regulation of cell migration
          positive regulation of cyclin-dependent protein serine/threonine kinase activity involved in G1/S transition of mitotic cell cycle
          positive regulation of catenin import into nucleus
          ERBB2 signaling pathway
          negative regulation of epidermal growth factor receptor signaling pathway
          negative regulation of protein catabolic process
          positive regulation of phosphorylation
          activation of phospholipase A2 activity by calcium-mediated signaling
          negative regulation of apoptotic process
          positive regulation of MAP kinase activity
          positive regulation of GTPase activity
          positive regulation of nitric oxide biosynthetic process
          positive regulation of DNA repair
          positive regulation of DNA replication
          positive regulation of transcription from RNA polymerase II promoter
          protein autophosphorylation
          phosphatidylinositol phosphorylation
          phosphatidylinositol-mediated signaling
          positive regulation of fibroblast proliferation
          digestive tract morphogenesis
          positive regulation of epithelial cell proliferation
          regulation of peptidyl-tyrosine phosphorylation
          regulation of nitric-oxide synthase activity
          protein insertion into membrane
          positive regulation of protein kinase B signaling
          morphogenesis of an epithelial fold
          eyelid development in camera-type eye
          response to UV-A
          positive regulation of ERK1 and ERK2 cascade
          cellular response to amino acid stimulus
          cellular response to epidermal growth factor stimulus
          cellular response to estradiol stimulus
          positive regulation of production of miRNAs involved in gene silencing by miRNA
          regulation of cell motility
          chromatin binding
          double-stranded DNA binding
          MAP kinase kinase kinase activity
          protein tyrosine kinase activity
          transmembrane receptor protein tyrosine kinase activity
          receptor signaling protein tyrosine kinase activity
          transmembrane signaling receptor activity
          epidermal growth factor-activated receptor activity
          Ras guanyl-nucleotide exchange factor activity
          protein binding
          ATP binding
          enzyme binding
          protein phosphatase binding
          nitric-oxide synthase regulator activity
          ubiquitin protein ligase binding
          identical protein binding
          phosphatidylinositol-4,5-bisphosphate 3-kinase activity
          protein heterodimerization activity
          actin filament binding
          cadherin binding involved in cell-cell adhesion

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR07004864.hg.1 TC07000328.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR07004864.hg.1 ENST00000275493.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07004864.hg.1 ENST00000342916.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07004864.hg.1 ENST00000344576.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07004864.hg.1 ENST00000395504.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07004864.hg.1 ENST00000420316.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07004864.hg.1 ENST00000442591.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07004864.hg.1 ENST00000454757.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07004864.hg.1 NM_005228.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07004864.hg.1 NM_201282.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07004864.hg.1 NM_201283.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07004864.hg.1 NM_201284.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07004862.hg.1 TC07000328.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR07004862.hg.1 ENST00000275493.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07004862.hg.1 ENST00000342916.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07004862.hg.1 ENST00000344576.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07004862.hg.1 ENST00000395504.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07004862.hg.1 ENST00000420316.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07004862.hg.1 ENST00000442591.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07004862.hg.1 ENST00000454757.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07004862.hg.1 NM_005228.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07004862.hg.1 NM_201282.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07004862.hg.1 NM_201283.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07004862.hg.1 NM_201284.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700152611.hg.1 ENST00000275493.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0700152611.hg.1 ENST00000342916.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0700152611.hg.1 ENST00000344576.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0700152611.hg.1 ENST00000420316.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0700152611.hg.1 ENST00000442591.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700152611.hg.1 ENST00000450046.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700152611.hg.1 NM_005228.4 Refseq
          Affymetrix Human Clariom D Assay PSR0700152611.hg.1 NM_201282.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700152611.hg.1 NM_201283.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700152611.hg.1 NM_201284.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700152611.hg.1 ENST00000275493.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0700152611.hg.1 ENST00000342916.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0700152611.hg.1 ENST00000344576.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0700152611.hg.1 ENST00000420316.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0700152611.hg.1 ENST00000442591.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700152611.hg.1 ENST00000450046.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700152611.hg.1 NM_005228.4 Refseq
          Affymetrix Human Clariom D Assay PSR0700152611.hg.1 NM_201282.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700152611.hg.1 NM_201283.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700152611.hg.1 NM_201284.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700152611.hg.1 TC0700007596.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0