Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01076119_m1
          See other RORC GE Assays ›
          Gene Symbol:
          RORC
          Gene Name:
          RAR related orphan receptor C
          Gene Aliases:
          IMD42, NR1F3, RORG, RZR-GAMMA, RZRG, TOR
          Chromosome Location:
          Chr.1: 151806071 - 151832450 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01076119_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RORC
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001001523.1 NP_001001523.1 5 - 6 1015 62
          NM_005060.3 NP_005051.2 6 - 7 1045 62
          XM_006711484.3 XP_006711547.2 7 - 8 1828 62
          GenBank mRNA AB307696.1 - 6 - 7 937 62
          AK097994.1 - 5 - 6 1029 62
          AK223137.1 - 6 - 7 971 62
          AK298460.1 - 7 - 8 1128 62
          AK315662.1 - 6 - 7 984 62
          AL834219.1 - 5 - 6 1015 62
          BC031554.1 - 6 - 7 963 5186655 62
          BC110571.2 - 6 - 7 997 40001043 62
          CR457436.1 - 6 - 7 937 62
          HQ692834.1 - 4 - 5 874 62
          U16997.1 - 6 - 7 1006 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100018491.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01006373.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene RORC

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription initiation from RNA polymerase II promoter
          xenobiotic metabolic process
          regulation of glucose metabolic process
          regulation of steroid metabolic process
          intracellular receptor signaling pathway
          circadian regulation of gene expression
          cellular response to sterol
          positive regulation of circadian rhythm
          steroid hormone mediated signaling pathway
          regulation of fat cell differentiation
          positive regulation of transcription, DNA-templated
          adipose tissue development
          T-helper 17 cell differentiation
          DNA-binding transcription factor activity
          steroid hormone receptor activity
          nuclear receptor activity
          protein binding
          oxysterol binding
          zinc ion binding
          sequence-specific DNA binding
          ligand-activated transcription factor activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0100018491.hg.1 ENST00000318247.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018491.hg.1 ENST00000318247.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018491.hg.1 ENST00000318247.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018491.hg.1 ENST00000318247.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018491.hg.1 ENST00000318247.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018491.hg.1 ENST00000318247.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018491.hg.1 ENST00000318247.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018491.hg.1 ENST00000318247.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018491.hg.1 ENST00000318247.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018491.hg.1 ENST00000318247.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018491.hg.1 ENST00000318247.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018491.hg.1 ENST00000318247.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018491.hg.1 ENST00000318247.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018491.hg.1 ENST00000318247.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018491.hg.1 ENST00000318247.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018491.hg.1 ENST00000318247.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018491.hg.1 ENST00000356728.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018491.hg.1 ENST00000356728.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018491.hg.1 ENST00000356728.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018491.hg.1 ENST00000356728.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018491.hg.1 ENST00000356728.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018491.hg.1 ENST00000356728.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018491.hg.1 ENST00000356728.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018491.hg.1 ENST00000356728.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018491.hg.1 ENST00000356728.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018491.hg.1 ENST00000356728.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018491.hg.1 ENST00000356728.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018491.hg.1 ENST00000356728.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018491.hg.1 ENST00000356728.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018491.hg.1 ENST00000480719.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018491.hg.1 ENST00000480719.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018491.hg.1 ENST00000480719.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018491.hg.1 ENST00000480719.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018491.hg.1 ENST00000480719.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018491.hg.1 ENST00000480719.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018491.hg.1 ENST00000480719.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018491.hg.1 ENST00000480719.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018491.hg.1 ENST00000480719.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018491.hg.1 ENST00000480719.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018491.hg.1 ENST00000480719.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018491.hg.1 ENST00000480719.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018491.hg.1 ENST00000480719.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018491.hg.1 NM_001001523.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018491.hg.1 NM_001001523.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018491.hg.1 NM_001001523.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018491.hg.1 NM_001001523.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018491.hg.1 NM_001001523.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018491.hg.1 NM_001001523.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018491.hg.1 NM_001001523.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018491.hg.1 NM_001001523.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018491.hg.1 NM_001001523.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018491.hg.1 NM_001001523.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018491.hg.1 NM_001001523.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018491.hg.1 NM_001001523.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018491.hg.1 NM_001001523.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018491.hg.1 NM_005060.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018491.hg.1 NM_005060.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018491.hg.1 NM_005060.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018491.hg.1 NM_005060.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018491.hg.1 NM_005060.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018491.hg.1 NM_005060.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018491.hg.1 NM_005060.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018491.hg.1 NM_005060.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018491.hg.1 NM_005060.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018491.hg.1 NM_005060.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018491.hg.1 NM_005060.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018491.hg.1 NM_005060.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018491.hg.1 NM_005060.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018491.hg.1 NM_005060.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018491.hg.1 NM_005060.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018491.hg.1 NM_005060.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018491.hg.1 NM_005060.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01049954.hg.1 ENST00000318247.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01049954.hg.1 ENST00000356728.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01049954.hg.1 ENST00000392697.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01049954.hg.1 ENST00000480719.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01049954.hg.1 NM_001001523.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01049954.hg.1 NM_005060.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01049954.hg.1 TC01006373.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC0100018491.hg.1 ENST00000318247.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100018491.hg.1 ENST00000318247.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100018491.hg.1 ENST00000318247.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100018491.hg.1 ENST00000318247.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100018491.hg.1 ENST00000318247.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100018491.hg.1 ENST00000318247.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100018491.hg.1 ENST00000318247.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100018491.hg.1 ENST00000318247.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100018491.hg.1 ENST00000318247.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100018491.hg.1 ENST00000318247.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100018491.hg.1 ENST00000318247.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100018491.hg.1 ENST00000318247.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100018491.hg.1 ENST00000318247.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100018491.hg.1 ENST00000318247.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100018491.hg.1 ENST00000318247.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100018491.hg.1 ENST00000318247.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100018491.hg.1 ENST00000356728.10 Ensembl
          Affymetrix Human Clariom D Assay TC0100018491.hg.1 ENST00000356728.10 Ensembl
          Affymetrix Human Clariom D Assay TC0100018491.hg.1 ENST00000356728.10 Ensembl
          Affymetrix Human Clariom D Assay TC0100018491.hg.1 ENST00000356728.10 Ensembl
          Affymetrix Human Clariom D Assay TC0100018491.hg.1 ENST00000356728.10 Ensembl
          Affymetrix Human Clariom D Assay TC0100018491.hg.1 ENST00000356728.10 Ensembl
          Affymetrix Human Clariom D Assay TC0100018491.hg.1 ENST00000356728.10 Ensembl
          Affymetrix Human Clariom D Assay TC0100018491.hg.1 ENST00000356728.10 Ensembl
          Affymetrix Human Clariom D Assay TC0100018491.hg.1 ENST00000356728.10 Ensembl
          Affymetrix Human Clariom D Assay TC0100018491.hg.1 ENST00000356728.10 Ensembl
          Affymetrix Human Clariom D Assay TC0100018491.hg.1 ENST00000356728.10 Ensembl
          Affymetrix Human Clariom D Assay TC0100018491.hg.1 ENST00000356728.10 Ensembl
          Affymetrix Human Clariom D Assay TC0100018491.hg.1 ENST00000356728.10 Ensembl
          Affymetrix Human Clariom D Assay TC0100018491.hg.1 ENST00000480719.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018491.hg.1 ENST00000480719.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018491.hg.1 ENST00000480719.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018491.hg.1 ENST00000480719.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018491.hg.1 ENST00000480719.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018491.hg.1 ENST00000480719.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018491.hg.1 ENST00000480719.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018491.hg.1 ENST00000480719.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018491.hg.1 ENST00000480719.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018491.hg.1 ENST00000480719.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018491.hg.1 ENST00000480719.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018491.hg.1 ENST00000480719.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018491.hg.1 ENST00000480719.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018491.hg.1 NM_001001523.1 Refseq
          Affymetrix Human Clariom D Assay TC0100018491.hg.1 NM_001001523.1 Refseq
          Affymetrix Human Clariom D Assay TC0100018491.hg.1 NM_001001523.1 Refseq
          Affymetrix Human Clariom D Assay TC0100018491.hg.1 NM_001001523.1 Refseq
          Affymetrix Human Clariom D Assay TC0100018491.hg.1 NM_001001523.1 Refseq
          Affymetrix Human Clariom D Assay TC0100018491.hg.1 NM_001001523.1 Refseq
          Affymetrix Human Clariom D Assay TC0100018491.hg.1 NM_001001523.1 Refseq
          Affymetrix Human Clariom D Assay TC0100018491.hg.1 NM_001001523.1 Refseq
          Affymetrix Human Clariom D Assay TC0100018491.hg.1 NM_001001523.1 Refseq
          Affymetrix Human Clariom D Assay TC0100018491.hg.1 NM_001001523.1 Refseq
          Affymetrix Human Clariom D Assay TC0100018491.hg.1 NM_001001523.1 Refseq
          Affymetrix Human Clariom D Assay TC0100018491.hg.1 NM_001001523.1 Refseq
          Affymetrix Human Clariom D Assay TC0100018491.hg.1 NM_001001523.1 Refseq
          Affymetrix Human Clariom D Assay TC0100018491.hg.1 NM_005060.3 Refseq
          Affymetrix Human Clariom D Assay TC0100018491.hg.1 NM_005060.3 Refseq
          Affymetrix Human Clariom D Assay TC0100018491.hg.1 NM_005060.3 Refseq
          Affymetrix Human Clariom D Assay TC0100018491.hg.1 NM_005060.3 Refseq
          Affymetrix Human Clariom D Assay TC0100018491.hg.1 NM_005060.3 Refseq
          Affymetrix Human Clariom D Assay TC0100018491.hg.1 NM_005060.3 Refseq
          Affymetrix Human Clariom D Assay TC0100018491.hg.1 NM_005060.3 Refseq
          Affymetrix Human Clariom D Assay TC0100018491.hg.1 NM_005060.3 Refseq
          Affymetrix Human Clariom D Assay TC0100018491.hg.1 NM_005060.3 Refseq
          Affymetrix Human Clariom D Assay TC0100018491.hg.1 NM_005060.3 Refseq
          Affymetrix Human Clariom D Assay TC0100018491.hg.1 NM_005060.3 Refseq
          Affymetrix Human Clariom D Assay TC0100018491.hg.1 NM_005060.3 Refseq
          Affymetrix Human Clariom D Assay TC0100018491.hg.1 NM_005060.3 Refseq
          Affymetrix Human Clariom D Assay TC0100018491.hg.1 NM_005060.3 Refseq
          Affymetrix Human Clariom D Assay TC0100018491.hg.1 NM_005060.3 Refseq
          Affymetrix Human Clariom D Assay TC0100018491.hg.1 NM_005060.3 Refseq
          Affymetrix Human Clariom D Assay TC0100018491.hg.1 NM_005060.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100243184.hg.1 ENST00000318247.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0100243184.hg.1 ENST00000356728.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0100243184.hg.1 ENST00000480719.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100243184.hg.1 NM_001001523.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100243184.hg.1 NM_005060.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100243184.hg.1 TC0100018491.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100243184.hg.1 NM_001001523.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100243184.hg.1 ENST00000318247.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0100243184.hg.1 ENST00000356728.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0100243184.hg.1 ENST00000480719.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100243184.hg.1 NM_005060.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-75d45cbfdf-4zq6t:80/100.66.130.62:80.
          git-commit: a3c66eacce7c9fa3045440ddf4a86215532ee526
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix