Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01080040_g1
          See other YES1 GE Assays ›
          Gene Symbol:
          YES1
          Gene Name:
          YES proto-oncogene 1, Src family tyrosine kinase
          Gene Aliases:
          HsT441, P61-YES, Yes, c-yes
          Chromosome Location:
          Chr.18: 721592 - 812326 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01080040_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: YES1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_005433.3 NP_005424.1 10 - 11 1515 90
          XM_017025960.1 XP_016881449.1 10 - 11 1357 90
          GenBank mRNA AK314063.1 - 10 - 11 1459 90
          BC048960.1 - 10 - 11 1511 5260751 90
          M15990.1 - 10 - 11 1501 90

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1800007868.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC18000278.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene YES1

          Gene Ontology Categories:

          Function(s) Process(es)

          cellular protein modification process
          transmembrane receptor protein tyrosine kinase signaling pathway
          cell differentiation
          T cell costimulation
          cellular response to platelet-derived growth factor stimulus
          peptidyl-tyrosine autophosphorylation
          Fc-gamma receptor signaling pathway involved in phagocytosis
          regulation of cell population proliferation
          regulation of vascular permeability
          innate immune response
          positive regulation of transcription by RNA polymerase II
          ephrin receptor signaling pathway
          positive regulation of peptidyl-tyrosine phosphorylation
          leukocyte migration
          cellular response to retinoic acid
          cellular response to transforming growth factor beta stimulus
          glucose transmembrane transport
          protein tyrosine kinase activity
          non-membrane spanning protein tyrosine kinase activity
          signaling receptor binding
          epidermal growth factor receptor binding
          protein binding
          ATP binding
          enzyme binding
          transmembrane transporter binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR18003283.hg.1 ENST00000314574.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR18003283.hg.1 ENST00000359834.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR18003283.hg.1 NM_005433.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR18003283.hg.1 TC18000278.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1800007868.hg.1 ENST00000314574.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007868.hg.1 ENST00000314574.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007868.hg.1 ENST00000314574.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007868.hg.1 ENST00000314574.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007868.hg.1 ENST00000314574.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007868.hg.1 ENST00000314574.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007868.hg.1 ENST00000314574.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007868.hg.1 ENST00000314574.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007868.hg.1 ENST00000314574.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007868.hg.1 ENST00000314574.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007868.hg.1 ENST00000314574.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007868.hg.1 ENST00000314574.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007868.hg.1 ENST00000314574.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007868.hg.1 ENST00000314574.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007868.hg.1 ENST00000577611.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007868.hg.1 ENST00000577611.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007868.hg.1 ENST00000577611.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007868.hg.1 ENST00000577611.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007868.hg.1 ENST00000577611.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007868.hg.1 ENST00000577611.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007868.hg.1 ENST00000577961.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007868.hg.1 ENST00000577961.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007868.hg.1 ENST00000577961.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007868.hg.1 ENST00000577961.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007868.hg.1 ENST00000577961.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007868.hg.1 ENST00000577961.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007868.hg.1 ENST00000577961.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007868.hg.1 ENST00000577961.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007868.hg.1 ENST00000577961.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007868.hg.1 ENST00000577961.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007868.hg.1 ENST00000577961.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007868.hg.1 ENST00000577961.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007868.hg.1 ENST00000577961.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007868.hg.1 ENST00000577961.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007868.hg.1 ENST00000577961.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007868.hg.1 ENST00000581960.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007868.hg.1 ENST00000581960.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007868.hg.1 ENST00000581960.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007868.hg.1 ENST00000581960.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007868.hg.1 ENST00000581960.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007868.hg.1 ENST00000584307.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007868.hg.1 ENST00000584307.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007868.hg.1 ENST00000584307.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007868.hg.1 ENST00000584307.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007868.hg.1 ENST00000584307.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007868.hg.1 ENST00000584307.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007868.hg.1 ENST00000584307.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007868.hg.1 ENST00000584307.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007868.hg.1 ENST00000584307.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007868.hg.1 ENST00000584307.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007868.hg.1 ENST00000584307.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007868.hg.1 ENST00000584307.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007868.hg.1 ENST00000584307.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007868.hg.1 ENST00000584307.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007868.hg.1 ENST00000584307.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800007868.hg.1 NM_005433.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800007868.hg.1 NM_005433.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800007868.hg.1 NM_005433.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800007868.hg.1 NM_005433.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800007868.hg.1 NM_005433.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800007868.hg.1 NM_005433.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800007868.hg.1 NM_005433.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800007868.hg.1 NM_005433.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800007868.hg.1 NM_005433.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800007868.hg.1 NM_005433.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800007868.hg.1 NM_005433.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800007868.hg.1 NM_005433.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800007868.hg.1 NM_005433.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800007868.hg.1 NM_005433.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800007868.hg.1 NM_005433.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800007868.hg.1 NM_005433.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800007868.hg.1 NM_005433.3 Refseq
          Affymetrix Human Clariom D Assay TC1800007868.hg.1 ENST00000314574.4 Ensembl
          Affymetrix Human Clariom D Assay TC1800007868.hg.1 ENST00000314574.4 Ensembl
          Affymetrix Human Clariom D Assay TC1800007868.hg.1 ENST00000314574.4 Ensembl
          Affymetrix Human Clariom D Assay TC1800007868.hg.1 ENST00000314574.4 Ensembl
          Affymetrix Human Clariom D Assay TC1800007868.hg.1 ENST00000314574.4 Ensembl
          Affymetrix Human Clariom D Assay TC1800007868.hg.1 ENST00000314574.4 Ensembl
          Affymetrix Human Clariom D Assay TC1800007868.hg.1 ENST00000314574.4 Ensembl
          Affymetrix Human Clariom D Assay TC1800007868.hg.1 ENST00000314574.4 Ensembl
          Affymetrix Human Clariom D Assay TC1800007868.hg.1 ENST00000314574.4 Ensembl
          Affymetrix Human Clariom D Assay TC1800007868.hg.1 ENST00000314574.4 Ensembl
          Affymetrix Human Clariom D Assay TC1800007868.hg.1 ENST00000314574.4 Ensembl
          Affymetrix Human Clariom D Assay TC1800007868.hg.1 ENST00000314574.4 Ensembl
          Affymetrix Human Clariom D Assay TC1800007868.hg.1 ENST00000314574.4 Ensembl
          Affymetrix Human Clariom D Assay TC1800007868.hg.1 ENST00000314574.4 Ensembl
          Affymetrix Human Clariom D Assay TC1800007868.hg.1 ENST00000577611.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800007868.hg.1 ENST00000577611.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800007868.hg.1 ENST00000577611.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800007868.hg.1 ENST00000577611.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800007868.hg.1 ENST00000577611.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800007868.hg.1 ENST00000577611.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800007868.hg.1 ENST00000577961.5 Ensembl
          Affymetrix Human Clariom D Assay TC1800007868.hg.1 ENST00000577961.5 Ensembl
          Affymetrix Human Clariom D Assay TC1800007868.hg.1 ENST00000577961.5 Ensembl
          Affymetrix Human Clariom D Assay TC1800007868.hg.1 ENST00000577961.5 Ensembl
          Affymetrix Human Clariom D Assay TC1800007868.hg.1 ENST00000577961.5 Ensembl
          Affymetrix Human Clariom D Assay TC1800007868.hg.1 ENST00000577961.5 Ensembl
          Affymetrix Human Clariom D Assay TC1800007868.hg.1 ENST00000577961.5 Ensembl
          Affymetrix Human Clariom D Assay TC1800007868.hg.1 ENST00000577961.5 Ensembl
          Affymetrix Human Clariom D Assay TC1800007868.hg.1 ENST00000577961.5 Ensembl
          Affymetrix Human Clariom D Assay TC1800007868.hg.1 ENST00000577961.5 Ensembl
          Affymetrix Human Clariom D Assay TC1800007868.hg.1 ENST00000577961.5 Ensembl
          Affymetrix Human Clariom D Assay TC1800007868.hg.1 ENST00000577961.5 Ensembl
          Affymetrix Human Clariom D Assay TC1800007868.hg.1 ENST00000577961.5 Ensembl
          Affymetrix Human Clariom D Assay TC1800007868.hg.1 ENST00000577961.5 Ensembl
          Affymetrix Human Clariom D Assay TC1800007868.hg.1 ENST00000577961.5 Ensembl
          Affymetrix Human Clariom D Assay TC1800007868.hg.1 ENST00000581960.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800007868.hg.1 ENST00000581960.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800007868.hg.1 ENST00000581960.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800007868.hg.1 ENST00000581960.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800007868.hg.1 ENST00000581960.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800007868.hg.1 ENST00000584307.5 Ensembl
          Affymetrix Human Clariom D Assay TC1800007868.hg.1 ENST00000584307.5 Ensembl
          Affymetrix Human Clariom D Assay TC1800007868.hg.1 ENST00000584307.5 Ensembl
          Affymetrix Human Clariom D Assay TC1800007868.hg.1 ENST00000584307.5 Ensembl
          Affymetrix Human Clariom D Assay TC1800007868.hg.1 ENST00000584307.5 Ensembl
          Affymetrix Human Clariom D Assay TC1800007868.hg.1 ENST00000584307.5 Ensembl
          Affymetrix Human Clariom D Assay TC1800007868.hg.1 ENST00000584307.5 Ensembl
          Affymetrix Human Clariom D Assay TC1800007868.hg.1 ENST00000584307.5 Ensembl
          Affymetrix Human Clariom D Assay TC1800007868.hg.1 ENST00000584307.5 Ensembl
          Affymetrix Human Clariom D Assay TC1800007868.hg.1 ENST00000584307.5 Ensembl
          Affymetrix Human Clariom D Assay TC1800007868.hg.1 ENST00000584307.5 Ensembl
          Affymetrix Human Clariom D Assay TC1800007868.hg.1 ENST00000584307.5 Ensembl
          Affymetrix Human Clariom D Assay TC1800007868.hg.1 ENST00000584307.5 Ensembl
          Affymetrix Human Clariom D Assay TC1800007868.hg.1 ENST00000584307.5 Ensembl
          Affymetrix Human Clariom D Assay TC1800007868.hg.1 ENST00000584307.5 Ensembl
          Affymetrix Human Clariom D Assay TC1800007868.hg.1 NM_005433.3 Refseq
          Affymetrix Human Clariom D Assay TC1800007868.hg.1 NM_005433.3 Refseq
          Affymetrix Human Clariom D Assay TC1800007868.hg.1 NM_005433.3 Refseq
          Affymetrix Human Clariom D Assay TC1800007868.hg.1 NM_005433.3 Refseq
          Affymetrix Human Clariom D Assay TC1800007868.hg.1 NM_005433.3 Refseq
          Affymetrix Human Clariom D Assay TC1800007868.hg.1 NM_005433.3 Refseq
          Affymetrix Human Clariom D Assay TC1800007868.hg.1 NM_005433.3 Refseq
          Affymetrix Human Clariom D Assay TC1800007868.hg.1 NM_005433.3 Refseq
          Affymetrix Human Clariom D Assay TC1800007868.hg.1 NM_005433.3 Refseq
          Affymetrix Human Clariom D Assay TC1800007868.hg.1 NM_005433.3 Refseq
          Affymetrix Human Clariom D Assay TC1800007868.hg.1 NM_005433.3 Refseq
          Affymetrix Human Clariom D Assay TC1800007868.hg.1 NM_005433.3 Refseq
          Affymetrix Human Clariom D Assay TC1800007868.hg.1 NM_005433.3 Refseq
          Affymetrix Human Clariom D Assay TC1800007868.hg.1 NM_005433.3 Refseq
          Affymetrix Human Clariom D Assay TC1800007868.hg.1 NM_005433.3 Refseq
          Affymetrix Human Clariom D Assay TC1800007868.hg.1 NM_005433.3 Refseq
          Affymetrix Human Clariom D Assay TC1800007868.hg.1 NM_005433.3 Refseq
          Affymetrix Human Clariom D Assay PSR1800153865.hg.1 ENST00000314574.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1800153865.hg.1 ENST00000577961.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1800153865.hg.1 ENST00000584307.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1800153865.hg.1 NM_005433.3 Refseq
          Affymetrix Human Clariom D Assay PSR1800153865.hg.1 TC1800007868.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1800153865.hg.1 NM_005433.3 Refseq
          Affymetrix Human Clariom D Assay PSR1800153865.hg.1 ENST00000314574.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1800153865.hg.1 ENST00000577961.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1800153865.hg.1 ENST00000584307.5 Ensembl

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline