Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09004949.hg.1 |
ENST00000277183.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09004949.hg.1 |
ENST00000375344.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09004949.hg.1 |
ENST00000428393.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09004949.hg.1 |
NM_032558.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09004949.hg.1 |
TC09000463.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008028.hg.1 |
ENST00000375344.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008028.hg.1 |
ENST00000375344.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008028.hg.1 |
ENST00000375344.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008028.hg.1 |
ENST00000375344.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008028.hg.1 |
ENST00000375344.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008028.hg.1 |
ENST00000375344.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008028.hg.1 |
ENST00000375344.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008028.hg.1 |
ENST00000375344.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008028.hg.1 |
ENST00000375344.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008028.hg.1 |
ENST00000375344.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008028.hg.1 |
ENST00000375344.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008028.hg.1 |
ENST00000375344.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008028.hg.1 |
ENST00000375344.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008028.hg.1 |
ENST00000375344.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008028.hg.1 |
ENST00000375344.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008028.hg.1 |
ENST00000375344.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008028.hg.1 |
ENST00000428393.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008028.hg.1 |
ENST00000428393.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008028.hg.1 |
ENST00000428393.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008028.hg.1 |
ENST00000428393.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008028.hg.1 |
ENST00000428393.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008028.hg.1 |
ENST00000428393.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008028.hg.1 |
ENST00000428393.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008028.hg.1 |
NM_032558.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008028.hg.1 |
NM_032558.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008028.hg.1 |
NM_032558.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008028.hg.1 |
NM_032558.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008028.hg.1 |
NM_032558.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008028.hg.1 |
NM_032558.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008028.hg.1 |
NM_032558.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008028.hg.1 |
NM_032558.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008028.hg.1 |
NM_032558.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008028.hg.1 |
NM_032558.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008028.hg.1 |
NM_032558.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008028.hg.1 |
NM_032558.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008028.hg.1 |
NM_032558.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008028.hg.1 |
NM_032558.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008028.hg.1 |
NM_032558.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900154042.hg.1 |
ENST00000375344.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900154042.hg.1 |
ENST00000428393.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900154042.hg.1 |
NM_032558.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900154042.hg.1 |
TC0900008028.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900154042.hg.1 |
ENST00000375344.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900154042.hg.1 |
ENST00000428393.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900154042.hg.1 |
NM_032558.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008028.hg.1 |
ENST00000375344.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008028.hg.1 |
ENST00000375344.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008028.hg.1 |
ENST00000375344.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008028.hg.1 |
ENST00000375344.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008028.hg.1 |
ENST00000375344.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008028.hg.1 |
ENST00000375344.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008028.hg.1 |
ENST00000375344.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008028.hg.1 |
ENST00000375344.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008028.hg.1 |
ENST00000375344.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008028.hg.1 |
ENST00000375344.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008028.hg.1 |
ENST00000375344.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008028.hg.1 |
ENST00000375344.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008028.hg.1 |
ENST00000375344.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008028.hg.1 |
ENST00000375344.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008028.hg.1 |
ENST00000375344.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008028.hg.1 |
ENST00000375344.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008028.hg.1 |
ENST00000428393.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008028.hg.1 |
ENST00000428393.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008028.hg.1 |
ENST00000428393.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008028.hg.1 |
ENST00000428393.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008028.hg.1 |
ENST00000428393.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008028.hg.1 |
ENST00000428393.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008028.hg.1 |
ENST00000428393.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008028.hg.1 |
NM_032558.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008028.hg.1 |
NM_032558.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008028.hg.1 |
NM_032558.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008028.hg.1 |
NM_032558.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008028.hg.1 |
NM_032558.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008028.hg.1 |
NM_032558.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008028.hg.1 |
NM_032558.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008028.hg.1 |
NM_032558.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008028.hg.1 |
NM_032558.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008028.hg.1 |
NM_032558.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008028.hg.1 |
NM_032558.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008028.hg.1 |
NM_032558.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008028.hg.1 |
NM_032558.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008028.hg.1 |
NM_032558.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008028.hg.1 |
NM_032558.2 |
Refseq |