Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05017840.hg.1 |
ENST00000274258.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05017840.hg.1 |
ENST00000425232.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05017840.hg.1 |
ENST00000508244.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05017840.hg.1 |
ENST00000512288.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05017840.hg.1 |
NM_023073.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05017840.hg.1 |
TC05001274.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0500173190.hg.1 |
ENST00000425232.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500173190.hg.1 |
ENST00000508244.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500173190.hg.1 |
ENST00000509849.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500173190.hg.1 |
ENST00000512288.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500173190.hg.1 |
ENST00000514429.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500173190.hg.1 |
NM_023073.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500173190.hg.1 |
TC0500010511.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500173190.hg.1 |
ENST00000509849.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500173190.hg.1 |
ENST00000425232.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500173190.hg.1 |
ENST00000508244.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500173190.hg.1 |
ENST00000512288.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500173190.hg.1 |
ENST00000514429.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500173190.hg.1 |
NM_023073.3 |
Refseq |