Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022949.hg.1 |
ENST00000265663.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022949.hg.1 |
ENST00000339554.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022949.hg.1 |
ENST00000371468.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022949.hg.1 |
ENST00000371472.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022949.hg.1 |
ENST00000371473.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022949.hg.1 |
ENST00000371474.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022949.hg.1 |
ENST00000371475.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022949.hg.1 |
ENST00000371482.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022949.hg.1 |
ENST00000392812.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022949.hg.1 |
ENST00000437259.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022949.hg.1 |
ENST00000484316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022949.hg.1 |
ENST00000541195.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022949.hg.1 |
NM_001130969.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022949.hg.1 |
NM_001130970.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022949.hg.1 |
NM_001130971.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022949.hg.1 |
NM_001178064.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022949.hg.1 |
NM_015537.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022949.hg.1 |
TC09001772.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0900183117.hg.1 |
ENST00000265663.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900183117.hg.1 |
ENST00000339554.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900183117.hg.1 |
ENST00000371468.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900183117.hg.1 |
ENST00000371472.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900183117.hg.1 |
ENST00000371473.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900183117.hg.1 |
ENST00000371474.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900183117.hg.1 |
ENST00000371475.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900183117.hg.1 |
ENST00000371482.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900183117.hg.1 |
ENST00000437259.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900183117.hg.1 |
ENST00000482448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900183117.hg.1 |
ENST00000484316.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900183117.hg.1 |
NM_001130969.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900183117.hg.1 |
NM_001130970.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900183117.hg.1 |
NM_001130971.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900183117.hg.1 |
NM_001178064.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900183117.hg.1 |
NM_015537.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900183117.hg.1 |
TC0900012066.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900183117.hg.1 |
ENST00000265663.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900183117.hg.1 |
ENST00000339554.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900183117.hg.1 |
ENST00000371468.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900183117.hg.1 |
ENST00000371472.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900183117.hg.1 |
ENST00000371473.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900183117.hg.1 |
ENST00000371474.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900183117.hg.1 |
ENST00000371475.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900183117.hg.1 |
ENST00000371482.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900183117.hg.1 |
ENST00000437259.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900183117.hg.1 |
ENST00000482448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900183117.hg.1 |
ENST00000484316.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900183117.hg.1 |
NM_001130969.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900183117.hg.1 |
NM_001130970.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900183117.hg.1 |
NM_001130971.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900183117.hg.1 |
NM_001178064.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900183117.hg.1 |
NM_015537.4 |
Refseq |