Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL16A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001856.3 | NP_001847.3 | 11 - 12 | 1347 | 81 | |
XM_005270481.1 | XP_005270538.1 | 11 - 12 | 1342 | 81 | ||
XM_011540722.1 | XP_011539024.1 | 11 - 12 | 1342 | 81 | ||
XM_011540723.1 | XP_011539025.1 | 11 - 12 | 1342 | 81 | ||
XM_011540724.1 | XP_011539026.1 | 11 - 12 | 1342 | 81 | ||
XM_011540726.2 | XP_011539028.1 | 8 - 9 | 858 | 81 | ||
XM_011540729.1 | XP_011539031.1 | 11 - 12 | 1342 | 81 | ||
XM_011540730.1 | XP_011539032.1 | 11 - 12 | 1342 | 81 | ||
XM_017000338.1 | XP_016855827.1 | 11 - 12 | 1342 | 81 | ||
XM_017000339.1 | XP_016855828.1 | 11 - 12 | 1342 | 81 | ||
GenBank mRNA | AB209571.1 | - | 7 - 8 | 654 | 81 | |
M92642.1 | - | 12 - 13 | 1135 | 81 |
Set Membership: |
![]() ![]() ![]() ![]() |