Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008140.hg.1 |
ENST00000285393.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008140.hg.1 |
ENST00000285393.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008140.hg.1 |
ENST00000285393.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008140.hg.1 |
ENST00000285393.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008140.hg.1 |
ENST00000285393.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008140.hg.1 |
ENST00000285393.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008140.hg.1 |
ENST00000285393.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008140.hg.1 |
ENST00000285393.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008140.hg.1 |
ENST00000285393.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008140.hg.1 |
ENST00000285393.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008140.hg.1 |
ENST00000285393.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008140.hg.1 |
ENST00000521564.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008140.hg.1 |
ENST00000521564.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008140.hg.1 |
ENST00000521564.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008140.hg.1 |
ENST00000521564.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008140.hg.1 |
ENST00000521564.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008140.hg.1 |
ENST00000523635.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008140.hg.1 |
ENST00000523635.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008140.hg.1 |
ENST00000523635.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008140.hg.1 |
ENST00000523635.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008140.hg.1 |
NM_152565.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008140.hg.1 |
NM_152565.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008140.hg.1 |
NM_152565.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008140.hg.1 |
NM_152565.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008140.hg.1 |
NM_152565.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008140.hg.1 |
NM_152565.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008140.hg.1 |
NM_152565.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008140.hg.1 |
NM_152565.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008140.hg.1 |
NM_152565.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008140.hg.1 |
NM_152565.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08007248.hg.1 |
ENST00000285393.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08007248.hg.1 |
NM_152565.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08007248.hg.1 |
TC08000536.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0800155849.hg.1 |
ENST00000285393.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800155849.hg.1 |
NM_152565.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800155849.hg.1 |
TC0800008140.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800155849.hg.1 |
ENST00000285393.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800155849.hg.1 |
NM_152565.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008140.hg.1 |
ENST00000285393.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008140.hg.1 |
ENST00000285393.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008140.hg.1 |
ENST00000285393.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008140.hg.1 |
ENST00000285393.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008140.hg.1 |
ENST00000285393.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008140.hg.1 |
ENST00000285393.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008140.hg.1 |
ENST00000285393.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008140.hg.1 |
ENST00000285393.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008140.hg.1 |
ENST00000285393.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008140.hg.1 |
ENST00000285393.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008140.hg.1 |
ENST00000285393.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008140.hg.1 |
ENST00000521564.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008140.hg.1 |
ENST00000521564.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008140.hg.1 |
ENST00000521564.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008140.hg.1 |
ENST00000521564.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008140.hg.1 |
ENST00000521564.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008140.hg.1 |
ENST00000523635.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008140.hg.1 |
ENST00000523635.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008140.hg.1 |
ENST00000523635.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008140.hg.1 |
ENST00000523635.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008140.hg.1 |
NM_152565.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008140.hg.1 |
NM_152565.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008140.hg.1 |
NM_152565.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008140.hg.1 |
NM_152565.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008140.hg.1 |
NM_152565.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008140.hg.1 |
NM_152565.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008140.hg.1 |
NM_152565.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008140.hg.1 |
NM_152565.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008140.hg.1 |
NM_152565.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008140.hg.1 |
NM_152565.1 |
Refseq |