Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01084904_mH
          See other SLC4A10 GE Assays ›
          Gene Symbol:
          SLC4A10
          Gene Name:
          solute carrier family 4 member 10
          Gene Aliases:
          NBCn2, NCBE
          Chromosome Location:
          Chr.2: 161624335 - 161985276 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01084904_mH
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SLC4A10
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001178015.1 NP_001171486.1 21 - 22 3044 90
          NM_001178016.1 NP_001171487.1 21 - 22 3090 90
          NM_022058.3 NP_071341.2 20 - 21 2954 90
          XM_005246694.4 XP_005246751.1 22 - 23 3099 90
          XM_005246695.4 XP_005246752.1 21 - 22 3009 90
          XM_011511508.2 XP_011509810.1 21 - 22 2887 90
          XM_011511509.2 XP_011509811.1 21 - 22 2884 90
          XM_011511510.2 XP_011509812.1 21 - 22 2887 90
          XM_011511511.2 XP_011509813.1 20 - 21 2797 90
          XM_011511512.2 XP_011509814.1 20 - 21 2794 90
          XM_011511513.2 XP_011509815.1 24 - 25 3376 90
          XM_017004541.1 XP_016860030.1 21 - 22 2884 90
          XM_017004542.1 XP_016860031.1 20 - 21 2794 90
          XM_017004543.1 XP_016860032.1 20 - 21 2797 90
          XM_017004544.1 XP_016860033.1 20 - 21 2794 90
          XM_017004545.1 XP_016860034.1 23 - 24 3285 90
          XM_017004546.1 XP_016860035.1 23 - 24 3286 90
          XM_017004547.1 XP_016860036.1 22 - 23 3099 90
          XM_017004548.1 XP_016860037.1 22 - 23 3073 90
          XM_017004549.1 XP_016860038.1 22 - 23 3161 90
          XM_017004550.1 XP_016860039.1 21 - 22 3006 90
          XM_017004551.1 XP_016860040.1 17 - 18 2382 90
          XM_017004552.1 XP_016860041.1 21 - 22 2903 90
          XM_017004553.1 XP_016860042.1 21 - 22 2903 90
          XM_017004554.1 XP_016860043.1 11 - 12 2086 90
          GenBank mRNA AB040457.1 - 20 - 21 2862 90
          AK293793.1 - 21 - 22 3003 90
          AL713680.1 - 2 - 3 174 90
          AL832525.1 - 2 - 3 174 90
          AY376402.1 - 21 - 22 2860 90
          BC136269.1 - 20 - 21 2840 9020256 90
          BC143714.1 - 21 - 22 2930 9052228 90

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0200016556.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC02004999.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene SLC4A10

          Gene Ontology Categories:

          Function(s) Process(es)

          sodium ion transport
          chloride transport
          nervous system development
          response to light stimulus
          post-embryonic development
          bicarbonate transport
          pyramidal neuron development
          multicellular organism growth
          locomotory exploration behavior
          brain morphogenesis
          regulation of intracellular pH
          anion transmembrane transport
          proton transmembrane transport
          inorganic anion exchanger activity
          anion transmembrane transporter activity
          symporter activity
          anion:anion antiporter activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR02014328.hg.1 ENST00000272716.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02014328.hg.1 ENST00000375514.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02014328.hg.1 ENST00000415711.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02014328.hg.1 ENST00000415876.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02014328.hg.1 ENST00000421911.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02014328.hg.1 ENST00000446997.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02014328.hg.1 ENST00000449513.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02014328.hg.1 NM_001178015.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02014328.hg.1 NM_001178016.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02014328.hg.1 NM_022058.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02014328.hg.1 TC02004999.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0200223924.hg.1 ENST00000272716.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0200223924.hg.1 ENST00000375514.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0200223924.hg.1 ENST00000415876.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0200223924.hg.1 ENST00000421911.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200223924.hg.1 ENST00000446228.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200223924.hg.1 ENST00000446997.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200223924.hg.1 NM_001178015.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200223924.hg.1 NM_001178016.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200223924.hg.1 NM_022058.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200223924.hg.1 TC0200016556.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200223924.hg.1 ENST00000272716.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0200223924.hg.1 ENST00000375514.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0200223924.hg.1 ENST00000415876.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0200223924.hg.1 ENST00000421911.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200223924.hg.1 ENST00000446228.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200223924.hg.1 ENST00000446997.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200223924.hg.1 NM_001178015.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200223924.hg.1 NM_001178016.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200223924.hg.1 NM_022058.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6659c69d8f-ztd5s:80/100.66.134.6:80.
          git-commit: 1827ea18100b76eacea45a9906e00cdefce2d263
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0