Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01085998_m1
          See other TGFB3 GE Assays ›
          Gene Symbol:
          TGFB3
          Gene Name:
          transforming growth factor beta 3
          Gene Aliases:
          ARVD, ARVD1, LDS5, RNHF, TGF-beta3
          Chromosome Location:
          Chr.14: 75958097 - 75982022 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01085998_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TGFB3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_003239.3 NP_003230.1 4 - 5 1879 82
          GenBank mRNA AK313564.1 - 4 - 5 960 82
          BC018503.1 - 4 - 5 850 4477175 82
          BT007287.1 - 4 - 5 750 82
          EF560714.1 - 4 - 5 863 82
          J03241.1 - 4 - 5 1012 82
          X14149.1 - 4 - 5 1003 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1400009738.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC14001326.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene TGFB3

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          in utero embryonic development
          platelet degranulation
          transforming growth factor beta receptor signaling pathway
          salivary gland morphogenesis
          female pregnancy
          aging
          negative regulation of DNA replication
          negative regulation of cell population proliferation
          positive regulation of epithelial to mesenchymal transition
          positive regulation of pathway-restricted SMAD protein phosphorylation
          negative regulation of macrophage cytokine production
          cell growth
          positive regulation of bone mineralization
          negative regulation of transforming growth factor beta receptor signaling pathway
          mammary gland development
          response to progesterone
          positive regulation of collagen biosynthetic process
          response to laminar fluid shear stress
          wound healing
          regulation of cell population proliferation
          odontogenesis
          uterine wall breakdown
          regulation of apoptotic process
          positive regulation of apoptotic process
          regulation of MAPK cascade
          negative regulation of neuron apoptotic process
          response to estrogen
          ossification involved in bone remodeling
          cell-cell junction organization
          positive regulation of DNA replication
          positive regulation of transcription, DNA-templated
          positive regulation of transcription by RNA polymerase II
          lung alveolus development
          digestive tract development
          embryonic neurocranium morphogenesis
          inner ear development
          positive regulation of protein secretion
          positive regulation of filopodium assembly
          positive regulation of stress fiber assembly
          positive regulation of cell division
          roof of mouth development
          face morphogenesis
          frontal suture morphogenesis
          regulation of SMAD protein signal transduction
          positive regulation of SMAD protein signal transduction
          SMAD protein signal transduction
          detection of hypoxia
          regulation of epithelial to mesenchymal transition involved in endocardial cushion formation
          positive regulation of tight junction disassembly
          type II transforming growth factor beta receptor binding
          cytokine activity
          protein binding
          growth factor activity
          type I transforming growth factor beta receptor binding
          type III transforming growth factor beta receptor binding
          identical protein binding
          protein heterodimerization activity
          transforming growth factor beta binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR14015136.hg.1 ENST00000238682.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14015136.hg.1 NM_003239.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR14015136.hg.1 TC14001326.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000238682.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000238682.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000238682.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000238682.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000238682.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000238682.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000238682.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000238682.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000238682.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000238682.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000238682.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000238682.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000238682.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000238682.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000554980.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000554980.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000554980.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000554980.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000554980.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000554980.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000554980.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000554980.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000554980.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000555193.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000555193.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000555193.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000555193.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000556285.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000556285.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000556285.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000556285.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000556285.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000556285.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000556285.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000556285.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000556507.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000556507.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000556507.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000556674.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000556674.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000556674.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000556674.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000557493.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000557493.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000557493.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000557493.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000557493.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 ENST00000557493.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 NM_003239.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 NM_003239.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 NM_003239.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 NM_003239.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 NM_003239.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 NM_003239.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 NM_003239.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 NM_003239.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 NM_003239.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 NM_003239.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 NM_003239.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 NM_003239.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 NM_003239.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 NM_003239.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 NM_003239.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 NM_003239.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 NM_003239.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 NM_003239.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400009738.hg.1 NM_003239.4 Refseq
          Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000238682.7 Ensembl
          Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000238682.7 Ensembl
          Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000238682.7 Ensembl
          Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000238682.7 Ensembl
          Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000238682.7 Ensembl
          Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000238682.7 Ensembl
          Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000238682.7 Ensembl
          Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000238682.7 Ensembl
          Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000238682.7 Ensembl
          Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000238682.7 Ensembl
          Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000238682.7 Ensembl
          Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000238682.7 Ensembl
          Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000238682.7 Ensembl
          Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000238682.7 Ensembl
          Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000554980.5 Ensembl
          Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000554980.5 Ensembl
          Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000554980.5 Ensembl
          Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000554980.5 Ensembl
          Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000554980.5 Ensembl
          Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000554980.5 Ensembl
          Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000554980.5 Ensembl
          Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000554980.5 Ensembl
          Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000554980.5 Ensembl
          Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000555193.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000555193.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000555193.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000555193.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000556285.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000556285.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000556285.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000556285.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000556285.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000556285.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000556285.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000556285.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000556507.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000556507.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000556507.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000556674.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000556674.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000556674.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000556674.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000557493.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000557493.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000557493.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000557493.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000557493.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400009738.hg.1 ENST00000557493.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400009738.hg.1 NM_003239.4 Refseq
          Affymetrix Human Clariom D Assay TC1400009738.hg.1 NM_003239.4 Refseq
          Affymetrix Human Clariom D Assay TC1400009738.hg.1 NM_003239.4 Refseq
          Affymetrix Human Clariom D Assay TC1400009738.hg.1 NM_003239.4 Refseq
          Affymetrix Human Clariom D Assay TC1400009738.hg.1 NM_003239.4 Refseq
          Affymetrix Human Clariom D Assay TC1400009738.hg.1 NM_003239.4 Refseq
          Affymetrix Human Clariom D Assay TC1400009738.hg.1 NM_003239.4 Refseq
          Affymetrix Human Clariom D Assay TC1400009738.hg.1 NM_003239.4 Refseq
          Affymetrix Human Clariom D Assay TC1400009738.hg.1 NM_003239.4 Refseq
          Affymetrix Human Clariom D Assay TC1400009738.hg.1 NM_003239.4 Refseq
          Affymetrix Human Clariom D Assay TC1400009738.hg.1 NM_003239.4 Refseq
          Affymetrix Human Clariom D Assay TC1400009738.hg.1 NM_003239.4 Refseq
          Affymetrix Human Clariom D Assay TC1400009738.hg.1 NM_003239.4 Refseq
          Affymetrix Human Clariom D Assay TC1400009738.hg.1 NM_003239.4 Refseq
          Affymetrix Human Clariom D Assay TC1400009738.hg.1 NM_003239.4 Refseq
          Affymetrix Human Clariom D Assay TC1400009738.hg.1 NM_003239.4 Refseq
          Affymetrix Human Clariom D Assay TC1400009738.hg.1 NM_003239.4 Refseq
          Affymetrix Human Clariom D Assay TC1400009738.hg.1 NM_003239.4 Refseq
          Affymetrix Human Clariom D Assay TC1400009738.hg.1 NM_003239.4 Refseq
          Affymetrix Human Clariom D Assay PSR1400171871.hg.1 ENST00000238682.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1400171871.hg.1 ENST00000554980.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1400171871.hg.1 ENST00000556285.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1400171871.hg.1 ENST00000557493.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1400171871.hg.1 NM_003239.4 Refseq
          Affymetrix Human Clariom D Assay PSR1400171871.hg.1 TC1400009738.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1400171871.hg.1 ENST00000557493.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1400171871.hg.1 ENST00000238682.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1400171871.hg.1 ENST00000554980.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1400171871.hg.1 ENST00000556285.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1400171871.hg.1 NM_003239.4 Refseq
          Affymetrix Human Clariom D Assay PSR1400171870.hg.1 ENST00000556285.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1400171870.hg.1 TC1400009738.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1400171870.hg.1 ENST00000556285.1 Ensembl

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0