Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19007803.hg.1 |
ENST00000263370.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19007803.hg.1 |
NM_025194.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19007803.hg.1 |
TC19000566.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008118.hg.1 |
ENST00000263370.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008118.hg.1 |
ENST00000263370.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008118.hg.1 |
ENST00000263370.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008118.hg.1 |
ENST00000263370.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008118.hg.1 |
ENST00000263370.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008118.hg.1 |
ENST00000263370.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008118.hg.1 |
ENST00000263370.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008118.hg.1 |
ENST00000263370.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008118.hg.1 |
ENST00000263370.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008118.hg.1 |
ENST00000263370.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008118.hg.1 |
ENST00000263370.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008118.hg.1 |
ENST00000263370.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008118.hg.1 |
ENST00000263370.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008118.hg.1 |
ENST00000597003.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008118.hg.1 |
ENST00000597003.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008118.hg.1 |
ENST00000597003.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008118.hg.1 |
ENST00000597003.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008118.hg.1 |
ENST00000597003.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008118.hg.1 |
ENST00000597003.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008118.hg.1 |
NM_025194.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008118.hg.1 |
NM_025194.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008118.hg.1 |
NM_025194.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008118.hg.1 |
NM_025194.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008118.hg.1 |
NM_025194.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008118.hg.1 |
NM_025194.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008118.hg.1 |
NM_025194.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008118.hg.1 |
NM_025194.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008118.hg.1 |
NM_025194.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008118.hg.1 |
NM_025194.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008118.hg.1 |
NM_025194.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008118.hg.1 |
NM_025194.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008118.hg.1 |
NM_025194.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19007804.hg.1 |
ENST00000263370.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19007804.hg.1 |
NM_025194.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19007804.hg.1 |
TC19000566.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1900161443.hg.1 |
ENST00000263370.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900161443.hg.1 |
ENST00000597003.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900161443.hg.1 |
NM_025194.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900161443.hg.1 |
TC1900008118.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900161443.hg.1 |
ENST00000263370.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900161443.hg.1 |
ENST00000597003.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900161443.hg.1 |
NM_025194.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900161446.hg.1 |
ENST00000263370.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900161446.hg.1 |
ENST00000597003.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900161446.hg.1 |
NM_025194.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900161446.hg.1 |
TC1900008118.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900161446.hg.1 |
ENST00000263370.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900161446.hg.1 |
ENST00000597003.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900161446.hg.1 |
NM_025194.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008118.hg.1 |
ENST00000263370.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008118.hg.1 |
ENST00000263370.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008118.hg.1 |
ENST00000263370.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008118.hg.1 |
ENST00000263370.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008118.hg.1 |
ENST00000263370.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008118.hg.1 |
ENST00000263370.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008118.hg.1 |
ENST00000263370.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008118.hg.1 |
ENST00000263370.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008118.hg.1 |
ENST00000263370.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008118.hg.1 |
ENST00000263370.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008118.hg.1 |
ENST00000263370.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008118.hg.1 |
ENST00000263370.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008118.hg.1 |
ENST00000263370.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008118.hg.1 |
ENST00000597003.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008118.hg.1 |
ENST00000597003.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008118.hg.1 |
ENST00000597003.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008118.hg.1 |
ENST00000597003.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008118.hg.1 |
ENST00000597003.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008118.hg.1 |
ENST00000597003.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008118.hg.1 |
NM_025194.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008118.hg.1 |
NM_025194.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008118.hg.1 |
NM_025194.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008118.hg.1 |
NM_025194.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008118.hg.1 |
NM_025194.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008118.hg.1 |
NM_025194.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008118.hg.1 |
NM_025194.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008118.hg.1 |
NM_025194.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008118.hg.1 |
NM_025194.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008118.hg.1 |
NM_025194.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008118.hg.1 |
NM_025194.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008118.hg.1 |
NM_025194.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008118.hg.1 |
NM_025194.2 |
Refseq |