Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01008323.hg.1 |
ENST00000304979.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01008323.hg.1 |
NM_006347.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01008323.hg.1 |
TC01000529.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01008322.hg.1 |
NM_006347.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01008322.hg.1 |
TC01000529.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012311.hg.1 |
ENST00000428428.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01008324.hg.1 |
ENST00000304979.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01008324.hg.1 |
ENST00000372550.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01008324.hg.1 |
NM_006347.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01008324.hg.1 |
TC01000529.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008023.hg.1 |
ENST00000461351.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008023.hg.1 |
ENST00000461351.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008023.hg.1 |
NM_006347.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008023.hg.1 |
NM_006347.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008023.hg.1 |
NM_006347.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008023.hg.1 |
NM_006347.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008023.hg.1 |
NM_006347.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008023.hg.1 |
NM_006347.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008023.hg.1 |
NM_006347.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008023.hg.1 |
NM_006347.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008023.hg.1 |
NM_006347.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008023.hg.1 |
NM_006347.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008023.hg.1 |
ENST00000304979.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008023.hg.1 |
ENST00000304979.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008023.hg.1 |
ENST00000304979.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008023.hg.1 |
ENST00000304979.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008023.hg.1 |
ENST00000304979.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008023.hg.1 |
ENST00000304979.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008023.hg.1 |
ENST00000304979.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008023.hg.1 |
ENST00000304979.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008023.hg.1 |
ENST00000304979.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008023.hg.1 |
ENST00000372549.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008023.hg.1 |
ENST00000372549.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008023.hg.1 |
ENST00000372549.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008023.hg.1 |
ENST00000372550.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008023.hg.1 |
ENST00000372550.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008023.hg.1 |
ENST00000372550.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008023.hg.1 |
ENST00000372550.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008023.hg.1 |
ENST00000372550.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008023.hg.1 |
ENST00000372550.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008023.hg.1 |
ENST00000372550.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008023.hg.1 |
ENST00000372550.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008023.hg.1 |
ENST00000372550.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008023.hg.1 |
ENST00000436387.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008023.hg.1 |
ENST00000436387.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008023.hg.1 |
ENST00000436387.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008023.hg.1 |
ENST00000436387.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008023.hg.1 |
ENST00000436387.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008023.hg.1 |
ENST00000440068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008023.hg.1 |
ENST00000440068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008023.hg.1 |
ENST00000440068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008023.hg.1 |
ENST00000440068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008023.hg.1 |
ENST00000440068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008023.hg.1 |
ENST00000440068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008023.hg.1 |
ENST00000440068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008023.hg.1 |
ENST00000440068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008023.hg.1 |
ENST00000461351.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008023.hg.1 |
ENST00000461351.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008023.hg.1 |
ENST00000461351.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008023.hg.1 |
ENST00000461351.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01008328.hg.1 |
ENST00000304979.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01008328.hg.1 |
ENST00000372549.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01008328.hg.1 |
ENST00000372550.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01008328.hg.1 |
NM_006347.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01008328.hg.1 |
TC01000529.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008161.hg.1 |
ENST00000403700.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100156533.hg.1 |
NM_006347.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100156533.hg.1 |
TC0100008023.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100156533.hg.1 |
NM_006347.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008023.hg.1 |
ENST00000461351.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008023.hg.1 |
ENST00000461351.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008023.hg.1 |
NM_006347.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008023.hg.1 |
NM_006347.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008023.hg.1 |
NM_006347.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008023.hg.1 |
NM_006347.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008023.hg.1 |
NM_006347.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008023.hg.1 |
NM_006347.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008023.hg.1 |
NM_006347.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008023.hg.1 |
NM_006347.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008023.hg.1 |
NM_006347.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008023.hg.1 |
NM_006347.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008023.hg.1 |
ENST00000304979.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008023.hg.1 |
ENST00000304979.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008023.hg.1 |
ENST00000304979.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008023.hg.1 |
ENST00000304979.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008023.hg.1 |
ENST00000304979.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008023.hg.1 |
ENST00000304979.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008023.hg.1 |
ENST00000304979.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008023.hg.1 |
ENST00000304979.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008023.hg.1 |
ENST00000304979.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008023.hg.1 |
ENST00000372549.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008023.hg.1 |
ENST00000372549.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008023.hg.1 |
ENST00000372549.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008023.hg.1 |
ENST00000372550.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008023.hg.1 |
ENST00000372550.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008023.hg.1 |
ENST00000372550.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008023.hg.1 |
ENST00000372550.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008023.hg.1 |
ENST00000372550.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008023.hg.1 |
ENST00000372550.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008023.hg.1 |
ENST00000372550.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008023.hg.1 |
ENST00000372550.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008023.hg.1 |
ENST00000372550.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008023.hg.1 |
ENST00000436387.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008023.hg.1 |
ENST00000436387.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008023.hg.1 |
ENST00000436387.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008023.hg.1 |
ENST00000436387.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008023.hg.1 |
ENST00000436387.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008023.hg.1 |
ENST00000440068.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008023.hg.1 |
ENST00000440068.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008023.hg.1 |
ENST00000440068.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008023.hg.1 |
ENST00000440068.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008023.hg.1 |
ENST00000440068.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008023.hg.1 |
ENST00000440068.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008023.hg.1 |
ENST00000440068.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008023.hg.1 |
ENST00000440068.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008023.hg.1 |
ENST00000461351.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008023.hg.1 |
ENST00000461351.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008023.hg.1 |
ENST00000461351.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008023.hg.1 |
ENST00000461351.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008161.hg.1 |
ENST00000403700.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100156541.hg.1 |
ENST00000304979.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100156541.hg.1 |
ENST00000372549.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100156541.hg.1 |
ENST00000372550.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100156541.hg.1 |
ENST00000440068.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100156541.hg.1 |
NM_006347.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100156541.hg.1 |
TC0100008023.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100156541.hg.1 |
ENST00000304979.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100156541.hg.1 |
ENST00000372549.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100156541.hg.1 |
ENST00000372550.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100156541.hg.1 |
ENST00000440068.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100156541.hg.1 |
NM_006347.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012311.hg.1 |
ENST00000428428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100156536.hg.1 |
ENST00000304979.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100156536.hg.1 |
ENST00000372550.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100156536.hg.1 |
NM_006347.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100156536.hg.1 |
TC0100008023.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100156536.hg.1 |
ENST00000304979.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100156536.hg.1 |
ENST00000372550.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100156536.hg.1 |
NM_006347.3 |
Refseq |