Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01088361_m1
          See other ANK3 GE Assays ›
          Gene Symbol:
          ANK3
          Gene Name:
          ankyrin 3, node of Ranvier (ankyrin G)
          Gene Aliases:
          ANKYRIN-G, MRT37
          Chromosome Location:
          Chr.10: 60026298 - 60733526 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01088361_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ANK3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001149.3 NP_001140.2 14 - 15 2537 149
          NM_001204403.1 NP_001191332.1 37 - 38 4675 149
          NM_001204404.1 NP_001191333.1 37 - 38 4918 149
          NM_001320874.1 NP_001307803.1 36 - 37 4932 149
          NM_020987.4 NP_066267.2 38 - 39 12771 149
          XM_005269715.3 XP_005269772.1 37 - 38 4819 149
          XM_006717796.3 XP_006717859.1 40 - 41 7441 149
          XM_006717802.3 XP_006717865.1 39 - 40 4858 149
          XM_011539708.2 XP_011538010.1 40 - 41 7441 149
          XM_011539709.2 XP_011538011.1 40 - 41 7441 149
          XM_017016110.1 XP_016871599.1 40 - 41 12676 149
          XM_017016111.1 XP_016871600.1 39 - 40 12664 149
          XM_017016112.1 XP_016871601.1 40 - 41 12661 149
          XM_017016113.1 XP_016871602.1 39 - 40 12649 149
          XM_017016114.1 XP_016871603.1 40 - 41 12775 149
          XM_017016115.1 XP_016871604.1 40 - 41 12505 149
          XM_017016116.1 XP_016871605.1 40 - 41 12676 149
          XM_017016117.1 XP_016871606.1 40 - 41 12676 149
          XM_017016118.1 XP_016871607.1 40 - 41 11335 149
          XM_017016119.1 XP_016871608.1 40 - 41 11335 149
          XM_017016120.1 XP_016871609.1 40 - 41 11335 149
          XM_017016121.1 XP_016871610.1 39 - 40 11308 149
          XM_017016122.1 XP_016871611.1 37 - 38 7348 149
          XM_017016123.1 XP_016871612.1 40 - 41 7441 149
          XM_017016124.1 XP_016871613.1 39 - 40 7414 149
          XM_017016125.1 XP_016871614.1 39 - 40 7414 149
          XM_017016126.1 XP_016871615.1 38 - 39 7309 149
          XM_017016127.1 XP_016871616.1 37 - 38 7282 149
          XM_017016128.1 XP_016871617.1 38 - 39 4831 149
          XM_017016129.1 XP_016871618.1 38 - 39 4831 149
          XM_017016130.1 XP_016871619.1 36 - 37 4765 149
          XM_017016131.1 XP_016871620.1 36 - 37 4750 149
          XM_017016132.1 XP_016871621.1 36 - 37 4729 149
          XM_017016133.1 XP_016871622.1 36 - 37 4864 149
          XM_017016134.1 XP_016871623.1 36 - 37 4699 149
          XM_017016135.1 XP_016871624.1 36 - 37 4828 149
          XM_017016136.1 XP_016871625.1 39 - 40 4858 149
          XM_017016137.1 XP_016871626.1 38 - 39 4831 149
          XM_017016138.1 XP_016871627.1 38 - 39 4831 149
          XM_017016139.1 XP_016871628.1 36 - 37 4765 149
          XM_017016140.1 XP_016871629.1 36 - 37 4765 149
          XM_017016141.1 XP_016871630.1 37 - 38 4726 149
          XM_017016142.1 XP_016871631.1 36 - 37 4714 149
          XM_017016143.1 XP_016871632.1 36 - 37 4699 149
          XM_017016144.1 XP_016871633.1 36 - 37 4699 149
          XM_017016145.1 XP_016871634.1 35 - 36 4687 149
          XM_017016146.1 XP_016871635.1 35 - 36 4687 149
          XM_017016147.1 XP_016871636.1 36 - 37 4678 149
          XM_017016148.1 XP_016871637.1 36 - 37 4581 149
          XM_017016149.1 XP_016871638.1 35 - 36 4786 149
          XM_017016150.1 XP_016871639.1 38 - 39 12610 149
          GenBank mRNA AK092361.1 - 4 - 5 262 149
          AK126851.1 - 18 - 19 2620 149
          AK292927.1 - 14 - 15 2423 149
          AK295661.1 - 37 - 38 4625 149
          AL136710.1 - 14 - 15 2196 149
          BX537917.1 - 37 - 38 4546 149
          BX648574.1 - 37 - 38 4640 149
          U13616.1 - 38 - 39 12517 149

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1000010727.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC10001319.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene ANK3

          Gene Ontology Categories:

          Function(s) Process(es)

          mitotic cytokinesis
          endoplasmic reticulum to Golgi vesicle-mediated transport
          plasma membrane organization
          signal transduction
          axonogenesis
          axon guidance
          neuromuscular junction development
          positive regulation of gene expression
          positive regulation of cell communication by electrical coupling
          positive regulation of sodium ion transport
          magnesium ion homeostasis
          neuronal action potential
          positive regulation of homotypic cell-cell adhesion
          Golgi to plasma membrane protein transport
          regulation of potassium ion transport
          establishment of protein localization
          positive regulation of membrane potential
          cellular response to magnesium ion
          membrane assembly
          protein localization to plasma membrane
          maintenance of protein location in plasma membrane
          positive regulation of protein targeting to membrane
          positive regulation of membrane depolarization during cardiac muscle cell action potential
          negative regulation of delayed rectifier potassium channel activity
          positive regulation of sodium ion transmembrane transporter activity
          positive regulation of cation channel activity
          structural constituent of cytoskeleton
          protein binding
          cytoskeletal protein binding
          cytoskeletal anchor activity
          spectrin binding
          protein-macromolecule adaptor activity
          transmembrane transporter binding
          cadherin binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR10016563.hg.1 ENST00000280772.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10016563.hg.1 ENST00000355288.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10016563.hg.1 ENST00000373817.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10016563.hg.1 ENST00000373827.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10016563.hg.1 ENST00000395299.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10016563.hg.1 ENST00000423532.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10016563.hg.1 ENST00000503366.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10016563.hg.1 NM_001149.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10016563.hg.1 NM_001204403.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10016563.hg.1 NM_001204404.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10016563.hg.1 NM_020987.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10016563.hg.1 TC10001319.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1000173162.hg.1 ENST00000280772.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1000173162.hg.1 ENST00000355288.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1000173162.hg.1 ENST00000373820.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1000173162.hg.1 ENST00000373827.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1000173162.hg.1 ENST00000459732.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1000173162.hg.1 ENST00000503366.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1000173162.hg.1 ENST00000511043.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1000173162.hg.1 ENST00000514197.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1000173162.hg.1 ENST00000610321.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1000173162.hg.1 ENST00000610901.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1000173162.hg.1 ENST00000613207.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1000173162.hg.1 ENST00000616444.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1000173162.hg.1 ENST00000618374.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1000173162.hg.1 ENST00000621739.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000173162.hg.1 NM_001149.3 Refseq
          Affymetrix Human Clariom D Assay PSR1000173162.hg.1 NM_001204403.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000173162.hg.1 NM_001204404.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000173162.hg.1 NM_020987.4 Refseq
          Affymetrix Human Clariom D Assay PSR1000173162.hg.1 TC1000010727.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000173162.hg.1 ENST00000280772.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1000173162.hg.1 ENST00000355288.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1000173162.hg.1 ENST00000373820.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1000173162.hg.1 ENST00000373827.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1000173162.hg.1 ENST00000459732.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1000173162.hg.1 ENST00000503366.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1000173162.hg.1 ENST00000511043.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1000173162.hg.1 ENST00000514197.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1000173162.hg.1 ENST00000610321.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1000173162.hg.1 ENST00000610901.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1000173162.hg.1 ENST00000613207.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1000173162.hg.1 ENST00000616444.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1000173162.hg.1 ENST00000618374.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1000173162.hg.1 ENST00000621739.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000173162.hg.1 NM_001149.3 Refseq
          Affymetrix Human Clariom D Assay PSR1000173162.hg.1 NM_001204403.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000173162.hg.1 NM_001204404.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000173162.hg.1 NM_020987.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline