Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01088632_m1
          See other ISG20 GE Assays ›
          Gene Symbol:
          ISG20
          Gene Name:
          interferon stimulated exonuclease gene 20
          Gene Aliases:
          CD25, HEM45
          Chromosome Location:
          Chr.15: 88635614 - 88656344 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01088632_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ISG20
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001303233.1 NP_001290162.1 3 - 4 523 76
          NM_001303234.1 NP_001290163.1 4 - 5 649 76
          NM_001303235.1 NP_001290164.1 2 - 3 2146 76
          NM_001303236.1 NP_001290165.1 3 - 4 2770 76
          NM_001303237.1 NP_001290166.1 3 - 4 733 76
          NM_002201.5 NP_002192.2 3 - 4 793 76
          NR_130134.1 2 - 3 271 76
          XM_005254899.2 XP_005254956.1 3 - 4 2056 76
          XM_006720488.3 XP_006720551.1 5 - 6 834 76
          XM_011521521.2 XP_011519823.1 2 - 3 1216 76
          XM_017022147.1 XP_016877636.1 3 - 4 3857 76
          XM_017022148.1 XP_016877637.1 4 - 5 612 76
          GenBank mRNA AB209639.1 - 1 - 2 5405 76
          AK122793.1 - 2 - 3 2110 76
          AK307769.1 - 3 - 4 591 76
          AV737297.1 - 2 - 3 293 76
          BC007922.2 - 3 - 4 728 4309120 76
          BC016341.1 - 3 - 4 2471 76
          BI838264.1 - 4 - 5 602 76
          BM691295.1 - 3 - 4 531 76
          BT006952.1 - 2 - 3 435 76
          BX339191.2 - 3 - 4 731 76
          CN265233.1 - 3 - 4 523 76
          CR456942.1 - 2 - 3 435 76
          U88964.1 - 2 - 3 472 76
          X89773.2 - 2 - 3 450 76

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene ISG20

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA catabolic process, exonucleolytic
          rRNA processing
          RNA catabolic process
          cell population proliferation
          response to virus
          negative regulation of viral genome replication
          defense response to virus
          type I interferon signaling pathway
          RNA phosphodiester bond hydrolysis, exonucleolytic
          3'-5'-exoribonuclease activity
          exonuclease activity
          single-stranded DNA 3'-5' exodeoxyribonuclease activity
          exoribonuclease II activity
          U1 snRNA binding
          U2 snRNA binding
          U3 snoRNA binding
          metal ion binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0