Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01088862_s1
          See other AGPAT4-IT1 GE Assays ›
          Gene Symbol:
          AGPAT4-IT1
          Gene Name:
          AGPAT4 intronic transcript 1
          Gene Aliases:
          C6orf59, NCRNA00241
          Chromosome Location:
          Chr.6: 161160114 - 161161982 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01088862_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: AGPAT4-IT1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_024277.1 1 1628 102
          GenBank mRNA AK026765.1 - 1 1637 102
          AL136708.1 - 1 1482 102
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BP250919.1 - 1 223 Chr. 6 - 161160015 - 161160577 [-] on Build GRCh38 102
          GenBankmRNA AW081591.1 - 1 Chr. 6 - 161160134 - 161160514 [+] on Build GRCh38 102
          GenBankmRNA AI937612.1 - 1 Chr. 6 - 161160115 - 161160499 [+] on Build GRCh38 102
          GenBankmRNA HY321190.1 - 1 Chr. 6 - 161160142 - 161160574 [+] on Build GRCh38 102
          GenBankmRNA HY348701.1 - 1 Chr. 6 - 161160142 - 161160568 [+] on Build GRCh38 102
          GenBankmRNA BP395531.1 - 1 143 Chr. 6 - 161160131 - 161160497 [-] on Build GRCh38 102
          GenBankmRNA AL566739.3 - 1 Chr. 6 - 161160179 - 161161178 [+] on Build GRCh38 102
          GenBankmRNA BE245723.1 - 1 Chr. 6 - 161160142 - 161160482 [+] on Build GRCh38 102
          GenBankmRNA HY322435.1 - 1 Chr. 6 - 161160142 - 161160602 [+] on Build GRCh38 102
          GenBankmRNA BP250941.1 - 1 222 Chr. 6 - 161160084 - 161160577 [-] on Build GRCh38 102
          GenBankmRNA HY324532.1 - 1 Chr. 6 - 161160142 - 161160593 [+] on Build GRCh38 102
          GenBankmRNA AA729747.1 - 1 Chr. 6 - 161160117 - 161160450 [+] on Build GRCh38 102

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream 1-acylglycerol-3-phosphate O-acyltransferase 4 AGPAT4 1-AGPAT4,
          LPAAT-delta,
          dJ473J16.2
          Sense Chr. 6 - 161129980 - 161274108 on NCBI Build GRCh38
          Downstream parkin RBR E3 ubiquitin protein ligase PARK2 AR-JP,
          LPRS2,
          PDJ,
          PRKN
          186.9kb Sense Chr. 6 - 161347417 - 162727802 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P70359 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P70359 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 17146 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 18604 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P70359 - -
          SurePrint G3 Human GE Microarray G4851A A_23_P70359 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215164 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25420203 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215126 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215158 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215146 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25420219 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215129 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215169 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25420189 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215136 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215111 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215143 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215168 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215124 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215173 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215157 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215131 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25420174 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215180 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215182 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25420198 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215175 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215149 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25420210 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215118 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215138 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215166 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215123 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215139 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25420168 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215135 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215125 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215155 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215150 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25420231 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25420234 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215121 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215177 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215174 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215165 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215133 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215171 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25420218 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25420177 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25420202 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25420203 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215182 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215164 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215175 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215138 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25420189 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215135 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25420174 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215165 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215125 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215180 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215131 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215146 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215139 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215136 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215169 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215150 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25420234 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215173 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215177 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215174 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215168 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215126 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215155 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215111 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215149 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25420202 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215158 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25420219 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215171 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25420231 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25420218 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215143 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215124 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215121 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215133 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25420177 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215166 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215157 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25420168 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25420198 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215118 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215123 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25420210 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215129 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P70359 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P70359 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05215121 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05215162 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05215150 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05215118 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05215167 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05215132 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05215180 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05215142 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05215137 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05215159 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05215146 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05215136 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05215125 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05215175 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05215115 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05215169 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05215123 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05215184 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05215165 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05215153 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05215176 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05215155 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05215178 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05215113 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05215173 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05215183 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05215111 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05215157 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05215109 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05215129 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05215133 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05215139 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8130643 - -
          Affymetrix Human Gene 1.0 ST Array 8130644 - -
          Affymetrix Human Gene 1.0 ST Array 8130642 - -
          Affymetrix Human Gene 1.1 ST Array 8130641 - -
          Affymetrix HT HG-U133+ PM Array Plate 223740_PM_at - -
          Human Genome U133 Plus 2.0 Array 223740_at - -
          Human Genome U133 Plus 2.0 Array 223740_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 223740_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 223740_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 223740_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 223740_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 223740_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 223740_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 223740_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 223740_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 223740_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 223740_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 223740_at9 - -
          Human Genome U95 Set 71617_PM_at - -
          Human Genome U95 Set 71617_at - -
          Human Genome U133 Set 223740_at - -
          Human Genome U133 Set 223740_PM_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A030787 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A030787 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 144531 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3241996 NR_024277.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3241996 AK026765.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1655572 NR_024277.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1655572 AK026765.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1655572 AL136708.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3241996 NR_024277.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3241996 AK026765.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3241996 NR_024277.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3241996 AK026765.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1655572 NR_024277.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1655572 AK026765.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1655572 AL136708.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3241996 NR_024277.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3241996 AK026765.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1655572 NR_024277.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1655572 AK026765.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1655572 AL136708.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1655572 NR_024277.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1655572 AK026765.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1655572 AL136708.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1655572 NR_024277.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1655572 AK026765.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1655572 AL136708.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1655572 NR_024277.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1655572 AK026765.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1655572 AL136708.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1655572 NR_024277.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1655572 AK026765.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1655572 AL136708.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0