Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01089157_s1
          See other LOC79160 GE Assays ›
          Gene Symbol:
          LOC79160
          Gene Name:
          uncharacterized LOC79160
          Gene Aliases:
          -
          Chromosome Location:
          Chr.20: 58634772 - 58635738 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01089157_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC79160
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_125359.1 1 929 85
          GenBank mRNA BC002831.2 - 1 929 3636069 85
          BU624279.1 - 1 85
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BU621514.1 - 1 Chr. 20 - 58635036 - 58635738 [-] on Build GRCh38 85
          GenBankmRNA BE301183.1 - 1 Chr. 20 - 58635402 - 58635738 [-] on Build GRCh38 85

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream APCDD1L antisense RNA 1 (head to head) APCDD1L-AS1 -15.1kb Anti-sense Chr. 20 - 58515379 - 58619892 on NCBI Build GRCh38
          Downstream syntaxin 16 STX16 SYN16 15.5kb Anti-sense Chr. 20 - 58651253 - 58679526 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC2000007905.hg.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11240424 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11240455 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17203919 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11240425 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11240497 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11240524 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11240508 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11240456 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11240496 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31890402 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11240527 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11240516 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11240440 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11240447 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11240442 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31890430 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11240454 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11240477 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11240482 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11240418 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31890448 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11240483 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17203914 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11240473 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17203915 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11240511 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11240434 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11240445 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11240491 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31890451 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31890452 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11240507 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31890400 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31890433 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11240421 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11240439 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11240522 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11240514 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11240428 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11240446 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31890437 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11240443 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11240528 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11240481 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11240452 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11240449 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11240485 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31890427 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11240479 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11240504 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11240450 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17203908 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31890440 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11240426 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31890412 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11240510 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11240419 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31890431 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11240523 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17203920 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17203911 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11240512 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11240480 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11240501 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11240417 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11240457 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11240475 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11240420 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31890428 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31890461 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11240422 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11240517 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11240458 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11240471 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11240487 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11240503 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31890483 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31890421 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11240490 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31890463 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11240478 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11240499 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11240520 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11240436 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11240431 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11240468 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11240493 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 152071 - -
          Agilent SurePrint G3 Human CGH Microarray 8x60K 20937 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P102681 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 43253 - -
          Agilent Human Genome CGH Microarray 44B A_14_P107706 - -
          Agilent Human Genome CGH Microarray 44A A_14_P107706 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 3752 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P102681 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_14_P107706 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_14_P107706 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 170245 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 99599 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_14_P107706 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_14_P107706 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.1 ST Array 8063634 - -
          Affymetrix HT HG-U133+ PM Array Plate 220698_PM_at - -
          Affymetrix Human Clariom D Assay PSR2000154840.hg.1 ENST00000598340.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2000154840.hg.1 NR_125359.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000154840.hg.1 TC2000007905.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2000154840.hg.1 ENST00000598340.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2000154840.hg.1 NR_125359.1 Refseq
          Human Genome U133 Plus 2.0 Array 220698_at - -
          Human Genome U133 Plus 2.0 Array 220698_PM_at - -
          Human Genome U133A 2.0 Array 220698_PM_at - -
          Human Genome U133A 2.0 Array 220698_at - -
          AffymetrixPathwork Diagnostics Pathchip pw529367 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220698_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220698_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220698_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220698_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220698_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220698_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220698_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220698_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220698_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220698_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220698_at5 - -
          Human Genome U133 Set 220698_PM_at - -
          Human Genome U133 Set 220698_at - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 143099 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1749414 BC002831.2 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1749414 BC002831.2 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1749414 BC002831.2 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1749414 BC002831.2 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline