Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03032757.hg.1 |
ENST00000510994.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03032757.hg.1 |
NM_001242374.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03032757.hg.1 |
TC03001805.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03032762.hg.1 |
ENST00000354910.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03032762.hg.1 |
ENST00000510994.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03032762.hg.1 |
NM_001242374.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03032762.hg.1 |
NM_015391.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03032762.hg.1 |
TC03001805.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012498.hg.1 |
ENST00000354910.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012498.hg.1 |
ENST00000354910.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012498.hg.1 |
ENST00000354910.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012498.hg.1 |
ENST00000354910.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012498.hg.1 |
ENST00000354910.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012498.hg.1 |
ENST00000354910.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012498.hg.1 |
ENST00000354910.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012498.hg.1 |
ENST00000354910.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012498.hg.1 |
ENST00000503439.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012498.hg.1 |
ENST00000503439.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012498.hg.1 |
ENST00000503439.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012498.hg.1 |
ENST00000503439.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012498.hg.1 |
ENST00000508755.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012498.hg.1 |
ENST00000508755.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012498.hg.1 |
ENST00000508755.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012498.hg.1 |
ENST00000508755.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012498.hg.1 |
ENST00000508755.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012498.hg.1 |
ENST00000510994.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012498.hg.1 |
ENST00000510994.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012498.hg.1 |
ENST00000510994.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012498.hg.1 |
ENST00000510994.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012498.hg.1 |
ENST00000510994.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012498.hg.1 |
ENST00000510994.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012498.hg.1 |
ENST00000510994.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012498.hg.1 |
ENST00000510994.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012498.hg.1 |
ENST00000510994.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012498.hg.1 |
ENST00000510994.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012498.hg.1 |
ENST00000511751.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012498.hg.1 |
ENST00000511751.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012498.hg.1 |
ENST00000511751.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012498.hg.1 |
ENST00000511751.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012498.hg.1 |
ENST00000511751.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012498.hg.1 |
ENST00000511751.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012498.hg.1 |
ENST00000514612.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012498.hg.1 |
ENST00000514612.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012498.hg.1 |
ENST00000514612.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012498.hg.1 |
ENST00000514612.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012498.hg.1 |
ENST00000514612.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012498.hg.1 |
ENST00000514612.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012498.hg.1 |
ENST00000514612.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012498.hg.1 |
ENST00000514612.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012498.hg.1 |
NM_001242374.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012498.hg.1 |
NM_001242374.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012498.hg.1 |
NM_001242374.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012498.hg.1 |
NM_001242374.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012498.hg.1 |
NM_001242374.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012498.hg.1 |
NM_001242374.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012498.hg.1 |
NM_001242374.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012498.hg.1 |
NM_001242374.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012498.hg.1 |
NM_001242374.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012498.hg.1 |
NM_001242374.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012498.hg.1 |
NM_001242375.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012498.hg.1 |
NM_001242375.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012498.hg.1 |
NM_001242375.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012498.hg.1 |
NM_001242375.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012498.hg.1 |
NM_001242375.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012498.hg.1 |
NM_001242375.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012498.hg.1 |
NM_001242375.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012498.hg.1 |
NM_001242375.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012498.hg.1 |
NM_015391.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012498.hg.1 |
NM_015391.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012498.hg.1 |
NM_015391.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012498.hg.1 |
NM_015391.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012498.hg.1 |
NM_015391.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012498.hg.1 |
NM_015391.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012498.hg.1 |
NM_015391.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012498.hg.1 |
NM_015391.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300193596.hg.1 |
ENST00000503439.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300193596.hg.1 |
ENST00000510994.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300193596.hg.1 |
NM_001242374.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300193596.hg.1 |
TC0300012498.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300193596.hg.1 |
ENST00000503439.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300193596.hg.1 |
ENST00000510994.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300193596.hg.1 |
NM_001242374.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012498.hg.1 |
ENST00000354910.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012498.hg.1 |
ENST00000354910.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012498.hg.1 |
ENST00000354910.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012498.hg.1 |
ENST00000354910.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012498.hg.1 |
ENST00000354910.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012498.hg.1 |
ENST00000354910.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012498.hg.1 |
ENST00000354910.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012498.hg.1 |
ENST00000354910.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012498.hg.1 |
ENST00000503439.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012498.hg.1 |
ENST00000503439.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012498.hg.1 |
ENST00000503439.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012498.hg.1 |
ENST00000503439.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012498.hg.1 |
ENST00000508755.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012498.hg.1 |
ENST00000508755.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012498.hg.1 |
ENST00000508755.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012498.hg.1 |
ENST00000508755.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012498.hg.1 |
ENST00000508755.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012498.hg.1 |
ENST00000510994.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012498.hg.1 |
ENST00000510994.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012498.hg.1 |
ENST00000510994.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012498.hg.1 |
ENST00000510994.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012498.hg.1 |
ENST00000510994.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012498.hg.1 |
ENST00000510994.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012498.hg.1 |
ENST00000510994.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012498.hg.1 |
ENST00000510994.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012498.hg.1 |
ENST00000510994.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012498.hg.1 |
ENST00000510994.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012498.hg.1 |
ENST00000511751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012498.hg.1 |
ENST00000511751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012498.hg.1 |
ENST00000511751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012498.hg.1 |
ENST00000511751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012498.hg.1 |
ENST00000511751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012498.hg.1 |
ENST00000511751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012498.hg.1 |
ENST00000514612.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012498.hg.1 |
ENST00000514612.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012498.hg.1 |
ENST00000514612.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012498.hg.1 |
ENST00000514612.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012498.hg.1 |
ENST00000514612.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012498.hg.1 |
ENST00000514612.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012498.hg.1 |
ENST00000514612.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012498.hg.1 |
ENST00000514612.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012498.hg.1 |
NM_001242374.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012498.hg.1 |
NM_001242374.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012498.hg.1 |
NM_001242374.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012498.hg.1 |
NM_001242374.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012498.hg.1 |
NM_001242374.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012498.hg.1 |
NM_001242374.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012498.hg.1 |
NM_001242374.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012498.hg.1 |
NM_001242374.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012498.hg.1 |
NM_001242374.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012498.hg.1 |
NM_001242374.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012498.hg.1 |
NM_001242375.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012498.hg.1 |
NM_001242375.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012498.hg.1 |
NM_001242375.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012498.hg.1 |
NM_001242375.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012498.hg.1 |
NM_001242375.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012498.hg.1 |
NM_001242375.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012498.hg.1 |
NM_001242375.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012498.hg.1 |
NM_001242375.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012498.hg.1 |
NM_015391.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012498.hg.1 |
NM_015391.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012498.hg.1 |
NM_015391.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012498.hg.1 |
NM_015391.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012498.hg.1 |
NM_015391.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012498.hg.1 |
NM_015391.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012498.hg.1 |
NM_015391.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012498.hg.1 |
NM_015391.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300193601.hg.1 |
ENST00000354910.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300193601.hg.1 |
ENST00000510994.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300193601.hg.1 |
NM_001242374.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300193601.hg.1 |
NM_015391.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300193601.hg.1 |
TC0300012498.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300193601.hg.1 |
ENST00000354910.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300193601.hg.1 |
ENST00000510994.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300193601.hg.1 |
NM_001242374.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300193601.hg.1 |
NM_015391.3 |
Refseq |