Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01091028_m1
          See other AKAP9 GE Assays ›
          Gene Symbol:
          AKAP9
          Gene Name:
          A-kinase anchoring protein 9
          Gene Aliases:
          AKAP-9, AKAP350, AKAP450, CG-NAP, HYPERION, LQT11, MU-RMS-40.16A, PPP1R45, PRKA9, YOTIAO
          Chromosome Location:
          Chr.7: 91940867 - 92110673 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01091028_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: AKAP9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_005751.4 NP_005742.4 44 - 45 11125 61
          NM_147185.2 NP_671714.1 44 - 45 11101 61
          XM_017011642.1 XP_016867131.1 46 - 47 11268 61
          XM_017011643.1 XP_016867132.1 45 - 46 11229 61
          XM_017011644.1 XP_016867133.1 46 - 47 11268 61
          XM_017011645.1 XP_016867134.1 45 - 46 11214 61
          XM_017011646.1 XP_016867135.1 45 - 46 11229 61
          XM_017011647.1 XP_016867136.1 45 - 46 11175 61
          XM_017011648.1 XP_016867137.1 45 - 46 11172 61
          XM_017011649.1 XP_016867138.1 45 - 46 11205 61
          XM_017011650.1 XP_016867139.1 44 - 45 11133 61
          XM_017011651.1 XP_016867140.1 45 - 46 11127 61
          XM_017011652.1 XP_016867141.1 45 - 46 11079 61
          XM_017011653.1 XP_016867142.1 43 - 44 11040 61
          XM_017011654.1 XP_016867143.1 43 - 44 10992 61
          XM_017011655.1 XP_016867144.1 46 - 47 10927 61
          XM_017011656.1 XP_016867145.1 45 - 46 10857 61
          XM_017011657.1 XP_016867146.1 31 - 32 6777 61
          XM_017011658.1 XP_016867147.1 24 - 25 6139 61
          XM_017011659.1 XP_016867148.1 23 - 24 6100 61
          XM_017011660.1 XP_016867149.1 23 - 24 6100 61
          GenBank mRNA AB018346.1 - 17 - 18 4304 61
          AB019691.1 - 44 - 45 10962 61
          AF083037.1 - 37 - 38 9964 61
          AF091711.1 - 18 - 19 4445 61
          AJ131693.1 - 44 - 45 11125 61
          AK025160.1 - 4 - 5 744 61
          AK026444.1 - 6 - 7 1824 61
          AK074869.1 - 14 - 15 3004 61
          BC040932.1 - 3 - 4 499 61

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0700008318.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC07000545.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene AKAP9

          Gene Ontology Categories:

          Function(s) Process(es)

          G2/M transition of mitotic cell cycle
          MAPK cascade
          transport
          microtubule nucleation
          signal transduction
          chemical synaptic transmission
          positive regulation of peptidyl-serine phosphorylation
          positive regulation of GTPase activity
          regulation of membrane repolarization
          regulation of ventricular cardiac muscle cell membrane repolarization
          cardiac conduction
          cellular response to cAMP
          regulation of heart rate by cardiac conduction
          positive regulation of potassium ion transmembrane transporter activity
          DNA binding
          signaling receptor binding
          protein binding
          potassium channel regulator activity
          protein kinase A regulatory subunit binding
          transmembrane transporter binding
          molecular adaptor activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR07007931.hg.1 ENST00000356239.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07007931.hg.1 ENST00000358100.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07007931.hg.1 ENST00000359028.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07007931.hg.1 ENST00000413120.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07007931.hg.1 NM_005751.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07007931.hg.1 NM_147185.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07007931.hg.1 TC07000545.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0700158021.hg.1 ENST00000356239.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0700158021.hg.1 ENST00000359028.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0700158021.hg.1 ENST00000394534.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0700158021.hg.1 ENST00000463118.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700158021.hg.1 ENST00000487258.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700158021.hg.1 NM_005751.4 Refseq
          Affymetrix Human Clariom D Assay PSR0700158021.hg.1 NM_147185.2 Refseq
          Affymetrix Human Clariom D Assay PSR0700158021.hg.1 TC0700008318.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700158021.hg.1 ENST00000359028.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0700158021.hg.1 ENST00000356239.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0700158021.hg.1 ENST00000394534.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0700158021.hg.1 ENST00000463118.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700158021.hg.1 ENST00000487258.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700158021.hg.1 NM_005751.4 Refseq
          Affymetrix Human Clariom D Assay PSR0700158021.hg.1 NM_147185.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0