Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01033408.hg.1 |
ENST00000377482.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01033408.hg.1 |
ENST00000467067.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01033408.hg.1 |
NM_018948.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01033408.hg.1 |
TC01002166.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01033410.hg.1 |
ENST00000377482.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01033410.hg.1 |
NM_018948.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01033410.hg.1 |
TC01002166.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018403.hg.1 |
ENST00000377482.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018403.hg.1 |
ENST00000377482.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018403.hg.1 |
ENST00000377482.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018403.hg.1 |
ENST00000377482.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018403.hg.1 |
ENST00000377482.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018403.hg.1 |
ENST00000377482.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018403.hg.1 |
ENST00000377482.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018403.hg.1 |
ENST00000377482.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018403.hg.1 |
ENST00000377482.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018403.hg.1 |
ENST00000467067.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018403.hg.1 |
ENST00000467067.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018403.hg.1 |
ENST00000467067.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018403.hg.1 |
ENST00000467067.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018403.hg.1 |
ENST00000467067.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018403.hg.1 |
ENST00000467067.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018403.hg.1 |
ENST00000469499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018403.hg.1 |
ENST00000469499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018403.hg.1 |
ENST00000469499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018403.hg.1 |
ENST00000469499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018403.hg.1 |
ENST00000469499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018403.hg.1 |
ENST00000487559.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018403.hg.1 |
ENST00000487559.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018403.hg.1 |
ENST00000487559.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018403.hg.1 |
ENST00000487559.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018403.hg.1 |
ENST00000487559.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018403.hg.1 |
NM_018948.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018403.hg.1 |
NM_018948.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018403.hg.1 |
NM_018948.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018403.hg.1 |
NM_018948.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018403.hg.1 |
NM_018948.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018403.hg.1 |
NM_018948.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018403.hg.1 |
NM_018948.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018403.hg.1 |
NM_018948.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018403.hg.1 |
NM_018948.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018403.hg.1 |
NM_018948.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01033409.hg.1 |
ENST00000377482.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01033409.hg.1 |
NM_018948.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01033409.hg.1 |
TC01002166.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0100241434.hg.1 |
ENST00000377482.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100241434.hg.1 |
ENST00000469499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100241434.hg.1 |
ENST00000487559.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100241434.hg.1 |
NM_018948.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100241434.hg.1 |
TC0100018403.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100241434.hg.1 |
NM_018948.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100241434.hg.1 |
ENST00000377482.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100241434.hg.1 |
ENST00000469499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100241434.hg.1 |
ENST00000487559.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100241432.hg.1 |
ENST00000377482.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100241432.hg.1 |
ENST00000467067.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100241432.hg.1 |
ENST00000469499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100241432.hg.1 |
ENST00000487559.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100241432.hg.1 |
NM_018948.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100241432.hg.1 |
TC0100018403.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100241432.hg.1 |
ENST00000377482.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100241432.hg.1 |
ENST00000467067.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100241432.hg.1 |
ENST00000469499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100241432.hg.1 |
ENST00000487559.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100241432.hg.1 |
NM_018948.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018403.hg.1 |
ENST00000377482.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018403.hg.1 |
ENST00000377482.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018403.hg.1 |
ENST00000377482.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018403.hg.1 |
ENST00000377482.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018403.hg.1 |
ENST00000377482.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018403.hg.1 |
ENST00000377482.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018403.hg.1 |
ENST00000377482.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018403.hg.1 |
ENST00000377482.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018403.hg.1 |
ENST00000377482.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018403.hg.1 |
ENST00000467067.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018403.hg.1 |
ENST00000467067.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018403.hg.1 |
ENST00000467067.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018403.hg.1 |
ENST00000467067.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018403.hg.1 |
ENST00000467067.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018403.hg.1 |
ENST00000467067.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018403.hg.1 |
ENST00000469499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018403.hg.1 |
ENST00000469499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018403.hg.1 |
ENST00000469499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018403.hg.1 |
ENST00000469499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018403.hg.1 |
ENST00000469499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018403.hg.1 |
ENST00000487559.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018403.hg.1 |
ENST00000487559.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018403.hg.1 |
ENST00000487559.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018403.hg.1 |
ENST00000487559.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018403.hg.1 |
ENST00000487559.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018403.hg.1 |
NM_018948.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018403.hg.1 |
NM_018948.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018403.hg.1 |
NM_018948.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018403.hg.1 |
NM_018948.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018403.hg.1 |
NM_018948.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018403.hg.1 |
NM_018948.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018403.hg.1 |
NM_018948.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018403.hg.1 |
NM_018948.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018403.hg.1 |
NM_018948.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018403.hg.1 |
NM_018948.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100241435.hg.1 |
ENST00000377482.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100241435.hg.1 |
ENST00000469499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100241435.hg.1 |
ENST00000487559.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100241435.hg.1 |
NM_018948.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100241435.hg.1 |
TC0100018403.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100241435.hg.1 |
ENST00000377482.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100241435.hg.1 |
ENST00000469499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100241435.hg.1 |
ENST00000487559.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100241435.hg.1 |
NM_018948.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100241437.hg.1 |
NM_018948.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100241437.hg.1 |
TC0100018403.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100241437.hg.1 |
NM_018948.3 |
Refseq |