Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01092435_m1
          See other DHCR24 GE Assays ›
          Gene Symbol:
          DHCR24
          Gene Name:
          24-dehydrocholesterol reductase
          Gene Aliases:
          DCE, Nbla03646, SELADIN1, seladin-1
          Chromosome Location:
          Chr.1: 54849627 - 54887248 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01092435_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: DHCR24
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_014762.3 NP_055577.1 3 - 4 618 58
          GenBank mRNA AB073391.1 - 4 - 5 493 58
          AF261758.1 - 3 - 4 588 58
          AK302774.1 - 3 - 4 449 58
          AK304302.1 - 1 - 2 50 58
          AK304437.1 - 3 - 4 335 58
          AK316419.1 - 3 - 4 380 58
          AK316427.1 - 3 - 4 319 58
          BC004375.1 - 3 - 4 524 3638639 58
          BC011669.2 - 3 - 4 551 4130396 58
          D13643.3 - 3 - 4 601 58

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100014276.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01002694.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the AK304302.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene DHCR24

          Gene Ontology Categories:

          Function(s) Process(es)

          cholesterol biosynthetic process
          apoptotic process
          response to oxidative stress
          Ras protein signal transduction
          protein localization
          negative regulation of cell population proliferation
          response to hormone
          tissue development
          male genitalia development
          plasminogen activation
          cholesterol biosynthetic process via desmosterol
          cholesterol biosynthetic process via lathosterol
          amyloid precursor protein catabolic process
          negative regulation of apoptotic process
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          skin development
          membrane organization
          regulation of neuron death
          delta24(24-1) sterol reductase activity
          oxidoreductase activity, acting on CH-OH group of donors
          oxidoreductase activity, acting on the CH-CH group of donors, NAD or NADP as acceptor
          enzyme binding
          peptide antigen binding
          delta24-sterol reductase activity
          flavin adenine dinucleotide binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000436604.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000436604.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000436604.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000436604.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000436604.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01042255.hg.1 ENST00000371269.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01042255.hg.1 ENST00000535035.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01042255.hg.1 ENST00000537443.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01042255.hg.1 NM_014762.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01042255.hg.1 TC01002694.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0100206993.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100206993.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100206993.hg.1 NM_014762.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100206993.hg.1 TC0100014276.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100206993.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100206993.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100206993.hg.1 NM_014762.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000436604.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000436604.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000436604.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000436604.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000436604.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 NM_014762.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-f85dfd666-px2hc:80/100.66.130.62:80.
          git-commit: 2ca7b3ab9a2195be9b417413331e4e963d93e17c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0