Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01093190_s1
          See other PRO1804 GE Assays ›
          Gene Symbol:
          PRO1804
          Gene Name:
          uncharacterized LOC100133319
          Gene Aliases:
          -
          Chromosome Location:
          Chr.17: 69232975 - 69235012 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01093190_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PRO1804
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_130736.1 1 1092 75
          GenBank mRNA AF132201.1 - 1 1092 75

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream ATP binding cassette subfamily A member 6 ABCA6 EST155051 -91.0kb Sense Chr. 17 - 69062044 - 69142188 on NCBI Build GRCh38
          Downstream ATP binding cassette subfamily A member 5 ABCA5 ABC13,
          EST90625
          9.4kb Sense Chr. 17 - 69244435 - 69327182 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1700011582.hg.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3398074 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P376599 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 28300 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 42521 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3398074 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P376599 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P376599 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 224110_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC1700011582.hg.1 ENST00000623780.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011582.hg.1 NR_130736.1 Refseq
          Affymetrix Human Clariom D Assay TC1700011582.hg.1 ENST00000623780.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700011582.hg.1 NR_130736.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700193966.hg.1 ENST00000623780.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700193966.hg.1 NR_130736.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700193966.hg.1 TC1700011582.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700193966.hg.1 NR_130736.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700193966.hg.1 ENST00000623780.1 Ensembl
          Human Genome U133 Plus 2.0 Array 224110_at - -
          Human Genome U133 Plus 2.0 Array 224110_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224110_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224110_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224110_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224110_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224110_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224110_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224110_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224110_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224110_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224110_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 224110_at4 - -
          Human Genome U133 Set 224110_PM_at - -
          Human Genome U133 Set 224110_at - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 707950 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0