Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01093197_m1
          See other BCL11A GE Assays ›
          Gene Symbol:
          BCL11A
          Gene Name:
          B-cell CLL/lymphoma 11A
          Gene Aliases:
          BCL11A-L, BCL11A-S, BCL11A-XL, BCL11a-M, CTIP1, EVI9, HBFQTL5, ZNF856
          Chromosome Location:
          Chr.2: 60451167 - 60553498 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01093197_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: BCL11A
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_018014.3 NP_060484.2 3 - 4 711 62
          NM_022893.3 NP_075044.2 3 - 4 711 62
          NM_138559.1 NP_612569.1 3 - 4 711 62
          XM_011532909.1 XP_011531211.1 3 - 4 711 62
          XM_011532910.1 XP_011531212.1 3 - 4 711 62
          XM_017004333.1 XP_016859822.1 3 - 4 479 62
          XM_017004336.1 XP_016859825.1 3 - 4 274 62
          XM_017004337.1 XP_016859826.1 2 - 3 7180 62
          XM_017004338.1 XP_016859827.1 2 - 3 2924 62
          GenBank mRNA AB058712.1 - 3 - 4 611 62
          AJ404611.1 - 3 - 4 711 62
          AJ404612.1 - 3 - 4 711 62
          AJ404613.1 - 3 - 4 711 62
          AK055251.1 - 3 - 4 711 62
          AK300012.1 - 4 - 5 939 62
          BC021098.1 - 3 - 4 592 5087967 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0200012713.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC02001885.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the AK300012.1, XM_017004337.1, XM_017004338.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene BCL11A

          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor zinc finger transcription factor C2H2 zinc finger transcription factor gene-specific transcriptional regulator

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          transcription, DNA-templated
          signal transduction
          protein sumoylation
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
          metal ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000335712.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000335712.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000335712.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000335712.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000335712.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000335712.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000335712.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000335712.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000335712.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000335712.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000335712.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000335712.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000335712.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000335712.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000335712.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000356842.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000356842.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000356842.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000356842.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000356842.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000356842.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000356842.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000356842.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000356842.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000356842.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000356842.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000356842.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000356842.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000356842.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000356842.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000358510.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000358510.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000358510.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000358510.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000358510.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000358510.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000358510.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000358510.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000358510.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000358510.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000358510.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000359629.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000359629.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000359629.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000359629.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000359629.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000359629.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000359629.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000359629.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000359629.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000359629.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000359629.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000359629.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000359629.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000359629.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000409351.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000409351.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000409351.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000409351.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000409351.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000409351.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000409351.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000409351.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000477659.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000477659.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000477659.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000477659.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000477659.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000477659.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000477659.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000477659.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000477659.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000479026.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000479026.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000479026.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000479026.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000489183.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000489183.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000489516.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000489516.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000489516.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000489516.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000489516.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000489516.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000489516.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000489516.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000489516.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000492272.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000492272.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000492272.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000492272.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000492272.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000631857.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000631857.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000631857.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000631857.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000631857.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000631857.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000631857.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000631857.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000631857.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000631857.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000631857.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 ENST00000631857.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 NM_018014.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 NM_018014.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 NM_018014.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 NM_018014.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 NM_018014.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 NM_018014.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 NM_018014.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 NM_018014.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 NM_018014.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 NM_018014.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 NM_018014.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 NM_018014.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 NM_018014.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 NM_018014.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 NM_018014.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 NM_022893.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 NM_022893.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 NM_022893.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 NM_022893.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 NM_022893.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 NM_022893.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 NM_022893.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 NM_022893.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 NM_022893.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 NM_022893.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 NM_022893.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 NM_022893.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 NM_022893.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 NM_022893.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 NM_022893.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 NM_138559.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 NM_138559.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 NM_138559.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 NM_138559.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 NM_138559.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 NM_138559.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 NM_138559.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 NM_138559.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 NM_138559.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 NM_138559.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 NM_138559.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 NM_138559.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012713.hg.1 NM_138559.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-865d7754c7-mcq8j:80/100.66.130.62:80.
          git-commit: 7ff36f61b459eb8f02ba5a263f0e3ce367ca82d3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0