Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01093364_s1
          See other MAPK8IP2 GE Assays ›
          Gene Symbol:
          MAPK8IP2 LOC284939
          Gene Name:
          mitogen-activated protein kinase 8 interacting protein 2
          uncharacterized LOC284939
          Gene Aliases:
          IB-2, IB2, JIP2, PRKM8IPL
          -
          Chromosome Location:
          Chr.22: 50600685 - 50613978 on Build GRCh38
          Chromosome Location:
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Hs01093364_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MAPK8IP2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_011530679.2 XP_011528981.1 12 5283 68
          XM_011530680.2 XP_011528982.1 12 5169 68
          XM_011530681.2 XP_011528983.1 12 5148 68
          Gene Symbol: LOC284939
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK094442.1 - 1 2000 68
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BU584643.1 - 1 456 Chr. 22 - 50613106 - 50613704 [+] on Build GRCh38 68
          GenBankmRNA AI553861.1 - 1 Chr. 22 - 50613480 - 50613978 [-] on Build GRCh38 68
          GenBankmRNA AI828539.1 - 1 Chr. 22 - 50613455 - 50613978 [-] on Build GRCh38 68
          GenBankmRNA AA365645.1 - 1 92 Chr. 22 - 50613470 - 50613765 [+] on Build GRCh38 68
          GenBankmRNA BI710915.1 - 1 266 Chr. 22 - 50613303 - 50613803 [+] on Build GRCh38 68
          GenBankmRNA BI711365.1 - 1 Chr. 22 - 50613479 - 50613978 [-] on Build GRCh38 68
          GenBankmRNA AA323408.1 - 1 Chr. 22 - 50613360 - 50613670 [+] on Build GRCh38 69
          GenBankmRNA DB297548.1 - 1 Chr. 22 - 50613439 - 50613974 [-] on Build GRCh38 68
          GenBankmRNA BX096179.1 - 1 43 Chr. 22 - 50613519 - 50613970 [+] on Build GRCh38 68
          GenBankmRNA AW959804.1 - 1 91 Chr. 22 - 50613471 - 50613978 [+] on Build GRCh38 68
          GenBankmRNA AA323016.1 - 1 Chr. 22 - 50613362 - 50613658 [+] on Build GRCh38 69
          GenBankmRNA AI908117.1 - 55 Chr. - - [-] on Build 68

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Downstream arylsulfatase A ARSA MLD 9.1kb Anti-sense Chr. 22 - 50622754 - 50628173 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          behavioral fear response
          signal complex assembly
          JNK cascade
          mating behavior
          regulation of signaling receptor activity
          positive regulation of stress-activated MAPK cascade
          social behavior
          regulation of JNK cascade
          nonassociative learning
          dendrite morphogenesis
          regulation of synaptic transmission, glutamatergic
          excitatory postsynaptic potential
          regulation of NMDA receptor activity
          regulation of AMPA receptor activity
          amyloid-beta binding
          MAP-kinase scaffold activity
          structural molecule activity
          protein binding
          kinesin binding
          protein kinase binding
          protein-containing complex binding

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human CGH Microarray 4x180K 139768 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 882098 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 278798 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3821494 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 58683 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P21355604 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P21355604 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3821494 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_16_P21355604 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P21355604 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 15122 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 81730 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P21355604 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P21355604 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 1556001_PM_at - -
          Human Genome U133 Plus 2.0 Array 1556001_PM_at - -
          Human Genome U133 Plus 2.0 Array 1556001_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556001_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556001_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556001_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556001_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556001_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556001_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556001_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556001_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556001_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556001_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556001_at4 - -
          Human Genome U95 Set 55506_PM_at - -
          Human Genome U95 Set 55506_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0