Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01095516_s1
          See other LOC150005 GE Assays ›
          Gene Symbol:
          LOC150005
          Gene Name:
          uncharacterized LOC150005
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01095516_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC150005
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK057475.1 - 1 682 74

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream chondrolectin CHODL C21orf68,
          MT75,
          PRED12
          Anti-sense Chr. 21 - 17819329 - 18267371 on NCBI Build GRCh38
          Downstream transmembrane protease,
          serine 15
          TMPRSS15 ENTK,
          PRSS7
          337.9kb Sense Chr. 21 - 18269107 - 18403806 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31946237 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11287904 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11287908 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31946243 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31946226 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31946240 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11287933 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11287911 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31946247 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31946221 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31946235 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11287934 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31946227 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31946236 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31946254 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31946249 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11287936 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11287912 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31946229 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31946223 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31946222 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11287910 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17222517 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11287895 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11287928 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31946218 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31946215 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31946241 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31946238 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31946230 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31946216 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31946246 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11287932 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31946239 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31946242 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3482461 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 44154 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3482461 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 1561343_PM_a_at - -
          Affymetrix HT HG-U133+ PM Array Plate 1561342_PM_at - -
          Human Genome U133 Plus 2.0 Array 1561343_PM_a_at - -
          Human Genome U133 Plus 2.0 Array 1561342_PM_at - -
          Human Genome U133 Plus 2.0 Array 1561342_at - -
          Human Genome U133 Plus 2.0 Array 1561343_a_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561342_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561343_a_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561342_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561342_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561343_a_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561342_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561343_a_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561343_a_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561343_a_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561342_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561343_a_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561343_a_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561343_a_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561342_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561342_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561342_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561342_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561342_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561343_a_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561342_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561343_a_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561343_a_at2 - -
          Array Name Probe IDs Targets Target Type
          Illumina human-6 v2.0 expression beadchip ILMN_1877412 - -
          Illumina Sentrix Human-6 v2 Expression BeadChip 5550349 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_75089 - -
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1824864 - -
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1824864 - -
          Illumina HumanWG-6_V2_0_R2 ILMN_1877412 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0