Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01099960_g1
          See other CNNM1 GE Assays ›
          Gene Symbol:
          CNNM1
          Gene Name:
          cyclin and CBS domain divalent metal cation transport mediator 1
          Gene Aliases:
          ACDP1, CLP-1
          Chromosome Location:
          Chr.10: 99329099 - 99394330 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01099960_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CNNM1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_020348.2 NP_065081.2 8 - 9 2812 93
          XM_011539629.1 XP_011537931.1 9 - 10 2875 93
          GenBank mRNA AF169226.1 - 8 - 9 1635 93
          AK056194.1 - 7 - 8 915 93
          AK296545.1 - 9 - 10 2043 93
          AK310486.1 - 7 - 8 1067 93
          AL137536.1 - 1 - 2 183 93
          BC098103.1 - 8 - 9 1586 40006972 93
          BC098279.1 - 8 - 9 1586 40006977 93
          BC098307.1 - 8 - 9 1586 40006978 93
          JF267782.1 - 6 - 7 742 93

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1000008613.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC10000708.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CNNM1

          Gene Ontology Categories:

          Function(s)

          ion transport

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR10008260.hg.1 ENST00000356713.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10008260.hg.1 ENST00000370534.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10008260.hg.1 ENST00000446890.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10008260.hg.1 ENST00000545665.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10008260.hg.1 NM_020348.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10008260.hg.1 TC10000708.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1000008613.hg.1 ENST00000356713.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008613.hg.1 ENST00000356713.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008613.hg.1 ENST00000356713.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008613.hg.1 ENST00000356713.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008613.hg.1 ENST00000356713.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008613.hg.1 ENST00000356713.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008613.hg.1 ENST00000356713.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008613.hg.1 ENST00000356713.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008613.hg.1 ENST00000356713.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008613.hg.1 ENST00000356713.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008613.hg.1 ENST00000356713.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008613.hg.1 ENST00000356713.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008613.hg.1 ENST00000356713.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008613.hg.1 ENST00000356713.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008613.hg.1 ENST00000356713.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008613.hg.1 ENST00000488090.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008613.hg.1 ENST00000488090.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008613.hg.1 ENST00000488090.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008613.hg.1 NM_020348.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008613.hg.1 NM_020348.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008613.hg.1 NM_020348.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008613.hg.1 NM_020348.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008613.hg.1 NM_020348.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008613.hg.1 NM_020348.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008613.hg.1 NM_020348.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008613.hg.1 NM_020348.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008613.hg.1 NM_020348.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008613.hg.1 NM_020348.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008613.hg.1 NM_020348.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008613.hg.1 NM_020348.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008613.hg.1 NM_020348.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008613.hg.1 NM_020348.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008613.hg.1 NM_020348.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000158727.hg.1 ENST00000356713.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1000158727.hg.1 NM_020348.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000158727.hg.1 TC1000008613.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000158727.hg.1 NM_020348.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000158727.hg.1 ENST00000356713.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008613.hg.1 ENST00000356713.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008613.hg.1 ENST00000356713.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008613.hg.1 ENST00000356713.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008613.hg.1 ENST00000356713.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008613.hg.1 ENST00000356713.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008613.hg.1 ENST00000356713.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008613.hg.1 ENST00000356713.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008613.hg.1 ENST00000356713.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008613.hg.1 ENST00000356713.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008613.hg.1 ENST00000356713.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008613.hg.1 ENST00000356713.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008613.hg.1 ENST00000356713.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008613.hg.1 ENST00000356713.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008613.hg.1 ENST00000356713.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008613.hg.1 ENST00000356713.4 Ensembl
          Affymetrix Human Clariom D Assay TC1000008613.hg.1 ENST00000488090.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000008613.hg.1 ENST00000488090.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000008613.hg.1 ENST00000488090.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000008613.hg.1 NM_020348.2 Refseq
          Affymetrix Human Clariom D Assay TC1000008613.hg.1 NM_020348.2 Refseq
          Affymetrix Human Clariom D Assay TC1000008613.hg.1 NM_020348.2 Refseq
          Affymetrix Human Clariom D Assay TC1000008613.hg.1 NM_020348.2 Refseq
          Affymetrix Human Clariom D Assay TC1000008613.hg.1 NM_020348.2 Refseq
          Affymetrix Human Clariom D Assay TC1000008613.hg.1 NM_020348.2 Refseq
          Affymetrix Human Clariom D Assay TC1000008613.hg.1 NM_020348.2 Refseq
          Affymetrix Human Clariom D Assay TC1000008613.hg.1 NM_020348.2 Refseq
          Affymetrix Human Clariom D Assay TC1000008613.hg.1 NM_020348.2 Refseq
          Affymetrix Human Clariom D Assay TC1000008613.hg.1 NM_020348.2 Refseq
          Affymetrix Human Clariom D Assay TC1000008613.hg.1 NM_020348.2 Refseq
          Affymetrix Human Clariom D Assay TC1000008613.hg.1 NM_020348.2 Refseq
          Affymetrix Human Clariom D Assay TC1000008613.hg.1 NM_020348.2 Refseq
          Affymetrix Human Clariom D Assay TC1000008613.hg.1 NM_020348.2 Refseq
          Affymetrix Human Clariom D Assay TC1000008613.hg.1 NM_020348.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline