Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06025597.hg.1 |
ENST00000368573.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06025597.hg.1 |
ENST00000368576.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06025597.hg.1 |
ENST00000471919.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06025597.hg.1 |
NM_145062.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06025597.hg.1 |
TC06002050.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06025598.hg.1 |
ENST00000368573.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06025598.hg.1 |
ENST00000368576.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06025598.hg.1 |
ENST00000471919.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06025598.hg.1 |
NM_145062.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06025598.hg.1 |
TC06002050.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012971.hg.1 |
ENST00000368573.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012971.hg.1 |
ENST00000368573.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012971.hg.1 |
ENST00000368573.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012971.hg.1 |
ENST00000368573.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012971.hg.1 |
ENST00000368573.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012971.hg.1 |
ENST00000368573.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012971.hg.1 |
ENST00000368576.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012971.hg.1 |
ENST00000368576.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012971.hg.1 |
ENST00000368576.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012971.hg.1 |
ENST00000368576.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012971.hg.1 |
ENST00000368576.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012971.hg.1 |
ENST00000368576.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012971.hg.1 |
ENST00000368576.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012971.hg.1 |
ENST00000368576.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012971.hg.1 |
ENST00000368576.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012971.hg.1 |
ENST00000368576.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012971.hg.1 |
ENST00000368576.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012971.hg.1 |
ENST00000368576.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012971.hg.1 |
ENST00000471919.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012971.hg.1 |
ENST00000471919.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012971.hg.1 |
ENST00000471919.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012971.hg.1 |
ENST00000471919.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012971.hg.1 |
ENST00000471919.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012971.hg.1 |
ENST00000471919.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012971.hg.1 |
ENST00000471919.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012971.hg.1 |
ENST00000485498.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012971.hg.1 |
ENST00000485498.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012971.hg.1 |
ENST00000485498.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012971.hg.1 |
ENST00000485498.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012971.hg.1 |
NM_145062.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012971.hg.1 |
NM_145062.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012971.hg.1 |
NM_145062.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012971.hg.1 |
NM_145062.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012971.hg.1 |
NM_145062.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012971.hg.1 |
NM_145062.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012971.hg.1 |
NM_145062.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012971.hg.1 |
NM_145062.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012971.hg.1 |
NM_145062.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012971.hg.1 |
NM_145062.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012971.hg.1 |
NM_145062.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012971.hg.1 |
NM_145062.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600189512.hg.1 |
ENST00000368573.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600189512.hg.1 |
ENST00000368576.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600189512.hg.1 |
ENST00000471919.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600189512.hg.1 |
NM_145062.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600189512.hg.1 |
TC0600012971.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600189512.hg.1 |
NM_145062.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600189512.hg.1 |
ENST00000368573.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600189512.hg.1 |
ENST00000368576.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600189512.hg.1 |
ENST00000471919.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600189513.hg.1 |
ENST00000368573.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600189513.hg.1 |
ENST00000368576.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600189513.hg.1 |
ENST00000471919.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600189513.hg.1 |
NM_145062.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600189513.hg.1 |
TC0600012971.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600189513.hg.1 |
ENST00000368573.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600189513.hg.1 |
ENST00000368576.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600189513.hg.1 |
ENST00000471919.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600189513.hg.1 |
NM_145062.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012971.hg.1 |
ENST00000368573.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012971.hg.1 |
ENST00000368573.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012971.hg.1 |
ENST00000368573.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012971.hg.1 |
ENST00000368573.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012971.hg.1 |
ENST00000368573.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012971.hg.1 |
ENST00000368573.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012971.hg.1 |
ENST00000368576.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012971.hg.1 |
ENST00000368576.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012971.hg.1 |
ENST00000368576.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012971.hg.1 |
ENST00000368576.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012971.hg.1 |
ENST00000368576.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012971.hg.1 |
ENST00000368576.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012971.hg.1 |
ENST00000368576.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012971.hg.1 |
ENST00000368576.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012971.hg.1 |
ENST00000368576.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012971.hg.1 |
ENST00000368576.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012971.hg.1 |
ENST00000368576.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012971.hg.1 |
ENST00000368576.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012971.hg.1 |
ENST00000471919.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012971.hg.1 |
ENST00000471919.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012971.hg.1 |
ENST00000471919.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012971.hg.1 |
ENST00000471919.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012971.hg.1 |
ENST00000471919.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012971.hg.1 |
ENST00000471919.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012971.hg.1 |
ENST00000471919.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012971.hg.1 |
ENST00000485498.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012971.hg.1 |
ENST00000485498.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012971.hg.1 |
ENST00000485498.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012971.hg.1 |
ENST00000485498.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012971.hg.1 |
NM_145062.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012971.hg.1 |
NM_145062.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012971.hg.1 |
NM_145062.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012971.hg.1 |
NM_145062.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012971.hg.1 |
NM_145062.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012971.hg.1 |
NM_145062.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012971.hg.1 |
NM_145062.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012971.hg.1 |
NM_145062.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012971.hg.1 |
NM_145062.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012971.hg.1 |
NM_145062.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012971.hg.1 |
NM_145062.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012971.hg.1 |
NM_145062.2 |
Refseq |