Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01100751_m1
          See other CACNB2 GE Assays ›
          Gene Symbol:
          CACNB2
          Gene Name:
          calcium voltage-gated channel auxiliary subunit beta 2
          Gene Aliases:
          CACNLB2, CAVB2, MYSB
          Chromosome Location:
          Chr.10: 18140560 - 18545881 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01100751_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CACNB2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_000724.3 NP_000715.2 6 - 7 1255 78
          NM_201570.2 NP_963864.1 6 - 7 1109 78
          NM_201571.3 NP_963865.2 7 - 8 1088 78
          NM_201590.2 NP_963884.2 6 - 7 885 78
          NM_201596.2 NP_963890.2 7 - 8 865 78
          XM_006717502.3 XP_006717565.1 6 - 7 918 78
          GenBank mRNA AB208917.1 - 6 - 7 1141 78
          AF137376.1 - 7 - 8 803 78
          AF137377.1 - 6 - 7 646 78
          AF285239.2 - 6 - 7 643 78
          AF423189.1 - 6 - 7 640 78
          AF423190.1 - 7 - 8 721 78
          AF423191.1 - 7 - 8 805 78
          AF423192.1 - 6 - 7 661 78
          AK296471.1 - 7 - 8 772 78
          AK299873.1 - 6 - 7 1011 78
          AK310943.1 - 6 - 7 885 78
          AY393858.1 - 7 - 8 721 78
          AY393859.1 - 6 - 7 643 78
          BC136409.1 - 7 - 8 828 9020397 78
          S60415.1 - 7 - 8 865 78
          U95019.1 - 6 - 7 1141 78

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1000006917.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC10000138.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CACNB2

          Gene Ontology Categories:

          Function(s) Process(es)

          chemical synaptic transmission
          neuromuscular junction development
          visual perception
          positive regulation of calcium ion transport
          cardiac conduction
          calcium ion import
          calcium ion transmembrane transport
          protein localization to plasma membrane
          membrane depolarization during AV node cell action potential
          regulation of heart rate by cardiac conduction
          membrane depolarization during atrial cardiac muscle cell action potential
          regulation of voltage-gated calcium channel activity
          positive regulation of high voltage-gated calcium channel activity
          positive regulation of calcium ion transmembrane transport via high voltage-gated calcium channel
          voltage-gated calcium channel activity
          calcium channel regulator activity
          calcium channel activity
          protein binding
          high voltage-gated calcium channel activity
          protein kinase binding
          protein domain specific binding
          actin filament binding
          phosphoprotein binding
          voltage-gated calcium channel activity involved in cardiac muscle cell action potential
          voltage-gated calcium channel activity involved in AV node cell action potential

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR10001600.hg.1 ENST00000282343.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10001600.hg.1 ENST00000324631.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10001600.hg.1 ENST00000352115.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10001600.hg.1 ENST00000377315.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10001600.hg.1 ENST00000377319.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10001600.hg.1 ENST00000377329.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10001600.hg.1 ENST00000377331.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10001600.hg.1 ENST00000396576.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10001600.hg.1 NM_000724.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10001600.hg.1 NM_001167945.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10001600.hg.1 NM_201570.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10001600.hg.1 NM_201571.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10001600.hg.1 NM_201572.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10001600.hg.1 NM_201590.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10001600.hg.1 NM_201593.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10001600.hg.1 NM_201596.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10001600.hg.1 NM_201597.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10001600.hg.1 TC10000138.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1000147387.hg.1 ENST00000282343.12 Ensembl
          Affymetrix Human Clariom D Assay PSR1000147387.hg.1 ENST00000324631.11 Ensembl
          Affymetrix Human Clariom D Assay PSR1000147387.hg.1 ENST00000352115.10 Ensembl
          Affymetrix Human Clariom D Assay PSR1000147387.hg.1 ENST00000377315.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1000147387.hg.1 ENST00000377319.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1000147387.hg.1 ENST00000377329.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1000147387.hg.1 ENST00000377331.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1000147387.hg.1 ENST00000396576.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1000147387.hg.1 ENST00000612134.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1000147387.hg.1 ENST00000617363.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1000147387.hg.1 NM_000724.3 Refseq
          Affymetrix Human Clariom D Assay PSR1000147387.hg.1 NM_001167945.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000147387.hg.1 NM_201570.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000147387.hg.1 NM_201571.3 Refseq
          Affymetrix Human Clariom D Assay PSR1000147387.hg.1 NM_201572.3 Refseq
          Affymetrix Human Clariom D Assay PSR1000147387.hg.1 NM_201590.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000147387.hg.1 NM_201593.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000147387.hg.1 NM_201596.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000147387.hg.1 NM_201597.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000147387.hg.1 TC1000006917.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000147387.hg.1 ENST00000282343.12 Ensembl
          Affymetrix Human Clariom D Assay PSR1000147387.hg.1 ENST00000324631.11 Ensembl
          Affymetrix Human Clariom D Assay PSR1000147387.hg.1 ENST00000352115.10 Ensembl
          Affymetrix Human Clariom D Assay PSR1000147387.hg.1 ENST00000377315.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1000147387.hg.1 ENST00000377319.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1000147387.hg.1 ENST00000377329.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1000147387.hg.1 ENST00000377331.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1000147387.hg.1 ENST00000396576.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1000147387.hg.1 ENST00000612134.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1000147387.hg.1 ENST00000617363.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1000147387.hg.1 NM_000724.3 Refseq
          Affymetrix Human Clariom D Assay PSR1000147387.hg.1 NM_001167945.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000147387.hg.1 NM_201570.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000147387.hg.1 NM_201571.3 Refseq
          Affymetrix Human Clariom D Assay PSR1000147387.hg.1 NM_201572.3 Refseq
          Affymetrix Human Clariom D Assay PSR1000147387.hg.1 NM_201590.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000147387.hg.1 NM_201593.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000147387.hg.1 NM_201596.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000147387.hg.1 NM_201597.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0