Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01101228_g1
          See other NR1H3 GE Assays ›
          Gene Symbol:
          NR1H3
          Gene Name:
          nuclear receptor subfamily 1 group H member 3
          Gene Aliases:
          LXR-a, LXRA, RLD-1
          Chromosome Location:
          Chr.11: 47248300 - 47269033 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01101228_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: NR1H3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001130101.2 NP_001123573.1 2 - 3 293 114
          NM_005693.3 NP_005684.2 2 - 3 293 114
          XM_005252705.1 XP_005252762.1 3 - 4 246 114
          XM_005252715.2 XP_005252772.1 2 - 3 240 114
          XM_006718112.1 XP_006718175.1 3 - 4 235 114
          XM_006718113.1 XP_006718176.1 3 - 4 246 114
          GenBank mRNA AK290614.1 - 2 - 3 248 114
          AK309027.1 - 2 - 3 233 114
          AK313311.1 - 2 - 3 241 114
          BC008819.2 - 2 - 3 212 3957848 114
          BP300071.1 - 2 - 3 264 114

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AK290614.1, AK313311.1, BC008819.2, NM_001130101.2, NM_005693.3, XM_005252705.1, XM_005252715.2, XM_006718112.1, XM_006718113.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene NR1H3

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          transcription initiation from RNA polymerase II promoter
          negative regulation of macrophage derived foam cell differentiation
          positive regulation of triglyceride biosynthetic process
          positive regulation of cholesterol efflux
          negative regulation of cholesterol storage
          intracellular receptor signaling pathway
          positive regulation of cellular protein metabolic process
          negative regulation of lipid transport
          positive regulation of cholesterol transport
          response to progesterone
          positive regulation of toll-like receptor 4 signaling pathway
          cholesterol homeostasis
          regulation of circadian rhythm
          negative regulation of macrophage activation
          apoptotic cell clearance
          steroid hormone mediated signaling pathway
          positive regulation of fatty acid biosynthetic process
          positive regulation of transcription, DNA-templated
          positive regulation of transcription by RNA polymerase II
          negative regulation of pinocytosis
          negative regulation of inflammatory response
          positive regulation of lipoprotein lipase activity
          lipid homeostasis
          sterol homeostasis
          negative regulation of interferon-gamma-mediated signaling pathway
          triglyceride homeostasis
          cellular response to lipopolysaccharide
          negative regulation of pancreatic juice secretion
          negative regulation of secretion of lysosomal enzymes
          transcription cis-regulatory region binding
          DNA binding
          steroid hormone receptor activity
          transcription coactivator activity
          nuclear receptor activity
          protein binding
          zinc ion binding
          cholesterol binding
          sterol response element binding
          sequence-specific DNA binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0