Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009303.hg.1 |
ENST00000343572.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009303.hg.1 |
ENST00000343572.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009303.hg.1 |
ENST00000343572.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009303.hg.1 |
ENST00000343572.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009303.hg.1 |
ENST00000609727.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009303.hg.1 |
ENST00000609727.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009303.hg.1 |
ENST00000609727.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009303.hg.1 |
ENST00000609727.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009303.hg.1 |
ENST00000613549.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009303.hg.1 |
ENST00000613549.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009303.hg.1 |
ENST00000613549.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009303.hg.1 |
ENST00000613549.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009303.hg.1 |
ENST00000613549.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009303.hg.1 |
ENST00000613549.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009303.hg.1 |
ENST00000613549.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009303.hg.1 |
ENST00000613549.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009303.hg.1 |
ENST00000613549.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009303.hg.1 |
ENST00000613549.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009303.hg.1 |
ENST00000613549.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009303.hg.1 |
ENST00000613549.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009303.hg.1 |
ENST00000613549.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009303.hg.1 |
NM_003585.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009303.hg.1 |
NM_003585.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009303.hg.1 |
NM_003585.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009303.hg.1 |
NM_003585.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009303.hg.1 |
NM_003585.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009303.hg.1 |
NM_003585.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009303.hg.1 |
NM_003585.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009303.hg.1 |
NM_003585.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009303.hg.1 |
NM_003585.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009303.hg.1 |
NM_003585.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009303.hg.1 |
NM_003585.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009303.hg.1 |
NM_003585.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009303.hg.1 |
ENST00000343572.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009303.hg.1 |
ENST00000343572.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009303.hg.1 |
ENST00000343572.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009303.hg.1 |
ENST00000343572.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009303.hg.1 |
ENST00000609727.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009303.hg.1 |
ENST00000609727.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009303.hg.1 |
ENST00000609727.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009303.hg.1 |
ENST00000609727.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009303.hg.1 |
ENST00000613549.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009303.hg.1 |
ENST00000613549.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009303.hg.1 |
ENST00000613549.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009303.hg.1 |
ENST00000613549.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009303.hg.1 |
ENST00000613549.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009303.hg.1 |
ENST00000613549.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009303.hg.1 |
ENST00000613549.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009303.hg.1 |
ENST00000613549.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009303.hg.1 |
ENST00000613549.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009303.hg.1 |
ENST00000613549.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009303.hg.1 |
ENST00000613549.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009303.hg.1 |
ENST00000613549.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009303.hg.1 |
ENST00000613549.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009303.hg.1 |
NM_003585.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009303.hg.1 |
NM_003585.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009303.hg.1 |
NM_003585.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009303.hg.1 |
NM_003585.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009303.hg.1 |
NM_003585.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009303.hg.1 |
NM_003585.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009303.hg.1 |
NM_003585.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009303.hg.1 |
NM_003585.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009303.hg.1 |
NM_003585.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009303.hg.1 |
NM_003585.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009303.hg.1 |
NM_003585.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009303.hg.1 |
NM_003585.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700170802.hg.1 |
ENST00000613549.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700170802.hg.1 |
NM_003585.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700170802.hg.1 |
TC1700009303.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700170802.hg.1 |
NM_003585.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700170802.hg.1 |
ENST00000613549.2 |
Ensembl |