Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01102590_g1
          See other DOC2B GE Assays ›
          Gene Symbol:
          DOC2B
          Gene Name:
          double C2 domain beta
          Gene Aliases:
          DOC2BL
          Chromosome Location:
          Chr.17: 146748 - 181630 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01102590_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: DOC2B
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_003585.4 NP_003576.2 8 - 9 1253 67
          XM_011535363.2 XP_011533665.1 7 - 8 1069 67
          GenBank mRNA BM713588.1 - 3 - 4 262 67
          D70830.1 - 8 - 9 1262 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1700009303.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Homo sapiens 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene DOC2B

          Gene Ontology Categories:

          Function(s) Process(es)

          vesicle fusion
          protein localization
          regulation of calcium ion-dependent exocytosis
          positive regulation of vesicle fusion
          positive regulation of insulin secretion
          positive regulation of calcium ion-dependent exocytosis
          calcium ion-regulated exocytosis of neurotransmitter
          calcium ion binding
          calcium-dependent phospholipid binding
          syntaxin binding
          clathrin binding

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human CGH Microarray 4x180K 149227 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K chr17_random - -
          Agilent SurePrint G3 Human CGH Microarray 8x60K 53302 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3256396 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P236753 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P236753 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 31194 - -
          Agilent Human Genome CGH Microarray 44B A_14_P113404 - -
          Agilent Human Genome CGH Microarray 44A A_14_P113404 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 28655 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 15237 - -
          Agilent Human Genome CGH + SNP Microarray A_18_P12553855 - -
          Agilent Human Genome CGH + SNP Microarray A_18_P12558599 - -
          Agilent Human Genome CGH + SNP Microarray A_18_P12555139 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P03199745 - -
          Agilent Human Genome CGH + SNP Microarray A_18_P12558827 - -
          Agilent Human Genome CGH + SNP Microarray A_18_P20752584 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3256396 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P236753 - -
          SurePrint G3 Human GE Microarray G4851A A_24_P236753 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3256396 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16932268 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30732949 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30732928 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10185394 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10185394 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30732949 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16932268 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30732928 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P236753 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_14_P113404 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_14_P113404 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P41920960 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P03809965 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P03809963 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P03809959 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P236753 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 39216 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03809960 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P41920960 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P41920951 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P21001273 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_14_P113404 - -
          Agilent Human 1B Microarray G4111A A_32_P413390 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P41920960 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_14_P113404 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P41920951 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NM_003585_at - -
          Affymetrix Human Gene 1.0 ST Array 8010902 - -
          Affymetrix Human Gene 1.1 ST Array 8010901 - -
          Affymetrix HT HG-U133+ PM Array Plate 207311_PM_at - -
          Affymetrix Human Genome U219 Array 11737636_PM_at - -
          Affymetrix Human Genome U219 Array 11737636_at - -
          Affymetrix Human Transcriptome Array 2.0 TC1700009303.hg.1 ENST00000343572.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009303.hg.1 ENST00000343572.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009303.hg.1 ENST00000343572.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009303.hg.1 ENST00000343572.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009303.hg.1 ENST00000609727.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009303.hg.1 ENST00000609727.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009303.hg.1 ENST00000609727.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009303.hg.1 ENST00000609727.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009303.hg.1 ENST00000613549.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009303.hg.1 ENST00000613549.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009303.hg.1 ENST00000613549.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009303.hg.1 ENST00000613549.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009303.hg.1 ENST00000613549.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009303.hg.1 ENST00000613549.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009303.hg.1 ENST00000613549.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009303.hg.1 ENST00000613549.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009303.hg.1 ENST00000613549.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009303.hg.1 ENST00000613549.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009303.hg.1 ENST00000613549.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009303.hg.1 ENST00000613549.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009303.hg.1 ENST00000613549.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009303.hg.1 NM_003585.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009303.hg.1 NM_003585.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009303.hg.1 NM_003585.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009303.hg.1 NM_003585.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009303.hg.1 NM_003585.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009303.hg.1 NM_003585.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009303.hg.1 NM_003585.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009303.hg.1 NM_003585.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009303.hg.1 NM_003585.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009303.hg.1 NM_003585.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009303.hg.1 NM_003585.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009303.hg.1 NM_003585.4 Refseq
          Affymetrix Human Clariom D Assay PSR1700170798.hg.1 ENST00000613549.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1700170798.hg.1 NM_003585.4 Refseq
          Affymetrix Human Clariom D Assay PSR1700170798.hg.1 TC1700009303.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700170798.hg.1 NM_003585.4 Refseq
          Affymetrix Human Clariom D Assay PSR1700170798.hg.1 ENST00000613549.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1700170800.hg.1 ENST00000613549.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1700170800.hg.1 NM_003585.4 Refseq
          Affymetrix Human Clariom D Assay PSR1700170800.hg.1 TC1700009303.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700170800.hg.1 ENST00000613549.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1700170800.hg.1 NM_003585.4 Refseq
          Affymetrix Human Clariom D Assay TC1700009303.hg.1 ENST00000343572.8 Ensembl
          Affymetrix Human Clariom D Assay TC1700009303.hg.1 ENST00000343572.8 Ensembl
          Affymetrix Human Clariom D Assay TC1700009303.hg.1 ENST00000343572.8 Ensembl
          Affymetrix Human Clariom D Assay TC1700009303.hg.1 ENST00000343572.8 Ensembl
          Affymetrix Human Clariom D Assay TC1700009303.hg.1 ENST00000609727.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700009303.hg.1 ENST00000609727.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700009303.hg.1 ENST00000609727.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700009303.hg.1 ENST00000609727.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700009303.hg.1 ENST00000613549.2 Ensembl
          Affymetrix Human Clariom D Assay TC1700009303.hg.1 ENST00000613549.2 Ensembl
          Affymetrix Human Clariom D Assay TC1700009303.hg.1 ENST00000613549.2 Ensembl
          Affymetrix Human Clariom D Assay TC1700009303.hg.1 ENST00000613549.2 Ensembl
          Affymetrix Human Clariom D Assay TC1700009303.hg.1 ENST00000613549.2 Ensembl
          Affymetrix Human Clariom D Assay TC1700009303.hg.1 ENST00000613549.2 Ensembl
          Affymetrix Human Clariom D Assay TC1700009303.hg.1 ENST00000613549.2 Ensembl
          Affymetrix Human Clariom D Assay TC1700009303.hg.1 ENST00000613549.2 Ensembl
          Affymetrix Human Clariom D Assay TC1700009303.hg.1 ENST00000613549.2 Ensembl
          Affymetrix Human Clariom D Assay TC1700009303.hg.1 ENST00000613549.2 Ensembl
          Affymetrix Human Clariom D Assay TC1700009303.hg.1 ENST00000613549.2 Ensembl
          Affymetrix Human Clariom D Assay TC1700009303.hg.1 ENST00000613549.2 Ensembl
          Affymetrix Human Clariom D Assay TC1700009303.hg.1 ENST00000613549.2 Ensembl
          Affymetrix Human Clariom D Assay TC1700009303.hg.1 NM_003585.4 Refseq
          Affymetrix Human Clariom D Assay TC1700009303.hg.1 NM_003585.4 Refseq
          Affymetrix Human Clariom D Assay TC1700009303.hg.1 NM_003585.4 Refseq
          Affymetrix Human Clariom D Assay TC1700009303.hg.1 NM_003585.4 Refseq
          Affymetrix Human Clariom D Assay TC1700009303.hg.1 NM_003585.4 Refseq
          Affymetrix Human Clariom D Assay TC1700009303.hg.1 NM_003585.4 Refseq
          Affymetrix Human Clariom D Assay TC1700009303.hg.1 NM_003585.4 Refseq
          Affymetrix Human Clariom D Assay TC1700009303.hg.1 NM_003585.4 Refseq
          Affymetrix Human Clariom D Assay TC1700009303.hg.1 NM_003585.4 Refseq
          Affymetrix Human Clariom D Assay TC1700009303.hg.1 NM_003585.4 Refseq
          Affymetrix Human Clariom D Assay TC1700009303.hg.1 NM_003585.4 Refseq
          Affymetrix Human Clariom D Assay TC1700009303.hg.1 NM_003585.4 Refseq
          Human Genome U133 Plus 2.0 Array 207311_PM_at - -
          Human Genome U133 Plus 2.0 Array 207311_at - -
          Human Genome U133A 2.0 Array 207311_at - -
          Human Genome U133A 2.0 Array 207311_PM_at - -
          Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) NM_003585_at - -
          AffymetrixPathwork Diagnostics Pathchip pw561481 - -
          Human Genome U95 Set 32422_PM_at - -
          Human Genome U95 Set 32422_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 207311_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 207311_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 207311_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 207311_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 207311_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 207311_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 207311_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 207311_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 207311_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 207311_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 207311_at4 - -
          Human Genome U133 Set 207311_at - -
          Human Genome U133 Set 207311_PM_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A054959 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A054959 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 121173 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1718401 NM_003585.3 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1718401 D70830.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1718401 NM_003585.3 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1718401 D70830.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1718401 NM_003585.3 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1718401 D70830.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1718401 NM_003585.3 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1718401 D70830.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1718401 NM_003585.3 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1718401 D70830.1 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_6005996-S NM_003585.3 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_6005996-S D70830.1 human_transcriptome
          Illumina HumanRef-8 v2 Expression BeadChip GI_3915-S - -
          Illumina human-6 v1.0 expression beadchip 0006370026 - -
          Illumina humanRef-8 v1.0 expression beadchip 0006370026 - -
          Illumina human-6 v2.0 expression beadchip ILMN_1718401 NM_003585.3 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1718401 D70830.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1718401 NM_003585.3 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1718401 D70830.1 human_transcriptome
          Illumina Sentrix Human-6 v2 Expression BeadChip ILMN_3915 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_3915 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1718401 NM_003585.3 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1718401 D70830.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1718401 NM_003585.3 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1718401 D70830.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1718401 NM_003585.3 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1718401 D70830.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1718401 NM_003585.3 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1718401 D70830.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1718401 NM_003585.3 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1718401 D70830.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0