Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01103021_m1
          See other CTNND2 GE Assays ›
          Gene Symbol:
          CTNND2
          Gene Name:
          catenin delta 2
          Gene Aliases:
          GT24, NPRAP
          Chromosome Location:
          Chr.5: 10971840 - 11904067 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01103021_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CTNND2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001288715.1 NP_001275644.1 17 - 18 3087 71
          NM_001288716.1 NP_001275645.1 15 - 16 2349 71
          NM_001288717.1 NP_001275646.1 17 - 18 2714 71
          NM_001332.3 NP_001323.1 18 - 19 3279 71
          NR_109988.1 17 - 18 2616 71
          XM_005248251.3 XP_005248308.1 19 - 20 3378 71
          XM_005248252.2 XP_005248309.1 19 - 20 3544 71
          XM_005248253.1 XP_005248310.1 18 - 19 3069 71
          XM_011513967.2 XP_011512269.1 18 - 19 3458 71
          XM_017009072.1 XP_016864561.1 17 - 18 2628 71
          XM_017009073.1 XP_016864562.1 17 - 18 2803 71
          XM_017009074.1 XP_016864563.1 16 - 17 2550 71
          XM_017009075.1 XP_016864564.1 16 - 17 2677 71
          XM_017009076.1 XP_016864565.1 16 - 17 2688 71
          GenBank mRNA AB013805.1 - 18 - 19 3235 71
          AF035302.1 - 6 - 7 856 71
          AK226124.1 - 17 - 18 2966 71
          AK294420.1 - 15 - 16 2142 71
          AK296232.1 - 17 - 18 2714 71
          AK299300.1 - 15 - 16 2280 71
          U52351.1 - 12 - 13 2044 71
          U52828.1 - 4 - 5 497 71
          U72665.1 - 7 - 8 1026 71
          U81004.1 - 13 - 14 2535 71
          U96136.1 - 18 - 19 3089 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0500010115.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CTNND2

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          cell adhesion
          signal transduction
          Wnt signaling pathway
          synapse organization
          regulation of canonical Wnt signaling pathway
          dendritic spine morphogenesis
          cell-cell adhesion
          protein binding
          beta-catenin binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Clariom D Assay PSR0500170916.hg.1 ENST00000304623.12 Ensembl
          Affymetrix Human Clariom D Assay PSR0500170916.hg.1 ENST00000458100.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0500170916.hg.1 ENST00000495388.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0500170916.hg.1 ENST00000503622.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500170916.hg.1 ENST00000504499.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500170916.hg.1 ENST00000506324.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500170916.hg.1 ENST00000511377.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500170916.hg.1 ENST00000513588.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500170916.hg.1 NM_001288715.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500170916.hg.1 NM_001288716.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500170916.hg.1 NM_001288717.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500170916.hg.1 NM_001332.3 Refseq
          Affymetrix Human Clariom D Assay PSR0500170916.hg.1 NR_109988.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500170916.hg.1 TC0500010115.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0500170916.hg.1 ENST00000503622.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500170916.hg.1 ENST00000304623.12 Ensembl
          Affymetrix Human Clariom D Assay PSR0500170916.hg.1 ENST00000458100.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0500170916.hg.1 ENST00000495388.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0500170916.hg.1 ENST00000504499.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500170916.hg.1 ENST00000506324.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500170916.hg.1 ENST00000511377.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500170916.hg.1 ENST00000513588.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500170916.hg.1 NM_001288715.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500170916.hg.1 NM_001288716.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500170916.hg.1 NM_001288717.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500170916.hg.1 NM_001332.3 Refseq
          Affymetrix Human Clariom D Assay PSR0500170916.hg.1 NR_109988.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0