Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01103156_m1
          See other ITGB4 GE Assays ›
          Gene Symbol:
          ITGB4
          Gene Name:
          integrin subunit beta 4
          Gene Aliases:
          CD104, GP150
          Chromosome Location:
          Chr.17: 75721328 - 75757819 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01103156_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ITGB4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_000213.4 NP_000204.3 23 - 24 2919 77
          NM_001005619.1 NP_001005619.1 22 - 23 2632 77
          NM_001005731.2 NP_001005731.1 23 - 24 2919 77
          NM_001321123.1 NP_001308052.1 23 - 24 2870 77
          XM_005257309.2 XP_005257366.1 23 - 24 2689 77
          XM_005257311.4 XP_005257368.1 23 - 24 2908 77
          XM_006721866.3 XP_006721929.1 23 - 24 2835 77
          XM_006721867.3 XP_006721930.1 23 - 24 2832 77
          XM_006721868.3 XP_006721931.1 23 - 24 2829 77
          XM_006721870.3 XP_006721933.1 23 - 24 2825 77
          XM_011524751.2 XP_011523053.1 23 - 24 2833 77
          XM_011524752.2 XP_011523054.1 8 - 9 1118 77
          GenBank mRNA AB208913.1 - 23 - 24 2721 77
          AF011375.1 - 22 - 23 2661 77
          BC118916.1 - 23 - 24 2681 40073279 77
          BC126411.1 - 23 - 24 2740 8992127 77
          BC143738.1 - 23 - 24 2740 9052252 77
          BC143742.1 - 23 - 24 2691 9052256 77
          X51841.1 - 23 - 24 2750 77
          X52186.1 - 22 - 23 2633 77
          X53587.1 - 23 - 24 2811 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1700008920.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC17000861.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene ITGB4

          Gene Ontology Categories:

          Function(s) Process(es)

          autophagy
          cell adhesion
          cell-matrix adhesion
          integrin-mediated signaling pathway
          response to wounding
          extracellular matrix organization
          hemidesmosome assembly
          nail development
          skin development
          mesodermal cell differentiation
          digestive tract development
          cell motility
          renal system development
          amelogenesis
          G protein-coupled receptor binding
          protein binding
          insulin-like growth factor I binding
          signaling receptor activity
          neuregulin binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR17011209.hg.1 ENST00000200181.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17011209.hg.1 ENST00000339591.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17011209.hg.1 ENST00000449880.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17011209.hg.1 ENST00000450894.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17011209.hg.1 NM_000213.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17011209.hg.1 NM_001005619.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17011209.hg.1 NM_001005731.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17011209.hg.1 TC17000861.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1700167128.hg.1 ENST00000200181.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1700167128.hg.1 ENST00000449880.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1700167128.hg.1 ENST00000450894.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1700167128.hg.1 ENST00000579662.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700167128.hg.1 ENST00000584558.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700167128.hg.1 NM_000213.4 Refseq
          Affymetrix Human Clariom D Assay PSR1700167128.hg.1 NM_001005619.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700167128.hg.1 NM_001005731.2 Refseq
          Affymetrix Human Clariom D Assay PSR1700167128.hg.1 TC1700008920.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700167128.hg.1 ENST00000200181.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1700167128.hg.1 ENST00000449880.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1700167128.hg.1 ENST00000450894.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1700167128.hg.1 ENST00000579662.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700167128.hg.1 ENST00000584558.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700167128.hg.1 NM_000213.4 Refseq
          Affymetrix Human Clariom D Assay PSR1700167128.hg.1 NM_001005619.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700167128.hg.1 NM_001005731.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-786bfc498d-47c8f:80/100.66.130.62:80.
          git-commit: 9eb01563f6748445c58f67612c67ecf6b1da7421
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0