Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008708.hg.1 |
ENST00000373615.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008708.hg.1 |
ENST00000373615.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008708.hg.1 |
ENST00000373615.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008708.hg.1 |
ENST00000373615.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008708.hg.1 |
ENST00000373615.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008708.hg.1 |
ENST00000373615.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008708.hg.1 |
ENST00000373615.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008708.hg.1 |
ENST00000373615.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008708.hg.1 |
ENST00000373615.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008708.hg.1 |
ENST00000373615.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008708.hg.1 |
ENST00000373615.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008708.hg.1 |
ENST00000373615.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008708.hg.1 |
ENST00000446480.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008708.hg.1 |
ENST00000446480.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008708.hg.1 |
ENST00000446480.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008708.hg.1 |
ENST00000446480.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008708.hg.1 |
ENST00000446480.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008708.hg.1 |
ENST00000446480.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008708.hg.1 |
ENST00000446480.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008708.hg.1 |
ENST00000446480.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008708.hg.1 |
ENST00000488674.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008708.hg.1 |
ENST00000488674.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008708.hg.1 |
ENST00000488674.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008708.hg.1 |
ENST00000488674.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008708.hg.1 |
ENST00000488674.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008708.hg.1 |
ENST00000488674.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008708.hg.1 |
ENST00000488674.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008708.hg.1 |
ENST00000560961.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008708.hg.1 |
ENST00000560961.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008708.hg.1 |
ENST00000560961.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008708.hg.1 |
NM_004789.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008708.hg.1 |
NM_004789.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008708.hg.1 |
NM_004789.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008708.hg.1 |
NM_004789.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008708.hg.1 |
NM_004789.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008708.hg.1 |
NM_004789.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008708.hg.1 |
NM_004789.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008708.hg.1 |
NM_004789.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008708.hg.1 |
NM_004789.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008708.hg.1 |
NM_004789.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008708.hg.1 |
NM_004789.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09007271.hg.1 |
ENST00000373615.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09007271.hg.1 |
ENST00000488674.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09007271.hg.1 |
NM_004789.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09007271.hg.1 |
TC09000633.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0900158096.hg.1 |
ENST00000373615.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900158096.hg.1 |
ENST00000446480.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900158096.hg.1 |
NM_004789.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900158096.hg.1 |
TC0900008708.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900158096.hg.1 |
ENST00000373615.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900158096.hg.1 |
ENST00000446480.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900158096.hg.1 |
NM_004789.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900158097.hg.1 |
ENST00000373615.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900158097.hg.1 |
ENST00000446480.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900158097.hg.1 |
ENST00000488674.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900158097.hg.1 |
NM_004789.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900158097.hg.1 |
TC0900008708.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900158097.hg.1 |
ENST00000373615.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900158097.hg.1 |
ENST00000446480.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900158097.hg.1 |
ENST00000488674.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900158097.hg.1 |
NM_004789.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008708.hg.1 |
ENST00000373615.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008708.hg.1 |
ENST00000373615.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008708.hg.1 |
ENST00000373615.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008708.hg.1 |
ENST00000373615.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008708.hg.1 |
ENST00000373615.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008708.hg.1 |
ENST00000373615.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008708.hg.1 |
ENST00000373615.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008708.hg.1 |
ENST00000373615.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008708.hg.1 |
ENST00000373615.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008708.hg.1 |
ENST00000373615.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008708.hg.1 |
ENST00000373615.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008708.hg.1 |
ENST00000373615.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008708.hg.1 |
ENST00000446480.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008708.hg.1 |
ENST00000446480.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008708.hg.1 |
ENST00000446480.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008708.hg.1 |
ENST00000446480.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008708.hg.1 |
ENST00000446480.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008708.hg.1 |
ENST00000446480.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008708.hg.1 |
ENST00000446480.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008708.hg.1 |
ENST00000446480.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008708.hg.1 |
ENST00000488674.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008708.hg.1 |
ENST00000488674.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008708.hg.1 |
ENST00000488674.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008708.hg.1 |
ENST00000488674.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008708.hg.1 |
ENST00000488674.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008708.hg.1 |
ENST00000488674.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008708.hg.1 |
ENST00000488674.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008708.hg.1 |
ENST00000560961.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008708.hg.1 |
ENST00000560961.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008708.hg.1 |
ENST00000560961.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008708.hg.1 |
NM_004789.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008708.hg.1 |
NM_004789.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008708.hg.1 |
NM_004789.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008708.hg.1 |
NM_004789.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008708.hg.1 |
NM_004789.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008708.hg.1 |
NM_004789.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008708.hg.1 |
NM_004789.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008708.hg.1 |
NM_004789.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008708.hg.1 |
NM_004789.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008708.hg.1 |
NM_004789.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008708.hg.1 |
NM_004789.3 |
Refseq |