Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010022.hg.1 |
ENST00000371744.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010022.hg.1 |
ENST00000371744.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010022.hg.1 |
ENST00000371744.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010022.hg.1 |
ENST00000371744.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010022.hg.1 |
ENST00000371744.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010022.hg.1 |
ENST00000371744.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010022.hg.1 |
ENST00000371744.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010022.hg.1 |
ENST00000371744.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010022.hg.1 |
ENST00000371744.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010022.hg.1 |
ENST00000371744.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010022.hg.1 |
ENST00000371744.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010022.hg.1 |
ENST00000371752.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010022.hg.1 |
ENST00000371752.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010022.hg.1 |
ENST00000371752.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010022.hg.1 |
ENST00000371752.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010022.hg.1 |
ENST00000371752.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010022.hg.1 |
ENST00000371752.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010022.hg.1 |
ENST00000371752.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010022.hg.1 |
ENST00000371752.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010022.hg.1 |
ENST00000371752.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010022.hg.1 |
ENST00000371752.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010022.hg.1 |
ENST00000371752.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010022.hg.1 |
ENST00000371752.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010022.hg.1 |
ENST00000371752.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010022.hg.1 |
ENST00000371752.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010022.hg.1 |
ENST00000371752.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010022.hg.1 |
ENST00000371752.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010022.hg.1 |
ENST00000396105.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010022.hg.1 |
ENST00000396105.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010022.hg.1 |
ENST00000396105.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010022.hg.1 |
ENST00000396105.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010022.hg.1 |
ENST00000396105.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010022.hg.1 |
ENST00000396105.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010022.hg.1 |
ENST00000396105.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010022.hg.1 |
ENST00000396105.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010022.hg.1 |
ENST00000396105.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010022.hg.1 |
ENST00000396105.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010022.hg.1 |
ENST00000396105.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010022.hg.1 |
ENST00000396105.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010022.hg.1 |
ENST00000396105.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010022.hg.1 |
ENST00000396105.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010022.hg.1 |
ENST00000396105.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010022.hg.1 |
ENST00000396105.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010022.hg.1 |
ENST00000396105.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010022.hg.1 |
ENST00000455070.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010022.hg.1 |
ENST00000455070.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010022.hg.1 |
ENST00000455070.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010022.hg.1 |
ENST00000455070.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010022.hg.1 |
ENST00000455070.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010022.hg.1 |
ENST00000455070.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010022.hg.1 |
ENST00000455070.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010022.hg.1 |
NM_021035.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010022.hg.1 |
NM_021035.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010022.hg.1 |
NM_021035.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010022.hg.1 |
NM_021035.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010022.hg.1 |
NM_021035.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010022.hg.1 |
NM_021035.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010022.hg.1 |
NM_021035.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010022.hg.1 |
NM_021035.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010022.hg.1 |
NM_021035.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010022.hg.1 |
NM_021035.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010022.hg.1 |
NM_021035.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010022.hg.1 |
NM_021035.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010022.hg.1 |
NM_021035.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010022.hg.1 |
NM_021035.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010022.hg.1 |
NM_021035.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010022.hg.1 |
NM_021035.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010022.hg.1 |
NM_021035.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012724.hg.1 |
TC20000924.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012724.hg.1 |
ENST00000371752.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012724.hg.1 |
ENST00000371754.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012724.hg.1 |
ENST00000396105.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012724.hg.1 |
NM_021035.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000010022.hg.1 |
ENST00000371744.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010022.hg.1 |
ENST00000371744.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010022.hg.1 |
ENST00000371744.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010022.hg.1 |
ENST00000371744.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010022.hg.1 |
ENST00000371744.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010022.hg.1 |
ENST00000371744.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010022.hg.1 |
ENST00000371744.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010022.hg.1 |
ENST00000371744.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010022.hg.1 |
ENST00000371744.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010022.hg.1 |
ENST00000371744.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010022.hg.1 |
ENST00000371744.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010022.hg.1 |
ENST00000371752.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010022.hg.1 |
ENST00000371752.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010022.hg.1 |
ENST00000371752.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010022.hg.1 |
ENST00000371752.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010022.hg.1 |
ENST00000371752.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010022.hg.1 |
ENST00000371752.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010022.hg.1 |
ENST00000371752.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010022.hg.1 |
ENST00000371752.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010022.hg.1 |
ENST00000371752.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010022.hg.1 |
ENST00000371752.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010022.hg.1 |
ENST00000371752.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010022.hg.1 |
ENST00000371752.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010022.hg.1 |
ENST00000371752.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010022.hg.1 |
ENST00000371752.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010022.hg.1 |
ENST00000371752.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010022.hg.1 |
ENST00000371752.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010022.hg.1 |
ENST00000396105.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010022.hg.1 |
ENST00000396105.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010022.hg.1 |
ENST00000396105.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010022.hg.1 |
ENST00000396105.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010022.hg.1 |
ENST00000396105.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010022.hg.1 |
ENST00000396105.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010022.hg.1 |
ENST00000396105.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010022.hg.1 |
ENST00000396105.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010022.hg.1 |
ENST00000396105.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010022.hg.1 |
ENST00000396105.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010022.hg.1 |
ENST00000396105.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010022.hg.1 |
ENST00000396105.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010022.hg.1 |
ENST00000396105.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010022.hg.1 |
ENST00000396105.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010022.hg.1 |
ENST00000396105.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010022.hg.1 |
ENST00000396105.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010022.hg.1 |
ENST00000396105.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010022.hg.1 |
ENST00000455070.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010022.hg.1 |
ENST00000455070.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010022.hg.1 |
ENST00000455070.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010022.hg.1 |
ENST00000455070.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010022.hg.1 |
ENST00000455070.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010022.hg.1 |
ENST00000455070.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010022.hg.1 |
ENST00000455070.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010022.hg.1 |
NM_021035.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000010022.hg.1 |
NM_021035.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000010022.hg.1 |
NM_021035.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000010022.hg.1 |
NM_021035.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000010022.hg.1 |
NM_021035.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000010022.hg.1 |
NM_021035.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000010022.hg.1 |
NM_021035.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000010022.hg.1 |
NM_021035.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000010022.hg.1 |
NM_021035.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000010022.hg.1 |
NM_021035.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000010022.hg.1 |
NM_021035.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000010022.hg.1 |
NM_021035.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000010022.hg.1 |
NM_021035.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000010022.hg.1 |
NM_021035.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000010022.hg.1 |
NM_021035.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000010022.hg.1 |
NM_021035.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000010022.hg.1 |
NM_021035.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000171792.hg.1 |
ENST00000371752.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000171792.hg.1 |
ENST00000396105.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000171792.hg.1 |
NM_021035.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000171792.hg.1 |
TC2000010022.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000171792.hg.1 |
NM_021035.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000171792.hg.1 |
ENST00000371752.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000171792.hg.1 |
ENST00000396105.5 |
Ensembl |